Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133532041-133532545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2E1
Entrez Gene ID
1571
Gene Name
cytochrome P450 family 2 subfamily E member 1
Gene Aliases
CPE1, CYP2E, P450-J, P450C2E
UniGene
Hs.12907
Gene Chromosome Location
Chr.10: 133527363-133539116 on Build GRCh38
Transcripts
NM_000773.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5929181 c.499G>A p.D167N Chr.10: 133532135-133532135 on Build GRCh38
6639239 c.632G>A p.S211N Chr.10: 133532268-133532268 on Build GRCh38
1703289 c.638C>T p.P213L Chr.10: 133532274-133532274 on Build GRCh38
355828 c.532A>C p.N178H Chr.10: 133532168-133532168 on Build GRCh38
4502792 c.622C>T p.H208Y Chr.10: 133532258-133532258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566294180 Chr.10: 133532150-133532150 on Build GRCh38
rs746192147 Chr.10: 133532227-133532227 on Build GRCh38
rs945598259 Chr.10: 133532456-133532456 on Build GRCh38
rs747523205 Chr.10: 133532073-133532073 on Build GRCh38
rs907445349 Chr.10: 133532274-133532274 on Build GRCh38
rs55744609 Chr.10: 133532487-133532487 on Build GRCh38
rs750987941 Chr.10: 133532258-133532258 on Build GRCh38
rs568558058 Chr.10: 133532170-133532170 on Build GRCh38
rs749371010 Chr.10: 133532161-133532161 on Build GRCh38
rs6413419 Chr.10: 133532171-133532171 on Build GRCh38
rs546804720 Chr.10: 133532071-133532071 on Build GRCh38
rs748243360 Chr.10: 133532486-133532486 on Build GRCh38
rs761787153 Chr.10: 133532310-133532310 on Build GRCh38
rs139133362 Chr.10: 133532152-133532152 on Build GRCh38
rs1028393353 Chr.10: 133532287-133532287 on Build GRCh38
rs1008314928 Chr.10: 133532492-133532492 on Build GRCh38
rs768213976 Chr.10: 133532269-133532269 on Build GRCh38
rs8192773 Chr.10: 133532470-133532470 on Build GRCh38
rs992246169 Chr.10: 133532430-133532430 on Build GRCh38
rs767704036 Chr.10: 133532315-133532315 on Build GRCh38
rs60452492 Chr.10: 133532153-133532153 on Build GRCh38
rs569909432 Chr.10: 133532407-133532407 on Build GRCh38
rs1027603262 Chr.10: 133532406-133532406 on Build GRCh38
rs540678270 Chr.10: 133532476-133532476 on Build GRCh38
rs758535504 Chr.10: 133532147-133532147 on Build GRCh38
rs774671018 Chr.10: 133532301-133532302 on Build GRCh38
rs971765663 Chr.10: 133532084-133532084 on Build GRCh38
rs371462264 Chr.10: 133532330-133532330 on Build GRCh38
rs12254222 Chr.10: 133532494-133532494 on Build GRCh38
rs577159401 Chr.10: 133532273-133532273 on Build GRCh38
rs576234120 Chr.10: 133532400-133532400 on Build GRCh38
rs752959099 Chr.10: 133532204-133532204 on Build GRCh38
rs140500826 Chr.10: 133532192-133532192 on Build GRCh38
rs923179921 Chr.10: 133532157-133532157 on Build GRCh38
rs769163444 Chr.10: 133532170-133532177 on Build GRCh38
rs368615861 Chr.10: 133532266-133532266 on Build GRCh38
rs769469905 Chr.10: 133532094-133532094 on Build GRCh38
rs964826963 Chr.10: 133532388-133532388 on Build GRCh38
rs56040284 Chr.10: 133532159-133532159 on Build GRCh38
rs897642069 Chr.10: 133532499-133532499 on Build GRCh38
rs980425140 Chr.10: 133532420-133532420 on Build GRCh38
rs749696114 Chr.10: 133532403-133532403 on Build GRCh38
rs773265206 Chr.10: 133532281-133532281 on Build GRCh38
rs780401304 Chr.10: 133532261-133532261 on Build GRCh38
rs199581521 Chr.10: 133532305-133532305 on Build GRCh38
rs774993741 Chr.10: 133532263-133532263 on Build GRCh38
rs770796390 Chr.10: 133532298-133532298 on Build GRCh38
rs563829665 Chr.10: 133532489-133532489 on Build GRCh38
rs12257054 Chr.10: 133532514-133532514 on Build GRCh38
rs932615346 Chr.10: 133532467-133532467 on Build GRCh38
rs751624559 Chr.10: 133532233-133532233 on Build GRCh38
rs760756985 Chr.10: 133532297-133532297 on Build GRCh38
rs780885158 Chr.10: 133532122-133532122 on Build GRCh38
rs75795289 Chr.10: 133532134-133532134 on Build GRCh38
rs747578103 Chr.10: 133532272-133532272 on Build GRCh38
rs386749499 Chr.10: 133532513-133532514 on Build GRCh38
rs12254225 Chr.10: 133532513-133532513 on Build GRCh38
rs755208649 Chr.10: 133532142-133532142 on Build GRCh38
rs778445040 Chr.10: 133532270-133532270 on Build GRCh38
rs568483402 Chr.10: 133532426-133532426 on Build GRCh38
rs79549556 Chr.10: 133532179-133532179 on Build GRCh38
rs181745542 Chr.10: 133532379-133532379 on Build GRCh38
rs972385548 Chr.10: 133532397-133532397 on Build GRCh38
rs763652251 Chr.10: 133532102-133532102 on Build GRCh38
rs776876434 Chr.10: 133532089-133532089 on Build GRCh38
rs945620366 Chr.10: 133532230-133532230 on Build GRCh38
rs772559232 Chr.10: 133532260-133532260 on Build GRCh38
rs753014113 Chr.10: 133532143-133532143 on Build GRCh38
rs771264192 Chr.10: 133532078-133532078 on Build GRCh38
rs779352805 Chr.10: 133532466-133532466 on Build GRCh38
rs557994567 Chr.10: 133532162-133532162 on Build GRCh38
rs376604022 Chr.10: 133532331-133532331 on Build GRCh38
rs537515736 Chr.10: 133532491-133532491 on Build GRCh38
rs6413421 Chr.10: 133532307-133532307 on Build GRCh38
rs757401077 Chr.10: 133532234-133532234 on Build GRCh38
rs201427783 Chr.10: 133532176-133532176 on Build GRCh38
rs112507169 Chr.10: 133532512-133532512 on Build GRCh38
rs936344704 Chr.10: 133532477-133532477 on Build GRCh38
rs761982981 Chr.10: 133532307-133532307 on Build GRCh38
rs914052836 Chr.10: 133532441-133532441 on Build GRCh38
rs763198251 Chr.10: 133532299-133532299 on Build GRCh38
rs28371743 Chr.10: 133532248-133532248 on Build GRCh38
rs76138620 Chr.10: 133532180-133532180 on Build GRCh38
rs541830914 Chr.10: 133532401-133532401 on Build GRCh38
rs984838269 Chr.10: 133532361-133532361 on Build GRCh38
rs919286179 Chr.10: 133532105-133532105 on Build GRCh38
rs945296363 Chr.10: 133532449-133532449 on Build GRCh38
rs374523768 Chr.10: 133532093-133532093 on Build GRCh38
rs548262477 Chr.10: 133532160-133532160 on Build GRCh38
rs79131749 Chr.10: 133532285-133532285 on Build GRCh38
rs951786242 Chr.10: 133532412-133532412 on Build GRCh38
rs756169972 Chr.10: 133532158-133532158 on Build GRCh38
rs939109597 Chr.10: 133532428-133532428 on Build GRCh38
rs200871390 Chr.10: 133532193-133532193 on Build GRCh38
rs750206115 Chr.10: 133532141-133532141 on Build GRCh38
rs904129171 Chr.10: 133532460-133532460 on Build GRCh38
rs763286285 Chr.10: 133532211-133532211 on Build GRCh38
rs764189163 Chr.10: 133532218-133532218 on Build GRCh38
rs775164673 Chr.10: 133532097-133532097 on Build GRCh38
rs201167793 Chr.10: 133532129-133532129 on Build GRCh38
rs9919386 Chr.10: 133532099-133532099 on Build GRCh38
rs1044354688 Chr.10: 133532459-133532459 on Build GRCh38

View Full Product Details