Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 203920-204430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BET1L
Entrez Gene ID
51272
Gene Name
Bet1 golgi vesicular membrane trafficking protein like
Gene Aliases
BET1L1, GOLIM3, GS15, HSPC197
UniGene
Hs.414418
Gene Chromosome Location
Chr.11: 202924-207422 on Build GRCh38
Transcripts
NM_001098787.1 NM_016526.4

SNP Detail

SNP ID Chromosome Location
rs573254580 Chr.11: 204194-204194 on Build GRCh38
rs557677185 Chr.11: 204084-204084 on Build GRCh38
rs756837118 Chr.11: 203968-203968 on Build GRCh38
rs954844152 Chr.11: 203985-203985 on Build GRCh38
rs1017959480 Chr.11: 204237-204237 on Build GRCh38
rs866876636 Chr.11: 204048-204048 on Build GRCh38
rs544143830 Chr.11: 204306-204306 on Build GRCh38
rs924738457 Chr.11: 204341-204343 on Build GRCh38
rs1021738610 Chr.11: 203945-203945 on Build GRCh38
rs963413905 Chr.11: 204300-204300 on Build GRCh38
rs12282672 Chr.11: 204134-204134 on Build GRCh38
rs758999824 Chr.11: 204228-204230 on Build GRCh38
rs11551935 Chr.11: 204293-204293 on Build GRCh38
rs560989941 Chr.11: 204275-204275 on Build GRCh38
rs551920515 Chr.11: 204031-204031 on Build GRCh38
rs907899964 Chr.11: 204012-204012 on Build GRCh38
rs936299679 Chr.11: 204101-204101 on Build GRCh38
rs528692148 Chr.11: 204002-204002 on Build GRCh38
rs2280545 Chr.11: 204147-204147 on Build GRCh38
rs928484907 Chr.11: 204346-204346 on Build GRCh38
rs923554438 Chr.11: 203969-203969 on Build GRCh38
rs948734000 Chr.11: 204375-204375 on Build GRCh38
rs942080960 Chr.11: 204016-204016 on Build GRCh38
rs977577507 Chr.11: 204328-204328 on Build GRCh38
rs1037825906 Chr.11: 204030-204030 on Build GRCh38
rs948533845 Chr.11: 204325-204325 on Build GRCh38
rs1054690109 Chr.11: 204349-204349 on Build GRCh38
rs773498980 Chr.11: 204181-204181 on Build GRCh38
rs564707966 Chr.11: 204199-204199 on Build GRCh38
rs78309322 Chr.11: 204357-204357 on Build GRCh38
rs566518900 Chr.11: 203997-203997 on Build GRCh38
rs748629384 Chr.11: 203959-203960 on Build GRCh38
rs143145314 Chr.11: 204343-204343 on Build GRCh38
rs954966217 Chr.11: 204256-204256 on Build GRCh38
rs546428004 Chr.11: 203980-203980 on Build GRCh38
rs61876180 Chr.11: 204100-204100 on Build GRCh38
rs976182164 Chr.11: 204288-204288 on Build GRCh38
rs1003224912 Chr.11: 204087-204087 on Build GRCh38
rs560585594 Chr.11: 203955-203955 on Build GRCh38
rs934135535 Chr.11: 204079-204079 on Build GRCh38
rs948585914 Chr.11: 204037-204037 on Build GRCh38
rs922998450 Chr.11: 204298-204298 on Build GRCh38
rs768555886 Chr.11: 204152-204152 on Build GRCh38
rs1055124296 Chr.11: 204120-204120 on Build GRCh38
rs371621630 Chr.11: 204082-204082 on Build GRCh38
rs1045762014 Chr.11: 204172-204172 on Build GRCh38
rs947089079 Chr.11: 204394-204394 on Build GRCh38
rs2280544 Chr.11: 204062-204062 on Build GRCh38
rs891461562 Chr.11: 204368-204368 on Build GRCh38
rs964815041 Chr.11: 204165-204165 on Build GRCh38
rs771030115 Chr.11: 204324-204324 on Build GRCh38
rs745831935 Chr.11: 204070-204070 on Build GRCh38
rs748386703 Chr.11: 204135-204135 on Build GRCh38
rs964738033 Chr.11: 204008-204008 on Build GRCh38
rs540189277 Chr.11: 204399-204399 on Build GRCh38
rs910187211 Chr.11: 204007-204007 on Build GRCh38
rs111689552 Chr.11: 204208-204208 on Build GRCh38
rs772473719 Chr.11: 204373-204373 on Build GRCh38
rs1053003969 Chr.11: 204363-204363 on Build GRCh38
rs575081562 Chr.11: 204238-204238 on Build GRCh38
rs760087710 Chr.11: 204369-204369 on Build GRCh38
rs1045454 Chr.11: 204228-204228 on Build GRCh38
rs984138397 Chr.11: 204267-204267 on Build GRCh38
rs891494590 Chr.11: 204123-204123 on Build GRCh38
rs753217066 Chr.11: 203954-203954 on Build GRCh38
rs61996345 Chr.11: 203965-203965 on Build GRCh38
rs1006100310 Chr.11: 204236-204236 on Build GRCh38

View Full Product Details