Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 555083-555611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMNTD2
Entrez Gene ID
256329
Gene Name
lamin tail domain containing 2
Gene Aliases
C11orf35
UniGene
Hs.669395
Gene Chromosome Location
Chr.11: 554850-560780 on Build GRCh38
Transcripts
NM_173573.2 XM_011519965.2 XM_011519967.2 XM_017017476.1 XM_017017477.1 XM_017017478.1 XM_017017479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3415951 c.1761A>G p.E587E Chr.11: 555317-555317 on Build GRCh38
1289566 c.1699A>C p.T567P Chr.11: 555379-555379 on Build GRCh38
6432074 c.1723G>A p.A575T Chr.11: 555355-555355 on Build GRCh38
7175768 c.1792G>A p.D598N Chr.11: 555093-555093 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764944318 Chr.11: 555345-555345 on Build GRCh38
rs763414061 Chr.11: 555536-555536 on Build GRCh38
rs58187194 Chr.11: 555134-555134 on Build GRCh38
rs202207219 Chr.11: 555376-555376 on Build GRCh38
rs377072542 Chr.11: 555354-555354 on Build GRCh38
rs746470566 Chr.11: 555324-555324 on Build GRCh38
rs759668783 Chr.11: 555517-555517 on Build GRCh38
rs746587921 Chr.11: 555381-555381 on Build GRCh38
rs780390220 Chr.11: 555310-555310 on Build GRCh38
rs543736884 Chr.11: 555122-555122 on Build GRCh38
rs760816828 Chr.11: 555417-555417 on Build GRCh38
rs779373767 Chr.11: 555367-555367 on Build GRCh38
rs776974660 Chr.11: 555416-555416 on Build GRCh38
rs530471704 Chr.11: 555306-555306 on Build GRCh38
rs766315280 Chr.11: 555419-555419 on Build GRCh38
rs778778910 Chr.11: 555277-555277 on Build GRCh38
rs140287963 Chr.11: 555577-555577 on Build GRCh38
rs375981150 Chr.11: 555290-555290 on Build GRCh38
rs370993619 Chr.11: 555401-555401 on Build GRCh38
rs934783071 Chr.11: 555569-555569 on Build GRCh38
rs867124318 Chr.11: 555210-555210 on Build GRCh38
rs916181509 Chr.11: 555501-555501 on Build GRCh38
rs755394395 Chr.11: 555312-555312 on Build GRCh38
rs766120708 Chr.11: 555274-555274 on Build GRCh38
rs951145759 Chr.11: 555466-555466 on Build GRCh38
rs747652093 Chr.11: 555379-555380 on Build GRCh38
rs535651529 Chr.11: 555399-555399 on Build GRCh38
rs756337165 Chr.11: 555333-555333 on Build GRCh38
rs1014436967 Chr.11: 555339-555339 on Build GRCh38
rs781712695 Chr.11: 555137-555137 on Build GRCh38
rs760082547 Chr.11: 555427-555427 on Build GRCh38
rs929590630 Chr.11: 555288-555288 on Build GRCh38
rs35167891 Chr.11: 555508-555508 on Build GRCh38
rs779940792 Chr.11: 555481-555481 on Build GRCh38
rs764103795 Chr.11: 555146-555170 on Build GRCh38
rs934435826 Chr.11: 555503-555503 on Build GRCh38
rs892823596 Chr.11: 555526-555528 on Build GRCh38
rs967321499 Chr.11: 555246-555246 on Build GRCh38
rs771077970 Chr.11: 555499-555499 on Build GRCh38
rs976618221 Chr.11: 555445-555445 on Build GRCh38
rs758823025 Chr.11: 555447-555447 on Build GRCh38
rs963818026 Chr.11: 555275-555275 on Build GRCh38
rs983153438 Chr.11: 555467-555467 on Build GRCh38
rs770188983 Chr.11: 555111-555111 on Build GRCh38
rs768391075 Chr.11: 555154-555154 on Build GRCh38
rs749707473 Chr.11: 555311-555311 on Build GRCh38
rs528138936 Chr.11: 555220-555220 on Build GRCh38
rs746420167 Chr.11: 555528-555529 on Build GRCh38
rs749136208 Chr.11: 555484-555484 on Build GRCh38
rs751245029 Chr.11: 555540-555540 on Build GRCh38
rs950222920 Chr.11: 555507-555507 on Build GRCh38
rs780862830 Chr.11: 555549-555549 on Build GRCh38
rs952649353 Chr.11: 555155-555155 on Build GRCh38
rs757393634 Chr.11: 555157-555161 on Build GRCh38
rs773298033 Chr.11: 555263-555263 on Build GRCh38
rs771564850 Chr.11: 555110-555111 on Build GRCh38
rs777548733 Chr.11: 555454-555454 on Build GRCh38
rs1030835143 Chr.11: 555249-555249 on Build GRCh38
rs774567649 Chr.11: 555347-555347 on Build GRCh38
rs777338850 Chr.11: 555113-555113 on Build GRCh38
rs760393767 Chr.11: 555119-555119 on Build GRCh38
rs867955738 Chr.11: 555177-555177 on Build GRCh38
rs770065598 Chr.11: 555141-555141 on Build GRCh38
rs903266486 Chr.11: 555559-555559 on Build GRCh38
rs767612828 Chr.11: 555352-555352 on Build GRCh38
rs776635035 Chr.11: 555389-555389 on Build GRCh38
rs755410710 Chr.11: 555364-555364 on Build GRCh38
rs772810284 Chr.11: 555323-555323 on Build GRCh38
rs923126950 Chr.11: 555329-555329 on Build GRCh38
rs201776188 Chr.11: 555224-555224 on Build GRCh38
rs559097180 Chr.11: 555135-555135 on Build GRCh38
rs867176854 Chr.11: 555172-555172 on Build GRCh38
rs780413295 Chr.11: 555146-555146 on Build GRCh38
rs750385179 Chr.11: 555117-555117 on Build GRCh38
rs759800288 Chr.11: 555113-555114 on Build GRCh38
rs893112186 Chr.11: 555585-555585 on Build GRCh38
rs759440828 Chr.11: 555337-555337 on Build GRCh38
rs375461388 Chr.11: 555444-555444 on Build GRCh38
rs1007579662 Chr.11: 555587-555587 on Build GRCh38
rs755831562 Chr.11: 555157-555158 on Build GRCh38
rs200292905 Chr.11: 555130-555130 on Build GRCh38
rs746304507 Chr.11: 555139-555139 on Build GRCh38
rs758606853 Chr.11: 555125-555125 on Build GRCh38
rs756306236 Chr.11: 555523-555523 on Build GRCh38
rs761485065 Chr.11: 555259-555259 on Build GRCh38
rs578072346 Chr.11: 555115-555115 on Build GRCh38
rs773501230 Chr.11: 555491-555491 on Build GRCh38
rs368266893 Chr.11: 555472-555472 on Build GRCh38
rs373015093 Chr.11: 555285-555285 on Build GRCh38
rs756803723 Chr.11: 555470-555470 on Build GRCh38
rs185023785 Chr.11: 555170-555170 on Build GRCh38
rs748839518 Chr.11: 555158-555159 on Build GRCh38
rs773290029 Chr.11: 555410-555410 on Build GRCh38
rs745588289 Chr.11: 555182-555183 on Build GRCh38
rs369845778 Chr.11: 555114-555114 on Build GRCh38
rs751106521 Chr.11: 555464-555464 on Build GRCh38
rs750422378 Chr.11: 555157-555166 on Build GRCh38
rs1002404372 Chr.11: 555564-555564 on Build GRCh38
rs563639204 Chr.11: 555124-555124 on Build GRCh38
rs763260175 Chr.11: 555146-555185 on Build GRCh38
rs74959245 Chr.11: 555547-555547 on Build GRCh38
rs867632543 Chr.11: 555236-555236 on Build GRCh38
rs769597750 Chr.11: 555397-555397 on Build GRCh38
rs771644690 Chr.11: 555262-555262 on Build GRCh38
rs753708200 Chr.11: 555278-555278 on Build GRCh38
rs941292837 Chr.11: 555320-555320 on Build GRCh38
rs777270650 Chr.11: 555378-555378 on Build GRCh38
rs754686413 Chr.11: 555159-555178 on Build GRCh38
rs767427197 Chr.11: 555157-555171 on Build GRCh38
rs761972005 Chr.11: 555350-555350 on Build GRCh38
rs570865581 Chr.11: 555302-555302 on Build GRCh38
rs9704177 Chr.11: 555182-555182 on Build GRCh38
rs769760756 Chr.11: 555479-555479 on Build GRCh38
rs765676722 Chr.11: 555118-555119 on Build GRCh38
rs764306537 Chr.11: 555538-555538 on Build GRCh38
rs770343572 Chr.11: 555386-555386 on Build GRCh38
rs759230499 Chr.11: 555281-555281 on Build GRCh38
rs143504198 Chr.11: 555317-555317 on Build GRCh38
rs1035201015 Chr.11: 555568-555568 on Build GRCh38
rs775536215 Chr.11: 555138-555139 on Build GRCh38
rs756120788 Chr.11: 555355-555355 on Build GRCh38
rs1029260643 Chr.11: 555105-555105 on Build GRCh38
rs745732985 Chr.11: 555390-555390 on Build GRCh38
rs770951133 Chr.11: 555330-555330 on Build GRCh38
rs748921018 Chr.11: 555147-555147 on Build GRCh38
rs781648173 Chr.11: 555102-555102 on Build GRCh38
rs984221101 Chr.11: 555156-555156 on Build GRCh38
rs746288086 Chr.11: 555104-555104 on Build GRCh38
rs1022480434 Chr.11: 555142-555142 on Build GRCh38
rs768273111 Chr.11: 555490-555490 on Build GRCh38
rs757332291 Chr.11: 555295-555295 on Build GRCh38
rs911279319 Chr.11: 555165-555165 on Build GRCh38
rs745834493 Chr.11: 555476-555476 on Build GRCh38
rs890779103 Chr.11: 555369-555369 on Build GRCh38
rs753253168 Chr.11: 555525-555525 on Build GRCh38
rs559302850 Chr.11: 555208-555208 on Build GRCh38
rs776616259 Chr.11: 555512-555512 on Build GRCh38
rs998776610 Chr.11: 555362-555362 on Build GRCh38
rs775221939 Chr.11: 555398-555398 on Build GRCh38
rs754393160 Chr.11: 555425-555425 on Build GRCh38
rs138037086 Chr.11: 555298-555298 on Build GRCh38
rs370145696 Chr.11: 555360-555360 on Build GRCh38
rs771759449 Chr.11: 555414-555415 on Build GRCh38
rs778715756 Chr.11: 555313-555313 on Build GRCh38
rs55731724 Chr.11: 555224-555224 on Build GRCh38
rs1022449395 Chr.11: 555438-555438 on Build GRCh38
rs528952627 Chr.11: 555231-555232 on Build GRCh38
rs1002278256 Chr.11: 555453-555453 on Build GRCh38
rs957955029 Chr.11: 555254-555254 on Build GRCh38
rs751686846 Chr.11: 555132-555132 on Build GRCh38
rs372552187 Chr.11: 555498-555498 on Build GRCh38
rs375596832 Chr.11: 555371-555371 on Build GRCh38
rs998830261 Chr.11: 555248-555248 on Build GRCh38
rs751643562 Chr.11: 555151-555170 on Build GRCh38
rs774137497 Chr.11: 555257-555257 on Build GRCh38
rs541559521 Chr.11: 555113-555113 on Build GRCh38
rs765811023 Chr.11: 555446-555446 on Build GRCh38
rs760615118 Chr.11: 555269-555269 on Build GRCh38

View Full Product Details