Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 557197-557724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMNTD2
Entrez Gene ID
256329
Gene Name
lamin tail domain containing 2
Gene Aliases
C11orf35
UniGene
Hs.669395
Gene Chromosome Location
Chr.11: 554850-560780 on Build GRCh38
Transcripts
NM_173573.2 XM_011519965.2 XM_011519967.2 XM_017017476.1 XM_017017477.1 XM_017017478.1 XM_017017479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4647350 c.616A>C p.T206P Chr.11: 557580-557580 on Build GRCh38
5372739 c.565G>C p.A189P Chr.11: 557631-557631 on Build GRCh38
3666388 c.637G>C p.E213Q Chr.11: 557475-557475 on Build GRCh38
5425958 c.612C>T p.A204A Chr.11: 557584-557584 on Build GRCh38
159459 c.576G>A p.L192L Chr.11: 557620-557620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760458910 Chr.11: 557599-557600 on Build GRCh38
rs751786475 Chr.11: 557618-557618 on Build GRCh38
rs1006217163 Chr.11: 557538-557538 on Build GRCh38
rs773922907 Chr.11: 557591-557591 on Build GRCh38
rs761325985 Chr.11: 557593-557593 on Build GRCh38
rs781576587 Chr.11: 557579-557579 on Build GRCh38
rs150372027 Chr.11: 557632-557632 on Build GRCh38
rs202084192 Chr.11: 557665-557665 on Build GRCh38
rs530319559 Chr.11: 557491-557491 on Build GRCh38
rs764804747 Chr.11: 557484-557484 on Build GRCh38
rs752511586 Chr.11: 557671-557671 on Build GRCh38
rs561554716 Chr.11: 557235-557235 on Build GRCh38
rs377636681 Chr.11: 557528-557528 on Build GRCh38
rs148920876 Chr.11: 557469-557469 on Build GRCh38
rs750580837 Chr.11: 557682-557682 on Build GRCh38
rs1003490315 Chr.11: 557302-557302 on Build GRCh38
rs755870757 Chr.11: 557502-557502 on Build GRCh38
rs138095963 Chr.11: 557633-557633 on Build GRCh38
rs553954486 Chr.11: 557344-557344 on Build GRCh38
rs756077393 Chr.11: 557688-557688 on Build GRCh38
rs779855574 Chr.11: 557355-557355 on Build GRCh38
rs750316389 Chr.11: 557551-557551 on Build GRCh38
rs936466 Chr.11: 557342-557342 on Build GRCh38
rs771634359 Chr.11: 557510-557510 on Build GRCh38
rs778137078 Chr.11: 557573-557573 on Build GRCh38
rs945505955 Chr.11: 557701-557701 on Build GRCh38
rs568252351 Chr.11: 557308-557308 on Build GRCh38
rs921719868 Chr.11: 557221-557221 on Build GRCh38
rs1013407773 Chr.11: 557251-557251 on Build GRCh38
rs1010137523 Chr.11: 557316-557316 on Build GRCh38
rs561284051 Chr.11: 557487-557487 on Build GRCh38
rs775714829 Chr.11: 557411-557411 on Build GRCh38
rs747073649 Chr.11: 557509-557509 on Build GRCh38
rs768359717 Chr.11: 557585-557585 on Build GRCh38
rs895719253 Chr.11: 557314-557314 on Build GRCh38
rs574684726 Chr.11: 557676-557676 on Build GRCh38
rs758206846 Chr.11: 557253-557253 on Build GRCh38
rs754384425 Chr.11: 557360-557360 on Build GRCh38
rs146558617 Chr.11: 557577-557577 on Build GRCh38
rs746903237 Chr.11: 557454-557454 on Build GRCh38
rs775969063 Chr.11: 557583-557583 on Build GRCh38
rs374265452 Chr.11: 557503-557503 on Build GRCh38
rs552037187 Chr.11: 557279-557279 on Build GRCh38
rs773788316 Chr.11: 557435-557435 on Build GRCh38
rs1046323602 Chr.11: 557438-557438 on Build GRCh38
rs1017837563 Chr.11: 557281-557281 on Build GRCh38
rs138662694 Chr.11: 557613-557613 on Build GRCh38
rs750748414 Chr.11: 557539-557539 on Build GRCh38
rs761305513 Chr.11: 557541-557541 on Build GRCh38
rs530104427 Chr.11: 557236-557236 on Build GRCh38
rs751034366 Chr.11: 557490-557490 on Build GRCh38
rs774522907 Chr.11: 557399-557399 on Build GRCh38
rs912147396 Chr.11: 557265-557265 on Build GRCh38
rs749469150 Chr.11: 557414-557414 on Build GRCh38
rs532417751 Chr.11: 557273-557273 on Build GRCh38
rs11825368 Chr.11: 557540-557540 on Build GRCh38
rs781002402 Chr.11: 557458-557458 on Build GRCh38
rs115430352 Chr.11: 557617-557617 on Build GRCh38
rs746348764 Chr.11: 557580-557580 on Build GRCh38
rs1020212317 Chr.11: 557329-557329 on Build GRCh38
rs139930536 Chr.11: 557433-557433 on Build GRCh38
rs768889768 Chr.11: 557416-557416 on Build GRCh38
rs757072911 Chr.11: 557455-557455 on Build GRCh38
rs372445124 Chr.11: 557359-557359 on Build GRCh38
rs757863407 Chr.11: 557450-557450 on Build GRCh38
rs762051630 Chr.11: 557573-557574 on Build GRCh38
rs76495701 Chr.11: 557660-557660 on Build GRCh38
rs770149970 Chr.11: 557526-557526 on Build GRCh38
rs576897294 Chr.11: 557364-557364 on Build GRCh38
rs913978541 Chr.11: 557700-557700 on Build GRCh38
rs552611047 Chr.11: 557274-557274 on Build GRCh38
rs1017555566 Chr.11: 557464-557464 on Build GRCh38
rs968930845 Chr.11: 557330-557330 on Build GRCh38
rs538109219 Chr.11: 557670-557670 on Build GRCh38
rs764909249 Chr.11: 557530-557530 on Build GRCh38
rs368263411 Chr.11: 557636-557636 on Build GRCh38
rs141302254 Chr.11: 557578-557578 on Build GRCh38
rs773115550 Chr.11: 557218-557218 on Build GRCh38
rs535003414 Chr.11: 557393-557393 on Build GRCh38
rs777687029 Chr.11: 557377-557377 on Build GRCh38
rs759941043 Chr.11: 557483-557483 on Build GRCh38
rs373453237 Chr.11: 557470-557470 on Build GRCh38
rs903394423 Chr.11: 557220-557220 on Build GRCh38
rs894659382 Chr.11: 557250-557250 on Build GRCh38
rs566483760 Chr.11: 557266-557266 on Build GRCh38
rs760705799 Chr.11: 557609-557609 on Build GRCh38
rs145884999 Chr.11: 557445-557445 on Build GRCh38
rs1054734386 Chr.11: 557394-557394 on Build GRCh38
rs183355273 Chr.11: 557306-557306 on Build GRCh38
rs747757050 Chr.11: 557652-557652 on Build GRCh38
rs759835653 Chr.11: 557525-557525 on Build GRCh38
rs371678978 Chr.11: 557645-557645 on Build GRCh38
rs987118244 Chr.11: 557368-557368 on Build GRCh38
rs142820502 Chr.11: 557477-557477 on Build GRCh38
rs767445647 Chr.11: 557544-557544 on Build GRCh38
rs776653940 Chr.11: 557662-557662 on Build GRCh38
rs569083434 Chr.11: 557280-557280 on Build GRCh38
rs59604070 Chr.11: 557234-557234 on Build GRCh38
rs565033812 Chr.11: 557686-557686 on Build GRCh38
rs78974264 Chr.11: 557292-557292 on Build GRCh38
rs61877803 Chr.11: 557252-557252 on Build GRCh38
rs200625515 Chr.11: 557472-557472 on Build GRCh38
rs772308841 Chr.11: 557505-557505 on Build GRCh38
rs555291385 Chr.11: 557675-557675 on Build GRCh38
rs1054597795 Chr.11: 557247-557247 on Build GRCh38
rs757739525 Chr.11: 557387-557387 on Build GRCh38
rs763558598 Chr.11: 557489-557489 on Build GRCh38
rs774340667 Chr.11: 557420-557420 on Build GRCh38
rs771590874 Chr.11: 557655-557655 on Build GRCh38
rs773351908 Chr.11: 557586-557587 on Build GRCh38
rs191916495 Chr.11: 557656-557656 on Build GRCh38
rs772086986 Chr.11: 557581-557581 on Build GRCh38
rs369889704 Chr.11: 557494-557494 on Build GRCh38
rs766214947 Chr.11: 557610-557610 on Build GRCh38
rs763711586 Chr.11: 557537-557537 on Build GRCh38
rs750357468 Chr.11: 557608-557608 on Build GRCh38
rs745712730 Chr.11: 557501-557501 on Build GRCh38
rs997319400 Chr.11: 557468-557468 on Build GRCh38
rs554990782 Chr.11: 557448-557448 on Build GRCh38
rs746815228 Chr.11: 557384-557384 on Build GRCh38
rs778051009 Chr.11: 557507-557507 on Build GRCh38
rs570289556 Chr.11: 557228-557228 on Build GRCh38
rs753481773 Chr.11: 557356-557356 on Build GRCh38
rs199973483 Chr.11: 557641-557641 on Build GRCh38
rs4963199 Chr.11: 557661-557661 on Build GRCh38
rs766330650 Chr.11: 557671-557671 on Build GRCh38
rs149449830 Chr.11: 557640-557640 on Build GRCh38
rs117790851 Chr.11: 557294-557294 on Build GRCh38
rs767618400 Chr.11: 557677-557677 on Build GRCh38
rs940947935 Chr.11: 557272-557272 on Build GRCh38
rs11822657 Chr.11: 557584-557584 on Build GRCh38
rs761133499 Chr.11: 557437-557437 on Build GRCh38
rs200143254 Chr.11: 557553-557553 on Build GRCh38
rs757871273 Chr.11: 557488-557488 on Build GRCh38
rs992292245 Chr.11: 557687-557687 on Build GRCh38
rs745649725 Chr.11: 557461-557461 on Build GRCh38
rs774014340 Chr.11: 557647-557647 on Build GRCh38
rs754636615 Chr.11: 557447-557447 on Build GRCh38
rs777193780 Chr.11: 557481-557481 on Build GRCh38
rs369030115 Chr.11: 557465-557465 on Build GRCh38
rs559184519 Chr.11: 557434-557434 on Build GRCh38
rs777703405 Chr.11: 557451-557451 on Build GRCh38
rs770172312 Chr.11: 557581-557581 on Build GRCh38
rs61329792 Chr.11: 557444-557444 on Build GRCh38
rs964603639 Chr.11: 557288-557288 on Build GRCh38
rs201477233 Chr.11: 557392-557392 on Build GRCh38
rs201277609 Chr.11: 557607-557607 on Build GRCh38
rs758753270 Chr.11: 557571-557571 on Build GRCh38
rs545223557 Chr.11: 557521-557521 on Build GRCh38
rs929375389 Chr.11: 557512-557512 on Build GRCh38
rs761305378 Chr.11: 557474-557474 on Build GRCh38
rs930845349 Chr.11: 557275-557275 on Build GRCh38

View Full Product Details