Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 710331-710825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPS8L2
Entrez Gene ID
64787
Gene Name
EPS8 like 2
Gene Aliases
EPS8R2
UniGene
Hs.55016
Gene Chromosome Location
Chr.11: 705217-727727 on Build GRCh38
Transcripts
NM_022772.3 XM_017018130.1 XM_017018131.1 XM_017018132.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3359378 c.134T>C p.I45T Chr.11: 710455-710455 on Build GRCh38
5635026 c.129C>G p.N43K Chr.11: 710450-710450 on Build GRCh38
6243582 c.108G>A p.R36R Chr.11: 710429-710429 on Build GRCh38
1605174 c.152A>C p.Q51P Chr.11: 710473-710473 on Build GRCh38
5012262 c.149C>A p.S50* Chr.11: 710470-710470 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200967975 Chr.11: 710462-710462 on Build GRCh38
rs560613629 Chr.11: 710627-710627 on Build GRCh38
rs779468624 Chr.11: 710464-710464 on Build GRCh38
rs780426922 Chr.11: 710393-710393 on Build GRCh38
rs767635733 Chr.11: 710377-710377 on Build GRCh38
rs7395822 Chr.11: 710558-710558 on Build GRCh38
rs775984254 Chr.11: 710493-710493 on Build GRCh38
rs191132967 Chr.11: 710684-710684 on Build GRCh38
rs551232786 Chr.11: 710361-710361 on Build GRCh38
rs922891230 Chr.11: 710479-710479 on Build GRCh38
rs1010681922 Chr.11: 710607-710607 on Build GRCh38
rs773326764 Chr.11: 710515-710515 on Build GRCh38
rs1052330633 Chr.11: 710752-710752 on Build GRCh38
rs1052285784 Chr.11: 710590-710590 on Build GRCh38
rs145656059 Chr.11: 710635-710635 on Build GRCh38
rs1033064926 Chr.11: 710360-710360 on Build GRCh38
rs1039533947 Chr.11: 710787-710787 on Build GRCh38
rs983430913 Chr.11: 710658-710658 on Build GRCh38
rs1013367300 Chr.11: 710587-710587 on Build GRCh38
rs750492293 Chr.11: 710380-710380 on Build GRCh38
rs971962773 Chr.11: 710614-710614 on Build GRCh38
rs753096193 Chr.11: 710474-710474 on Build GRCh38
rs796905301 Chr.11: 710637-710637 on Build GRCh38
rs777481965 Chr.11: 710419-710419 on Build GRCh38
rs769618001 Chr.11: 710440-710440 on Build GRCh38
rs539628025 Chr.11: 710550-710550 on Build GRCh38
rs371399082 Chr.11: 710431-710431 on Build GRCh38
rs950155850 Chr.11: 710723-710723 on Build GRCh38
rs540969909 Chr.11: 710610-710610 on Build GRCh38
rs760681573 Chr.11: 710455-710455 on Build GRCh38
rs35555625 Chr.11: 710767-710767 on Build GRCh38
rs751811382 Chr.11: 710373-710373 on Build GRCh38
rs199874167 Chr.11: 710503-710503 on Build GRCh38
rs562666163 Chr.11: 710680-710680 on Build GRCh38
rs756713353 Chr.11: 710389-710389 on Build GRCh38
rs894589509 Chr.11: 710778-710778 on Build GRCh38
rs376531230 Chr.11: 710542-710542 on Build GRCh38
rs1042005707 Chr.11: 710744-710744 on Build GRCh38
rs766430379 Chr.11: 710460-710460 on Build GRCh38
rs761996508 Chr.11: 710375-710375 on Build GRCh38
rs936179997 Chr.11: 710755-710755 on Build GRCh38
rs570696750 Chr.11: 710539-710539 on Build GRCh38
rs765732672 Chr.11: 710452-710454 on Build GRCh38
rs376112350 Chr.11: 710496-710496 on Build GRCh38
rs754310926 Chr.11: 710394-710394 on Build GRCh38
rs555303803 Chr.11: 710568-710568 on Build GRCh38
rs931652936 Chr.11: 710760-710775 on Build GRCh38
rs991562970 Chr.11: 710700-710700 on Build GRCh38
rs377044318 Chr.11: 710414-710414 on Build GRCh38
rs531660875 Chr.11: 710681-710681 on Build GRCh38
rs777583989 Chr.11: 710477-710477 on Build GRCh38
rs1010650859 Chr.11: 710395-710395 on Build GRCh38
rs767725231 Chr.11: 710446-710446 on Build GRCh38
rs779375860 Chr.11: 710400-710400 on Build GRCh38
rs772042929 Chr.11: 710685-710685 on Build GRCh38
rs943127402 Chr.11: 710711-710711 on Build GRCh38
rs376203143 Chr.11: 710495-710495 on Build GRCh38
rs758189540 Chr.11: 710475-710475 on Build GRCh38
rs762744153 Chr.11: 710511-710511 on Build GRCh38
rs781464231 Chr.11: 710447-710447 on Build GRCh38
rs919093439 Chr.11: 710471-710471 on Build GRCh38
rs746624745 Chr.11: 710480-710480 on Build GRCh38
rs7396652 Chr.11: 710567-710567 on Build GRCh38
rs759479219 Chr.11: 710435-710435 on Build GRCh38
rs893638616 Chr.11: 710600-710600 on Build GRCh38
rs760908504 Chr.11: 710520-710520 on Build GRCh38
rs7396314 Chr.11: 710724-710724 on Build GRCh38
rs747842098 Chr.11: 710405-710405 on Build GRCh38
rs576177004 Chr.11: 710562-710562 on Build GRCh38
rs976194984 Chr.11: 710722-710722 on Build GRCh38
rs370812183 Chr.11: 710687-710687 on Build GRCh38
rs201710721 Chr.11: 710730-710731 on Build GRCh38
rs377007390 Chr.11: 710763-710770 on Build GRCh38
rs899585328 Chr.11: 710790-710790 on Build GRCh38
rs998309732 Chr.11: 710794-710794 on Build GRCh38
rs903412385 Chr.11: 710745-710748 on Build GRCh38
rs1015744955 Chr.11: 710466-710466 on Build GRCh38
rs770418057 Chr.11: 710429-710429 on Build GRCh38
rs935299612 Chr.11: 710746-710746 on Build GRCh38
rs750589182 Chr.11: 710453-710453 on Build GRCh38
rs143848078 Chr.11: 710484-710484 on Build GRCh38
rs139916138 Chr.11: 710742-710749 on Build GRCh38
rs386749608 Chr.11: 710763-710767 on Build GRCh38
rs935207740 Chr.11: 710585-710585 on Build GRCh38
rs776750117 Chr.11: 710526-710526 on Build GRCh38
rs766405129 Chr.11: 710521-710521 on Build GRCh38
rs138277885 Chr.11: 710444-710444 on Build GRCh38
rs545969515 Chr.11: 710649-710649 on Build GRCh38
rs763461101 Chr.11: 710495-710511 on Build GRCh38
rs755316478 Chr.11: 710396-710396 on Build GRCh38
rs951691684 Chr.11: 710483-710483 on Build GRCh38
rs377097002 Chr.11: 710519-710519 on Build GRCh38
rs910320369 Chr.11: 710699-710699 on Build GRCh38
rs768380012 Chr.11: 710513-710513 on Build GRCh38
rs775334536 Chr.11: 710506-710506 on Build GRCh38
rs941483903 Chr.11: 710510-710510 on Build GRCh38
rs374333852 Chr.11: 710463-710463 on Build GRCh38
rs35626095 Chr.11: 710763-710763 on Build GRCh38
rs746494874 Chr.11: 710425-710425 on Build GRCh38
rs575036700 Chr.11: 710593-710593 on Build GRCh38
rs562157688 Chr.11: 710688-710688 on Build GRCh38
rs760094235 Chr.11: 710401-710402 on Build GRCh38
rs951670466 Chr.11: 710654-710654 on Build GRCh38
rs916010802 Chr.11: 710705-710705 on Build GRCh38
rs547702297 Chr.11: 710741-710741 on Build GRCh38
rs775248199 Chr.11: 710441-710441 on Build GRCh38

View Full Product Details