Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 720390-720917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPS8L2
Entrez Gene ID
64787
Gene Name
EPS8 like 2
Gene Aliases
EPS8R2
UniGene
Hs.55016
Gene Chromosome Location
Chr.11: 705217-727727 on Build GRCh38
Transcripts
NM_022772.3 XM_017018130.1 XM_017018131.1 XM_017018132.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3703675 c.361C>T p.Q121* Chr.11: 720630-720630 on Build GRCh38
6497221 c.542G>A p.R181Q Chr.11: 720894-720894 on Build GRCh38
5480329 c.519C>A p.D173E Chr.11: 720871-720871 on Build GRCh38
6995177 c.328-4_335delGCAGGAGGAGCT p.? Chr.11: 720593-720604 on Build GRCh38
5984826 c.385C>T p.Q129* Chr.11: 720654-720654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201129457 Chr.11: 720609-720609 on Build GRCh38
rs766015412 Chr.11: 720538-720538 on Build GRCh38
rs754127157 Chr.11: 720703-720703 on Build GRCh38
rs6597989 Chr.11: 720763-720763 on Build GRCh38
rs1022643706 Chr.11: 720631-720631 on Build GRCh38
rs772551632 Chr.11: 720635-720635 on Build GRCh38
rs539592756 Chr.11: 720868-720868 on Build GRCh38
rs765381407 Chr.11: 720866-720866 on Build GRCh38
rs773632598 Chr.11: 720648-720648 on Build GRCh38
rs1037569908 Chr.11: 720507-720507 on Build GRCh38
rs1031619757 Chr.11: 720470-720470 on Build GRCh38
rs764853873 Chr.11: 720676-720676 on Build GRCh38
rs372636952 Chr.11: 720713-720713 on Build GRCh38
rs780440827 Chr.11: 720599-720599 on Build GRCh38
rs779909774 Chr.11: 720881-720882 on Build GRCh38
rs1039829888 Chr.11: 720782-720782 on Build GRCh38
rs568385251 Chr.11: 720689-720689 on Build GRCh38
rs968838132 Chr.11: 720645-720645 on Build GRCh38
rs777769940 Chr.11: 720568-720568 on Build GRCh38
rs527505904 Chr.11: 720416-720416 on Build GRCh38
rs778824922 Chr.11: 720625-720625 on Build GRCh38
rs1010990097 Chr.11: 720503-720503 on Build GRCh38
rs559611623 Chr.11: 720428-720428 on Build GRCh38
rs200177008 Chr.11: 720867-720867 on Build GRCh38
rs899967190 Chr.11: 720795-720795 on Build GRCh38
rs188385344 Chr.11: 720505-720505 on Build GRCh38
rs1000302540 Chr.11: 720827-720827 on Build GRCh38
rs909888133 Chr.11: 720744-720744 on Build GRCh38
rs964084335 Chr.11: 720524-720524 on Build GRCh38
rs866506246 Chr.11: 720499-720499 on Build GRCh38
rs867054985 Chr.11: 720850-720850 on Build GRCh38
rs371905684 Chr.11: 720665-720665 on Build GRCh38
rs774848969 Chr.11: 720678-720678 on Build GRCh38
rs867500914 Chr.11: 720592-720592 on Build GRCh38
rs758406101 Chr.11: 720753-720753 on Build GRCh38
rs767652699 Chr.11: 720751-720760 on Build GRCh38
rs749057351 Chr.11: 720884-720886 on Build GRCh38
rs368045203 Chr.11: 720547-720547 on Build GRCh38
rs760373853 Chr.11: 720533-720533 on Build GRCh38
rs748168915 Chr.11: 720735-720735 on Build GRCh38
rs755290745 Chr.11: 720845-720845 on Build GRCh38
rs763014969 Chr.11: 720680-720680 on Build GRCh38
rs775142661 Chr.11: 720773-720773 on Build GRCh38
rs745594494 Chr.11: 720764-720764 on Build GRCh38
rs866382650 Chr.11: 720878-720878 on Build GRCh38
rs955864304 Chr.11: 720666-720666 on Build GRCh38
rs750257597 Chr.11: 720777-720778 on Build GRCh38
rs748734159 Chr.11: 720633-720633 on Build GRCh38
rs932639124 Chr.11: 720454-720454 on Build GRCh38
rs577356107 Chr.11: 720778-720778 on Build GRCh38
rs768814580 Chr.11: 720617-720617 on Build GRCh38
rs907755581 Chr.11: 720620-720620 on Build GRCh38
rs751397220 Chr.11: 720580-720580 on Build GRCh38
rs761814265 Chr.11: 720781-720781 on Build GRCh38
rs963279459 Chr.11: 720895-720895 on Build GRCh38
rs779103363 Chr.11: 720717-720717 on Build GRCh38
rs1012973631 Chr.11: 720622-720622 on Build GRCh38
rs139173072 Chr.11: 720668-720668 on Build GRCh38
rs554029920 Chr.11: 720767-720767 on Build GRCh38
rs549844759 Chr.11: 720455-720455 on Build GRCh38
rs934957546 Chr.11: 720736-720736 on Build GRCh38
rs767466392 Chr.11: 720812-720812 on Build GRCh38
rs894875717 Chr.11: 720431-720431 on Build GRCh38
rs752722738 Chr.11: 720564-720564 on Build GRCh38
rs983729499 Chr.11: 720417-720417 on Build GRCh38
rs762570304 Chr.11: 720777-720777 on Build GRCh38
rs753421157 Chr.11: 720540-720540 on Build GRCh38
rs985834266 Chr.11: 720743-720743 on Build GRCh38
rs904185089 Chr.11: 720820-720820 on Build GRCh38
rs1056665536 Chr.11: 720796-720796 on Build GRCh38
rs906189716 Chr.11: 720722-720722 on Build GRCh38
rs973254815 Chr.11: 720897-720897 on Build GRCh38
rs368090270 Chr.11: 720426-720426 on Build GRCh38
rs772524621 Chr.11: 720508-720508 on Build GRCh38
rs1012510123 Chr.11: 720807-720807 on Build GRCh38
rs955810683 Chr.11: 720859-720859 on Build GRCh38
rs959635454 Chr.11: 720614-720614 on Build GRCh38
rs376502072 Chr.11: 720675-720675 on Build GRCh38
rs750167406 Chr.11: 720819-720819 on Build GRCh38
rs968516251 Chr.11: 720418-720418 on Build GRCh38
rs893983912 Chr.11: 720482-720482 on Build GRCh38
rs758249192 Chr.11: 720565-720565 on Build GRCh38
rs976529477 Chr.11: 720860-720860 on Build GRCh38
rs773538757 Chr.11: 720523-720523 on Build GRCh38
rs922556270 Chr.11: 720452-720452 on Build GRCh38
rs368598389 Chr.11: 720684-720684 on Build GRCh38
rs1046050659 Chr.11: 720442-720442 on Build GRCh38
rs532926976 Chr.11: 720450-720450 on Build GRCh38
rs757143018 Chr.11: 720697-720697 on Build GRCh38
rs201086107 Chr.11: 720553-720553 on Build GRCh38
rs947716869 Chr.11: 720432-720432 on Build GRCh38
rs1037113590 Chr.11: 720757-720757 on Build GRCh38
rs774464810 Chr.11: 720780-720780 on Build GRCh38
rs895372854 Chr.11: 720798-720798 on Build GRCh38
rs535067318 Chr.11: 720593-720593 on Build GRCh38
rs756072469 Chr.11: 720780-720781 on Build GRCh38
rs542585760 Chr.11: 720488-720488 on Build GRCh38
rs1039315639 Chr.11: 720769-720769 on Build GRCh38
rs778266262 Chr.11: 720759-720759 on Build GRCh38
rs1029663104 Chr.11: 720542-720542 on Build GRCh38
rs767384396 Chr.11: 720702-720702 on Build GRCh38
rs983397593 Chr.11: 720562-720562 on Build GRCh38
rs771226190 Chr.11: 720667-720667 on Build GRCh38
rs757238153 Chr.11: 720743-720745 on Build GRCh38
rs747463415 Chr.11: 720760-720760 on Build GRCh38
rs999777022 Chr.11: 720469-720469 on Build GRCh38
rs759096861 Chr.11: 720541-720541 on Build GRCh38
rs758493990 Chr.11: 720892-720892 on Build GRCh38
rs754806041 Chr.11: 720892-720893 on Build GRCh38
rs547570959 Chr.11: 720420-720420 on Build GRCh38
rs1014864959 Chr.11: 720884-720884 on Build GRCh38
rs755130479 Chr.11: 720714-720714 on Build GRCh38

View Full Product Details