Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 828530-829032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRACR2B
Entrez Gene ID
283229
Gene Name
calcium release activated channel regulator 2B
Gene Aliases
EFCAB4A
UniGene
Hs.660936
Gene Chromosome Location
Chr.11: 825558-831991 on Build GRCh38
Transcripts
NM_001286606.1 NM_173584.4 XM_011520011.1 XM_011520019.1 XM_017017581.1 XM_017017582.1 XM_017017583.1 XM_017017584.1 XM_017017585.1 XM_017017586.1 XM_017017587.1 XM_017017588.1 XM_017017589.1 XM_017017590.1 XM_017017591.1 XM_017017592.1 XM_017017593.1 XM_017017594.1 XM_017017595.1 XM_017017596.1 XM_017017597.1 XM_017017598.1 XM_017017599.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5542878 c.178G>A p.D60N Chr.11: 828864-828864 on Build GRCh38
932544 c.191C>T p.T64M Chr.11: 828877-828877 on Build GRCh38
932549 c.235C>A p.H79N Chr.11: 828921-828921 on Build GRCh38
932528 c.165+2T>C p.? Chr.11: 828774-828774 on Build GRCh38
5503695 c.134C>T p.A45V Chr.11: 828741-828741 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758800144 Chr.11: 828699-828699 on Build GRCh38
rs957216008 Chr.11: 828952-828952 on Build GRCh38
rs371562171 Chr.11: 828707-828707 on Build GRCh38
rs747843382 Chr.11: 828792-828792 on Build GRCh38
rs769099541 Chr.11: 828607-828607 on Build GRCh38
rs935946898 Chr.11: 828805-828805 on Build GRCh38
rs367958561 Chr.11: 828971-828971 on Build GRCh38
rs759154160 Chr.11: 828803-828803 on Build GRCh38
rs750880313 Chr.11: 828822-828822 on Build GRCh38
rs368077191 Chr.11: 828701-828701 on Build GRCh38
rs538494554 Chr.11: 828802-828802 on Build GRCh38
rs200686837 Chr.11: 828764-828764 on Build GRCh38
rs758309931 Chr.11: 828590-828590 on Build GRCh38
rs867555788 Chr.11: 828678-828678 on Build GRCh38
rs768727022 Chr.11: 828786-828786 on Build GRCh38
rs774046753 Chr.11: 828928-828928 on Build GRCh38
rs369982644 Chr.11: 828939-828939 on Build GRCh38
rs756350831 Chr.11: 828839-828840 on Build GRCh38
rs985978413 Chr.11: 828983-828983 on Build GRCh38
rs764981337 Chr.11: 828766-828766 on Build GRCh38
rs199837373 Chr.11: 828886-828886 on Build GRCh38
rs756145742 Chr.11: 828779-828779 on Build GRCh38
rs369130837 Chr.11: 828726-828726 on Build GRCh38
rs752818986 Chr.11: 828968-828968 on Build GRCh38
rs781207611 Chr.11: 828899-828899 on Build GRCh38
rs779890092 Chr.11: 828668-828668 on Build GRCh38
rs904439343 Chr.11: 828672-828672 on Build GRCh38
rs534373499 Chr.11: 828902-828902 on Build GRCh38
rs775747373 Chr.11: 828683-828683 on Build GRCh38
rs753549408 Chr.11: 828698-828698 on Build GRCh38
rs539844309 Chr.11: 829010-829010 on Build GRCh38
rs770556498 Chr.11: 828594-828594 on Build GRCh38
rs750751434 Chr.11: 828870-828870 on Build GRCh38
rs992642167 Chr.11: 828588-828588 on Build GRCh38
rs759661293 Chr.11: 828745-828745 on Build GRCh38
rs751837387 Chr.11: 828981-828981 on Build GRCh38
rs373015697 Chr.11: 828727-828727 on Build GRCh38
rs764543928 Chr.11: 828666-828667 on Build GRCh38
rs765492019 Chr.11: 828622-828622 on Build GRCh38
rs372435980 Chr.11: 828892-828892 on Build GRCh38
rs749362848 Chr.11: 828834-828834 on Build GRCh38
rs553411358 Chr.11: 828686-828686 on Build GRCh38
rs374787891 Chr.11: 828821-828821 on Build GRCh38
rs767925446 Chr.11: 828779-828779 on Build GRCh38
rs541669383 Chr.11: 828680-828680 on Build GRCh38
rs758616740 Chr.11: 828889-828889 on Build GRCh38
rs766562819 Chr.11: 828943-828955 on Build GRCh38
rs780596259 Chr.11: 828664-828664 on Build GRCh38
rs753941141 Chr.11: 828943-828943 on Build GRCh38
rs977266812 Chr.11: 828927-828927 on Build GRCh38
rs759086070 Chr.11: 828639-828640 on Build GRCh38
rs760069812 Chr.11: 828959-828959 on Build GRCh38
rs769873372 Chr.11: 828718-828718 on Build GRCh38
rs562150479 Chr.11: 828609-828609 on Build GRCh38
rs773218431 Chr.11: 828746-828746 on Build GRCh38
rs201478416 Chr.11: 828635-828635 on Build GRCh38
rs773400186 Chr.11: 828557-828557 on Build GRCh38
rs371173369 Chr.11: 828846-828846 on Build GRCh38
rs199782263 Chr.11: 828767-828767 on Build GRCh38
rs765707873 Chr.11: 828964-828964 on Build GRCh38
rs201730917 Chr.11: 828685-828685 on Build GRCh38
rs554285465 Chr.11: 828597-828597 on Build GRCh38
rs764005845 Chr.11: 828651-828651 on Build GRCh38
rs756950699 Chr.11: 828826-828826 on Build GRCh38
rs377168504 Chr.11: 828788-828788 on Build GRCh38
rs867288821 Chr.11: 828612-828612 on Build GRCh38
rs548517140 Chr.11: 828828-828828 on Build GRCh38
rs368464681 Chr.11: 828608-828608 on Build GRCh38
rs751464129 Chr.11: 828659-828659 on Build GRCh38
rs546209108 Chr.11: 828619-828619 on Build GRCh38
rs967564663 Chr.11: 828925-828925 on Build GRCh38
rs202043008 Chr.11: 828864-828864 on Build GRCh38
rs774859905 Chr.11: 828667-828667 on Build GRCh38
rs757077556 Chr.11: 828994-828994 on Build GRCh38
rs761091360 Chr.11: 828872-828872 on Build GRCh38
rs747504738 Chr.11: 828674-828674 on Build GRCh38
rs28507876 Chr.11: 828784-828784 on Build GRCh38
rs777727582 Chr.11: 828595-828595 on Build GRCh38
rs34871888 Chr.11: 828748-828748 on Build GRCh38
rs764476341 Chr.11: 828816-828816 on Build GRCh38
rs762105460 Chr.11: 828682-828683 on Build GRCh38
rs558310500 Chr.11: 828801-828801 on Build GRCh38
rs375669287 Chr.11: 828885-828885 on Build GRCh38
rs748435879 Chr.11: 828840-828840 on Build GRCh38
rs772583367 Chr.11: 828671-828671 on Build GRCh38
rs1005782765 Chr.11: 828891-828891 on Build GRCh38
rs770922466 Chr.11: 828618-828618 on Build GRCh38
rs771908014 Chr.11: 828793-828793 on Build GRCh38
rs774338646 Chr.11: 828710-828710 on Build GRCh38
rs1023822114 Chr.11: 828637-828637 on Build GRCh38
rs927021582 Chr.11: 828556-828556 on Build GRCh38
rs1041600245 Chr.11: 828667-828667 on Build GRCh38
rs375054445 Chr.11: 828763-828763 on Build GRCh38
rs911046431 Chr.11: 828896-828896 on Build GRCh38
rs576238806 Chr.11: 828643-828643 on Build GRCh38
rs776769935 Chr.11: 828857-828857 on Build GRCh38
rs745508251 Chr.11: 828600-828600 on Build GRCh38
rs764786031 Chr.11: 828804-828804 on Build GRCh38
rs368569122 Chr.11: 828845-828845 on Build GRCh38
rs760736195 Chr.11: 828558-828558 on Build GRCh38
rs748514649 Chr.11: 828669-828669 on Build GRCh38
rs375873951 Chr.11: 828715-828715 on Build GRCh38
rs770864140 Chr.11: 828894-828894 on Build GRCh38
rs1013793094 Chr.11: 828634-828636 on Build GRCh38
rs754351281 Chr.11: 828878-828878 on Build GRCh38
rs910472956 Chr.11: 828988-828988 on Build GRCh38
rs752584386 Chr.11: 828589-828589 on Build GRCh38
rs372499416 Chr.11: 828676-828676 on Build GRCh38
rs994747123 Chr.11: 828703-828703 on Build GRCh38
rs752459636 Chr.11: 828623-828623 on Build GRCh38
rs750040448 Chr.11: 828665-828665 on Build GRCh38
rs746945029 Chr.11: 828890-828890 on Build GRCh38
rs896536942 Chr.11: 828797-828797 on Build GRCh38
rs7126805 Chr.11: 828916-828916 on Build GRCh38
rs747450805 Chr.11: 828705-828705 on Build GRCh38
rs774965246 Chr.11: 828865-828865 on Build GRCh38
rs780838170 Chr.11: 828599-828599 on Build GRCh38
rs765049724 Chr.11: 828584-828584 on Build GRCh38
rs200385933 Chr.11: 828687-828687 on Build GRCh38
rs769137654 Chr.11: 828908-828908 on Build GRCh38
rs778085923 Chr.11: 828979-828979 on Build GRCh38
rs757700011 Chr.11: 828768-828768 on Build GRCh38
rs768289892 Chr.11: 828919-828919 on Build GRCh38
rs370281925 Chr.11: 828789-828789 on Build GRCh38
rs767348897 Chr.11: 828741-828741 on Build GRCh38
rs780126428 Chr.11: 828783-828783 on Build GRCh38
rs958324957 Chr.11: 828898-828898 on Build GRCh38
rs201725212 Chr.11: 828941-828941 on Build GRCh38
rs754738058 Chr.11: 828577-828577 on Build GRCh38
rs781492690 Chr.11: 828774-828774 on Build GRCh38
rs574851252 Chr.11: 828552-828552 on Build GRCh38
rs750816074 Chr.11: 828789-828790 on Build GRCh38
rs746524456 Chr.11: 828596-828596 on Build GRCh38
rs534257522 Chr.11: 828564-828564 on Build GRCh38
rs758914691 Chr.11: 828862-828862 on Build GRCh38
rs766854771 Chr.11: 828877-828877 on Build GRCh38
rs760968577 Chr.11: 828938-828938 on Build GRCh38
rs754151216 Chr.11: 828817-828817 on Build GRCh38
rs754023295 Chr.11: 828566-828566 on Build GRCh38
rs756587360 Chr.11: 828827-828827 on Build GRCh38
rs754975053 Chr.11: 828882-828882 on Build GRCh38
rs763781115 Chr.11: 828867-828867 on Build GRCh38
rs759700888 Chr.11: 828621-828621 on Build GRCh38
rs778279448 Chr.11: 828673-828673 on Build GRCh38
rs746378063 Chr.11: 828775-828775 on Build GRCh38
rs563825697 Chr.11: 828760-828760 on Build GRCh38
rs200216625 Chr.11: 828684-828684 on Build GRCh38
rs755142897 Chr.11: 829003-829015 on Build GRCh38
rs375526040 Chr.11: 828731-828731 on Build GRCh38
rs74385189 Chr.11: 828903-828903 on Build GRCh38
rs776097114 Chr.11: 828595-828596 on Build GRCh38
rs778939255 Chr.11: 828836-828836 on Build GRCh38
rs774698938 Chr.11: 828791-828791 on Build GRCh38
rs779798185 Chr.11: 828697-828697 on Build GRCh38
rs986570636 Chr.11: 828740-828740 on Build GRCh38
rs369737052 Chr.11: 828732-828732 on Build GRCh38

View Full Product Details