Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 836111-836635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD151
Entrez Gene ID
977
Gene Name
CD151 molecule (Raph blood group)
Gene Aliases
GP27, MER2, PETA-3, RAPH, SFA1, TSPAN24
UniGene
Hs.654379
Gene Chromosome Location
Chr.11: 832952-838835 on Build GRCh38
Transcripts
NM_001039490.1 NM_004357.4 NM_139029.1 NM_139030.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5957271 c.47T>G p.L16R Chr.11: 836116-836116 on Build GRCh38
4849335 c.188T>G p.I63S Chr.11: 836354-836354 on Build GRCh38
4608392 c.94C>A p.L32M Chr.11: 836260-836260 on Build GRCh38
6623402 c.63T>G p.F21L Chr.11: 836132-836132 on Build GRCh38
7256990 c.240C>T p.C80C Chr.11: 836406-836406 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771096074 Chr.11: 836397-836397 on Build GRCh38
rs374162358 Chr.11: 836367-836367 on Build GRCh38
rs761070004 Chr.11: 836355-836355 on Build GRCh38
rs771438118 Chr.11: 836202-836204 on Build GRCh38
rs895863238 Chr.11: 836459-836459 on Build GRCh38
rs774385687 Chr.11: 836207-836207 on Build GRCh38
rs542085660 Chr.11: 836445-836445 on Build GRCh38
rs780093397 Chr.11: 836473-836473 on Build GRCh38
rs950759437 Chr.11: 836466-836466 on Build GRCh38
rs768756178 Chr.11: 836481-836481 on Build GRCh38
rs760573926 Chr.11: 836182-836182 on Build GRCh38
rs754521405 Chr.11: 836189-836189 on Build GRCh38
rs973633541 Chr.11: 836551-836551 on Build GRCh38
rs751285028 Chr.11: 836349-836349 on Build GRCh38
rs781279920 Chr.11: 836454-836454 on Build GRCh38
rs763769216 Chr.11: 836341-836341 on Build GRCh38
rs545338675 Chr.11: 836226-836226 on Build GRCh38
rs768698548 Chr.11: 836171-836171 on Build GRCh38
rs943923697 Chr.11: 836213-836213 on Build GRCh38
rs748794395 Chr.11: 836173-836178 on Build GRCh38
rs768810190 Chr.11: 836204-836204 on Build GRCh38
rs753224244 Chr.11: 836435-836435 on Build GRCh38
rs138731111 Chr.11: 836338-836338 on Build GRCh38
rs767144167 Chr.11: 836422-836422 on Build GRCh38
rs370830997 Chr.11: 836198-836198 on Build GRCh38
rs745521783 Chr.11: 836172-836177 on Build GRCh38
rs772268648 Chr.11: 836303-836303 on Build GRCh38
rs1001136902 Chr.11: 836569-836569 on Build GRCh38
rs368857499 Chr.11: 836179-836179 on Build GRCh38
rs758951148 Chr.11: 836438-836438 on Build GRCh38
rs772319843 Chr.11: 836225-836225 on Build GRCh38
rs766369908 Chr.11: 836183-836183 on Build GRCh38
rs748378441 Chr.11: 836224-836224 on Build GRCh38
rs111768956 Chr.11: 836154-836154 on Build GRCh38
rs777003760 Chr.11: 836591-836591 on Build GRCh38
rs972683650 Chr.11: 836223-836223 on Build GRCh38
rs537372809 Chr.11: 836553-836553 on Build GRCh38
rs892029296 Chr.11: 836507-836507 on Build GRCh38
rs781668686 Chr.11: 836191-836191 on Build GRCh38
rs752191278 Chr.11: 836250-836250 on Build GRCh38
rs773994585 Chr.11: 836486-836486 on Build GRCh38
rs780382052 Chr.11: 836288-836288 on Build GRCh38
rs141569440 Chr.11: 836407-836407 on Build GRCh38
rs1030027506 Chr.11: 836248-836248 on Build GRCh38
rs774633764 Chr.11: 836173-836173 on Build GRCh38
rs757673547 Chr.11: 836135-836135 on Build GRCh38
rs113725314 Chr.11: 836444-836444 on Build GRCh38
rs917173152 Chr.11: 836585-836585 on Build GRCh38
rs948684526 Chr.11: 836586-836586 on Build GRCh38
rs762570483 Chr.11: 836339-836339 on Build GRCh38
rs753229468 Chr.11: 836185-836185 on Build GRCh38
rs775577440 Chr.11: 836409-836409 on Build GRCh38
rs866886079 Chr.11: 836351-836351 on Build GRCh38
rs201750899 Chr.11: 836166-836166 on Build GRCh38
rs774058678 Chr.11: 836470-836470 on Build GRCh38
rs772604592 Chr.11: 836185-836185 on Build GRCh38
rs985313285 Chr.11: 836174-836174 on Build GRCh38
rs754388960 Chr.11: 836289-836289 on Build GRCh38
rs777975809 Chr.11: 836306-836306 on Build GRCh38
rs781747523 Chr.11: 836138-836138 on Build GRCh38
rs376496318 Chr.11: 836315-836315 on Build GRCh38
rs755504233 Chr.11: 836366-836366 on Build GRCh38
rs963137891 Chr.11: 836548-836548 on Build GRCh38
rs112741073 Chr.11: 836443-836443 on Build GRCh38
rs147298858 Chr.11: 836377-836377 on Build GRCh38
rs571864895 Chr.11: 836503-836503 on Build GRCh38
rs375022791 Chr.11: 836546-836547 on Build GRCh38
rs200787857 Chr.11: 836177-836177 on Build GRCh38
rs557213758 Chr.11: 836554-836554 on Build GRCh38
rs887886783 Chr.11: 836299-836299 on Build GRCh38
rs780621314 Chr.11: 836199-836199 on Build GRCh38
rs761054844 Chr.11: 836199-836200 on Build GRCh38
rs562608258 Chr.11: 836549-836549 on Build GRCh38
rs948798762 Chr.11: 836460-836460 on Build GRCh38
rs770788850 Chr.11: 836230-836230 on Build GRCh38
rs1019424026 Chr.11: 836437-836437 on Build GRCh38
rs776341908 Chr.11: 836320-836320 on Build GRCh38
rs776929851 Chr.11: 836475-836476 on Build GRCh38
rs34987505 Chr.11: 836177-836178 on Build GRCh38
rs752707203 Chr.11: 836370-836370 on Build GRCh38
rs764784965 Chr.11: 836245-836245 on Build GRCh38
rs750755109 Chr.11: 836279-836279 on Build GRCh38
rs749256482 Chr.11: 836200-836200 on Build GRCh38
rs552112986 Chr.11: 836501-836501 on Build GRCh38
rs747742714 Chr.11: 836311-836311 on Build GRCh38
rs761528106 Chr.11: 836420-836420 on Build GRCh38
rs573935113 Chr.11: 836590-836590 on Build GRCh38
rs749218525 Chr.11: 836479-836479 on Build GRCh38
rs779521471 Chr.11: 836172-836173 on Build GRCh38
rs1017325058 Chr.11: 836547-836547 on Build GRCh38
rs772199120 Chr.11: 836436-836436 on Build GRCh38
rs748293136 Chr.11: 836301-836301 on Build GRCh38
rs142774509 Chr.11: 836376-836376 on Build GRCh38
rs756548739 Chr.11: 836280-836280 on Build GRCh38
rs770737519 Chr.11: 836313-836313 on Build GRCh38
rs377680693 Chr.11: 836423-836423 on Build GRCh38
rs762694458 Chr.11: 836266-836266 on Build GRCh38
rs773625647 Chr.11: 836189-836190 on Build GRCh38
rs759489509 Chr.11: 836327-836327 on Build GRCh38
rs150909098 Chr.11: 836417-836417 on Build GRCh38
rs779017734 Chr.11: 836295-836295 on Build GRCh38
rs747670309 Chr.11: 836487-836487 on Build GRCh38
rs747207481 Chr.11: 836391-836391 on Build GRCh38
rs758922995 Chr.11: 836235-836235 on Build GRCh38
rs546760064 Chr.11: 836463-836463 on Build GRCh38
rs142659029 Chr.11: 836499-836499 on Build GRCh38
rs746305303 Chr.11: 836193-836193 on Build GRCh38
rs200700576 Chr.11: 836167-836167 on Build GRCh38
rs751702824 Chr.11: 836452-836452 on Build GRCh38
rs763900802 Chr.11: 836268-836268 on Build GRCh38
rs769690275 Chr.11: 836330-836330 on Build GRCh38
rs756671568 Chr.11: 836195-836195 on Build GRCh38
rs374946921 Chr.11: 836178-836178 on Build GRCh38
rs746358515 Chr.11: 836150-836150 on Build GRCh38
rs200217814 Chr.11: 836426-836426 on Build GRCh38
rs1055137245 Chr.11: 836541-836541 on Build GRCh38
rs112992215 Chr.11: 836249-836249 on Build GRCh38
rs770467521 Chr.11: 836159-836159 on Build GRCh38
rs56063221 Chr.11: 836540-836540 on Build GRCh38
rs776663637 Chr.11: 836234-836234 on Build GRCh38
rs201499072 Chr.11: 836544-836544 on Build GRCh38
rs116211118 Chr.11: 836418-836418 on Build GRCh38
rs752418503 Chr.11: 836190-836190 on Build GRCh38
rs889464465 Chr.11: 836583-836583 on Build GRCh38
rs769300288 Chr.11: 836360-836360 on Build GRCh38
rs776351828 Chr.11: 836502-836502 on Build GRCh38
rs548885427 Chr.11: 836440-836440 on Build GRCh38
rs747271748 Chr.11: 836134-836134 on Build GRCh38
rs879089262 Chr.11: 836247-836247 on Build GRCh38
rs34726520 Chr.11: 836406-836406 on Build GRCh38
rs755485787 Chr.11: 836294-836294 on Build GRCh38
rs566560607 Chr.11: 836464-836464 on Build GRCh38
rs748770542 Chr.11: 836411-836411 on Build GRCh38
rs28434354 Chr.11: 836227-836227 on Build GRCh38
rs918074521 Chr.11: 836543-836543 on Build GRCh38
rs754193115 Chr.11: 836358-836358 on Build GRCh38
rs1053422326 Chr.11: 836577-836577 on Build GRCh38
rs770882120 Chr.11: 836453-836453 on Build GRCh38
rs398075671 Chr.11: 836548-836548 on Build GRCh38
rs370735173 Chr.11: 836543-836544 on Build GRCh38

View Full Product Details