Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 836591-837082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD151
Entrez Gene ID
977
Gene Name
CD151 molecule (Raph blood group)
Gene Aliases
GP27, MER2, PETA-3, RAPH, SFA1, TSPAN24
UniGene
Hs.654379
Gene Chromosome Location
Chr.11: 832952-838835 on Build GRCh38
Transcripts
NM_001039490.1 NM_004357.4 NM_139029.1 NM_139030.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3383892 c.349C>G p.Q117E Chr.11: 836841-836841 on Build GRCh38
3454021 c.331G>A p.A111T Chr.11: 836823-836823 on Build GRCh38
6489312 c.292C>A p.L98I Chr.11: 836784-836784 on Build GRCh38
127573 c.285C>T p.I95I Chr.11: 836777-836777 on Build GRCh38
7175330 c.307C>T p.L103L Chr.11: 836799-836799 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762055563 Chr.11: 836714-836714 on Build GRCh38
rs750481097 Chr.11: 836752-836752 on Build GRCh38
rs375930736 Chr.11: 837060-837060 on Build GRCh38
rs769034746 Chr.11: 836889-836889 on Build GRCh38
rs760805374 Chr.11: 836828-836828 on Build GRCh38
rs546772594 Chr.11: 836824-836824 on Build GRCh38
rs113718864 Chr.11: 836654-836654 on Build GRCh38
rs544660476 Chr.11: 836759-836759 on Build GRCh38
rs1040683509 Chr.11: 836984-836984 on Build GRCh38
rs1053678440 Chr.11: 836620-836620 on Build GRCh38
rs766766478 Chr.11: 836829-836829 on Build GRCh38
rs374490956 Chr.11: 836764-836764 on Build GRCh38
rs765435179 Chr.11: 836743-836743 on Build GRCh38
rs540411150 Chr.11: 836759-836760 on Build GRCh38
rs1024120527 Chr.11: 836936-836937 on Build GRCh38
rs573246601 Chr.11: 836673-836673 on Build GRCh38
rs146079952 Chr.11: 836820-836820 on Build GRCh38
rs965574058 Chr.11: 836627-836627 on Build GRCh38
rs200736365 Chr.11: 836822-836822 on Build GRCh38
rs28620453 Chr.11: 836971-836971 on Build GRCh38
rs890042570 Chr.11: 836678-836678 on Build GRCh38
rs1034261867 Chr.11: 836908-836908 on Build GRCh38
rs144204595 Chr.11: 836786-836786 on Build GRCh38
rs1006595615 Chr.11: 836825-836825 on Build GRCh38
rs763318055 Chr.11: 836808-836808 on Build GRCh38
rs1019400031 Chr.11: 836618-836618 on Build GRCh38
rs753629231 Chr.11: 836836-836836 on Build GRCh38
rs138730289 Chr.11: 836871-836871 on Build GRCh38
rs76638476 Chr.11: 837059-837059 on Build GRCh38
rs978590689 Chr.11: 836939-836939 on Build GRCh38
rs568625605 Chr.11: 836758-836759 on Build GRCh38
rs1047288801 Chr.11: 837007-837007 on Build GRCh38
rs368338565 Chr.11: 836894-836894 on Build GRCh38
rs777916346 Chr.11: 836875-836875 on Build GRCh38
rs763444209 Chr.11: 836749-836749 on Build GRCh38
rs769081481 Chr.11: 836810-836810 on Build GRCh38
rs757285151 Chr.11: 836760-836760 on Build GRCh38
rs952629472 Chr.11: 836686-836686 on Build GRCh38
rs143711290 Chr.11: 836914-836914 on Build GRCh38
rs750636942 Chr.11: 836762-836762 on Build GRCh38
rs1014015698 Chr.11: 836917-836917 on Build GRCh38
rs774746771 Chr.11: 836815-836815 on Build GRCh38
rs370471055 Chr.11: 836796-836796 on Build GRCh38
rs765563249 Chr.11: 836747-836747 on Build GRCh38
rs746609476 Chr.11: 836789-836789 on Build GRCh38
rs913313056 Chr.11: 836823-836823 on Build GRCh38
rs765757864 Chr.11: 836966-836966 on Build GRCh38
rs998834725 Chr.11: 836663-836663 on Build GRCh38
rs778944602 Chr.11: 836859-836859 on Build GRCh38
rs74045201 Chr.11: 836972-836972 on Build GRCh38
rs764041356 Chr.11: 836863-836866 on Build GRCh38
rs1041173388 Chr.11: 836837-836837 on Build GRCh38
rs867231591 Chr.11: 836777-836777 on Build GRCh38
rs970810482 Chr.11: 836935-836935 on Build GRCh38
rs551960866 Chr.11: 836885-836885 on Build GRCh38
rs539151436 Chr.11: 836973-836973 on Build GRCh38
rs775870497 Chr.11: 836748-836748 on Build GRCh38
rs777391775 Chr.11: 836787-836787 on Build GRCh38
rs984010079 Chr.11: 836687-836687 on Build GRCh38
rs369569073 Chr.11: 836750-836750 on Build GRCh38
rs199955739 Chr.11: 836888-836888 on Build GRCh38
rs185487588 Chr.11: 837006-837006 on Build GRCh38
rs113926671 Chr.11: 836767-836767 on Build GRCh38
rs959355483 Chr.11: 836913-836913 on Build GRCh38
rs754825324 Chr.11: 836848-836848 on Build GRCh38
rs759833283 Chr.11: 836736-836737 on Build GRCh38
rs771697606 Chr.11: 836720-836720 on Build GRCh38
rs545089055 Chr.11: 836685-836685 on Build GRCh38
rs751935271 Chr.11: 836905-836905 on Build GRCh38
rs990854325 Chr.11: 836718-836718 on Build GRCh38
rs762974959 Chr.11: 836781-836783 on Build GRCh38
rs375782528 Chr.11: 836895-836895 on Build GRCh38
rs950540477 Chr.11: 836915-836915 on Build GRCh38
rs142548410 Chr.11: 837061-837061 on Build GRCh38
rs574746390 Chr.11: 837049-837049 on Build GRCh38
rs770509073 Chr.11: 836881-836881 on Build GRCh38
rs148125849 Chr.11: 836993-836993 on Build GRCh38
rs924095086 Chr.11: 836947-836947 on Build GRCh38
rs536443951 Chr.11: 836633-836633 on Build GRCh38
rs747628175 Chr.11: 836772-836772 on Build GRCh38
rs758399675 Chr.11: 836868-836868 on Build GRCh38
rs775816996 Chr.11: 836799-836799 on Build GRCh38
rs565254857 Chr.11: 836694-836694 on Build GRCh38
rs561171137 Chr.11: 837032-837032 on Build GRCh38
rs536456894 Chr.11: 836961-836961 on Build GRCh38
rs34581917 Chr.11: 836864-836864 on Build GRCh38
rs772770951 Chr.11: 836734-836734 on Build GRCh38
rs751464320 Chr.11: 836868-836869 on Build GRCh38
rs936863228 Chr.11: 836958-836958 on Build GRCh38
rs1017311968 Chr.11: 836697-836697 on Build GRCh38
rs537436125 Chr.11: 836903-836903 on Build GRCh38
rs911434740 Chr.11: 837043-837046 on Build GRCh38
rs1051248865 Chr.11: 837038-837038 on Build GRCh38
rs376390859 Chr.11: 836765-836765 on Build GRCh38
rs1025416531 Chr.11: 836916-836916 on Build GRCh38
rs1025917173 Chr.11: 836684-836684 on Build GRCh38
rs558768047 Chr.11: 836987-836987 on Build GRCh38

View Full Product Details