Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1554113-1554603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP8
Entrez Gene ID
1850
Gene Name
dual specificity phosphatase 8
Gene Aliases
C11orf81, HB5, HVH-5, HVH8
UniGene
Hs.41688
Gene Chromosome Location
Chr.11: 1554051-1572839 on Build GRCh38
Transcripts
NM_004420.2 XM_011519932.2 XM_011519933.2

SNP Detail

SNP ID Chromosome Location
rs762925003 Chr.11: 1554202-1554202 on Build GRCh38
rs770068825 Chr.11: 1554531-1554531 on Build GRCh38
rs1004605998 Chr.11: 1554411-1554411 on Build GRCh38
rs907560069 Chr.11: 1554138-1554138 on Build GRCh38
rs985009605 Chr.11: 1554276-1554276 on Build GRCh38
rs909358618 Chr.11: 1554280-1554280 on Build GRCh38
rs576100381 Chr.11: 1554149-1554149 on Build GRCh38
rs139969275 Chr.11: 1554143-1554148 on Build GRCh38
rs1000747360 Chr.11: 1554150-1554150 on Build GRCh38
rs182379864 Chr.11: 1554480-1554480 on Build GRCh38
rs540555616 Chr.11: 1554197-1554197 on Build GRCh38
rs952522062 Chr.11: 1554537-1554537 on Build GRCh38
rs577157198 Chr.11: 1554415-1554415 on Build GRCh38
rs763970892 Chr.11: 1554216-1554216 on Build GRCh38
rs896502547 Chr.11: 1554403-1554403 on Build GRCh38
rs938298216 Chr.11: 1554332-1554332 on Build GRCh38
rs977233967 Chr.11: 1554208-1554208 on Build GRCh38
rs190205565 Chr.11: 1554478-1554478 on Build GRCh38
rs1042368627 Chr.11: 1554438-1554438 on Build GRCh38
rs1034287624 Chr.11: 1554139-1554139 on Build GRCh38
rs1001346950 Chr.11: 1554456-1554456 on Build GRCh38
rs1019509195 Chr.11: 1554500-1554500 on Build GRCh38
rs1006822537 Chr.11: 1554488-1554488 on Build GRCh38
rs965046535 Chr.11: 1554528-1554528 on Build GRCh38
rs764523044 Chr.11: 1554548-1554548 on Build GRCh38
rs977474001 Chr.11: 1554575-1554575 on Build GRCh38
rs887141470 Chr.11: 1554346-1554346 on Build GRCh38
rs1048624768 Chr.11: 1554313-1554313 on Build GRCh38
rs1020965310 Chr.11: 1554142-1554142 on Build GRCh38
rs1037439495 Chr.11: 1554147-1554147 on Build GRCh38
rs989824886 Chr.11: 1554165-1554165 on Build GRCh38
rs34698106 Chr.11: 1554440-1554440 on Build GRCh38
rs921526295 Chr.11: 1554252-1554252 on Build GRCh38
rs912285612 Chr.11: 1554219-1554219 on Build GRCh38
rs543613998 Chr.11: 1554240-1554240 on Build GRCh38
rs748210524 Chr.11: 1554253-1554253 on Build GRCh38
rs1032584765 Chr.11: 1554151-1554151 on Build GRCh38
rs12284550 Chr.11: 1554377-1554377 on Build GRCh38
rs1040247113 Chr.11: 1554233-1554233 on Build GRCh38
rs1011541527 Chr.11: 1554525-1554525 on Build GRCh38
rs954019416 Chr.11: 1554167-1554167 on Build GRCh38
rs565097050 Chr.11: 1554268-1554268 on Build GRCh38
rs371389211 Chr.11: 1554269-1554269 on Build GRCh38
rs149565066 Chr.11: 1554530-1554530 on Build GRCh38
rs941690088 Chr.11: 1554143-1554143 on Build GRCh38
rs12288630 Chr.11: 1554206-1554206 on Build GRCh38
rs1034153217 Chr.11: 1554494-1554494 on Build GRCh38
rs985762625 Chr.11: 1554189-1554189 on Build GRCh38
rs944243769 Chr.11: 1554220-1554220 on Build GRCh38

View Full Product Details