Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1557195-1557692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP8
Entrez Gene ID
1850
Gene Name
dual specificity phosphatase 8
Gene Aliases
C11orf81, HB5, HVH-5, HVH8
UniGene
Hs.41688
Gene Chromosome Location
Chr.11: 1554051-1572839 on Build GRCh38
Transcripts
NM_004420.2 XM_011519932.2 XM_011519933.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6431836 c.929C>T p.P310L Chr.11: 1557467-1557467 on Build GRCh38
6431645 c.842C>T p.P281L Chr.11: 1557554-1557554 on Build GRCh38
3967439 c.1095G>A p.P365P Chr.11: 1557301-1557301 on Build GRCh38
3445676 c.861C>T p.F287F Chr.11: 1557535-1557535 on Build GRCh38
3445675 c.1008C>T p.T336T Chr.11: 1557388-1557388 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768257298 Chr.11: 1557301-1557301 on Build GRCh38
rs780509893 Chr.11: 1557492-1557492 on Build GRCh38
rs754672813 Chr.11: 1557336-1557336 on Build GRCh38
rs754622728 Chr.11: 1557613-1557662 on Build GRCh38
rs779482451 Chr.11: 1557291-1557291 on Build GRCh38
rs1017293753 Chr.11: 1557651-1557651 on Build GRCh38
rs373215160 Chr.11: 1557508-1557508 on Build GRCh38
rs764293878 Chr.11: 1557412-1557412 on Build GRCh38
rs948747359 Chr.11: 1557579-1557579 on Build GRCh38
rs774637879 Chr.11: 1557413-1557413 on Build GRCh38
rs765066671 Chr.11: 1557580-1557580 on Build GRCh38
rs766369836 Chr.11: 1557467-1557467 on Build GRCh38
rs773418051 Chr.11: 1557544-1557544 on Build GRCh38
rs764391300 Chr.11: 1557325-1557325 on Build GRCh38
rs770485803 Chr.11: 1557437-1557437 on Build GRCh38
rs766237952 Chr.11: 1557337-1557338 on Build GRCh38
rs377156761 Chr.11: 1557484-1557484 on Build GRCh38
rs368626733 Chr.11: 1557235-1557235 on Build GRCh38
rs755331036 Chr.11: 1557466-1557466 on Build GRCh38
rs762369591 Chr.11: 1557585-1557585 on Build GRCh38
rs949214990 Chr.11: 1557319-1557319 on Build GRCh38
rs541590220 Chr.11: 1557274-1557274 on Build GRCh38
rs779617998 Chr.11: 1557583-1557583 on Build GRCh38
rs771327377 Chr.11: 1557361-1557361 on Build GRCh38
rs763115741 Chr.11: 1557405-1557405 on Build GRCh38
rs753693414 Chr.11: 1557578-1557579 on Build GRCh38
rs753776093 Chr.11: 1557468-1557468 on Build GRCh38
rs551148356 Chr.11: 1557606-1557606 on Build GRCh38
rs764823387 Chr.11: 1557473-1557473 on Build GRCh38
rs754263942 Chr.11: 1557372-1557372 on Build GRCh38
rs758411475 Chr.11: 1557616-1557616 on Build GRCh38
rs750180238 Chr.11: 1557421-1557421 on Build GRCh38
rs374762328 Chr.11: 1557220-1557220 on Build GRCh38
rs1007295218 Chr.11: 1557650-1557650 on Build GRCh38
rs780598168 Chr.11: 1557607-1557607 on Build GRCh38
rs72843290 Chr.11: 1557264-1557264 on Build GRCh38
rs528607630 Chr.11: 1557445-1557445 on Build GRCh38
rs770721538 Chr.11: 1557574-1557574 on Build GRCh38
rs558316726 Chr.11: 1557631-1557631 on Build GRCh38
rs768795799 Chr.11: 1557446-1557446 on Build GRCh38
rs757936636 Chr.11: 1557434-1557434 on Build GRCh38
rs771396419 Chr.11: 1557310-1557310 on Build GRCh38
rs759328894 Chr.11: 1557472-1557472 on Build GRCh38
rs748963371 Chr.11: 1557295-1557295 on Build GRCh38
rs770396091 Chr.11: 1557378-1557378 on Build GRCh38
rs761883251 Chr.11: 1557417-1557417 on Build GRCh38
rs776800306 Chr.11: 1557362-1557362 on Build GRCh38
rs754476393 Chr.11: 1557427-1557427 on Build GRCh38
rs745437361 Chr.11: 1557272-1557272 on Build GRCh38
rs765416750 Chr.11: 1557316-1557316 on Build GRCh38
rs372718516 Chr.11: 1557335-1557335 on Build GRCh38
rs753404435 Chr.11: 1557424-1557424 on Build GRCh38
rs760049622 Chr.11: 1557311-1557311 on Build GRCh38
rs189061887 Chr.11: 1557578-1557578 on Build GRCh38
rs756083394 Chr.11: 1557423-1557423 on Build GRCh38
rs1009210849 Chr.11: 1557342-1557342 on Build GRCh38
rs190550919 Chr.11: 1557438-1557438 on Build GRCh38
rs756555115 Chr.11: 1557489-1557489 on Build GRCh38
rs747848347 Chr.11: 1557432-1557432 on Build GRCh38
rs762127578 Chr.11: 1557449-1557449 on Build GRCh38
rs757072093 Chr.11: 1557331-1557331 on Build GRCh38
rs890117456 Chr.11: 1557637-1557637 on Build GRCh38
rs552159665 Chr.11: 1557337-1557337 on Build GRCh38
rs200566171 Chr.11: 1557610-1557610 on Build GRCh38
rs760712584 Chr.11: 1557315-1557316 on Build GRCh38
rs763634422 Chr.11: 1557602-1557602 on Build GRCh38
rs772152722 Chr.11: 1557461-1557461 on Build GRCh38
rs775675442 Chr.11: 1557322-1557322 on Build GRCh38
rs772394327 Chr.11: 1557528-1557528 on Build GRCh38
rs761937966 Chr.11: 1557301-1557302 on Build GRCh38
rs778215704 Chr.11: 1557428-1557428 on Build GRCh38
rs773181723 Chr.11: 1557309-1557310 on Build GRCh38
rs749835351 Chr.11: 1557498-1557498 on Build GRCh38
rs182736278 Chr.11: 1557511-1557511 on Build GRCh38
rs752105174 Chr.11: 1557474-1557474 on Build GRCh38
rs781652418 Chr.11: 1557436-1557436 on Build GRCh38
rs963901940 Chr.11: 1557659-1557659 on Build GRCh38
rs767534995 Chr.11: 1557310-1557318 on Build GRCh38
rs576750165 Chr.11: 1557662-1557662 on Build GRCh38
rs767368858 Chr.11: 1557420-1557420 on Build GRCh38
rs748327126 Chr.11: 1557613-1557613 on Build GRCh38
rs775763578 Chr.11: 1557394-1557394 on Build GRCh38
rs746397157 Chr.11: 1557370-1557370 on Build GRCh38
rs773039099 Chr.11: 1557464-1557464 on Build GRCh38
rs1051332776 Chr.11: 1557632-1557632 on Build GRCh38
rs751372236 Chr.11: 1557327-1557327 on Build GRCh38
rs769005181 Chr.11: 1557500-1557500 on Build GRCh38
rs376022990 Chr.11: 1557603-1557603 on Build GRCh38
rs569581529 Chr.11: 1557609-1557609 on Build GRCh38
rs563264932 Chr.11: 1557323-1557323 on Build GRCh38
rs773490281 Chr.11: 1557308-1557308 on Build GRCh38
rs750888163 Chr.11: 1557486-1557486 on Build GRCh38
rs750387823 Chr.11: 1557332-1557332 on Build GRCh38
rs929308558 Chr.11: 1557254-1557254 on Build GRCh38
rs755998260 Chr.11: 1557334-1557334 on Build GRCh38
rs752312186 Chr.11: 1557584-1557584 on Build GRCh38
rs779272914 Chr.11: 1557611-1557611 on Build GRCh38
rs774248843 Chr.11: 1557448-1557448 on Build GRCh38
rs747396196 Chr.11: 1557344-1557344 on Build GRCh38
rs756645112 Chr.11: 1557558-1557558 on Build GRCh38
rs917316720 Chr.11: 1557298-1557298 on Build GRCh38
rs753120497 Chr.11: 1557553-1557553 on Build GRCh38
rs777280891 Chr.11: 1557485-1557485 on Build GRCh38
rs974090448 Chr.11: 1557458-1557458 on Build GRCh38
rs374530145 Chr.11: 1557253-1557253 on Build GRCh38

View Full Product Details