Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1563720-1564210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP8
Entrez Gene ID
1850
Gene Name
dual specificity phosphatase 8
Gene Aliases
C11orf81, HB5, HVH-5, HVH8
UniGene
Hs.41688
Gene Chromosome Location
Chr.11: 1554051-1572839 on Build GRCh38
Transcripts
NM_004420.2 XM_011519932.2 XM_011519933.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6395475 c.244G>T p.E82* Chr.11: 1563977-1563977 on Build GRCh38
4449895 c.370+1G>A p.? Chr.11: 1563850-1563850 on Build GRCh38
6988787 c.294G>A p.V98V Chr.11: 1563927-1563927 on Build GRCh38
6379615 c.340G>T p.G114C Chr.11: 1563881-1563881 on Build GRCh38
5524875 c.288C>T p.A96A Chr.11: 1563933-1563933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150227968 Chr.11: 1563921-1563921 on Build GRCh38
rs868803301 Chr.11: 1563863-1563863 on Build GRCh38
rs200755114 Chr.11: 1563866-1563866 on Build GRCh38
rs865928413 Chr.11: 1563887-1563887 on Build GRCh38
rs781640826 Chr.11: 1563842-1563842 on Build GRCh38
rs753251957 Chr.11: 1563811-1563811 on Build GRCh38
rs770394046 Chr.11: 1563857-1563857 on Build GRCh38
rs766200658 Chr.11: 1563920-1563920 on Build GRCh38
rs758964260 Chr.11: 1563936-1563936 on Build GRCh38
rs900945069 Chr.11: 1564186-1564186 on Build GRCh38
rs1053664977 Chr.11: 1564117-1564117 on Build GRCh38
rs749740242 Chr.11: 1564003-1564003 on Build GRCh38
rs188919009 Chr.11: 1564115-1564115 on Build GRCh38
rs545757883 Chr.11: 1564034-1564034 on Build GRCh38
rs145547062 Chr.11: 1563966-1563966 on Build GRCh38
rs562169059 Chr.11: 1564129-1564129 on Build GRCh38
rs368821890 Chr.11: 1563844-1563844 on Build GRCh38
rs906987753 Chr.11: 1564155-1564155 on Build GRCh38
rs902456677 Chr.11: 1563881-1563881 on Build GRCh38
rs772858992 Chr.11: 1563780-1563785 on Build GRCh38
rs951808300 Chr.11: 1563741-1563741 on Build GRCh38
rs533450708 Chr.11: 1563743-1563743 on Build GRCh38
rs756742980 Chr.11: 1563999-1563999 on Build GRCh38
rs769065497 Chr.11: 1563846-1563847 on Build GRCh38
rs552663309 Chr.11: 1563882-1563882 on Build GRCh38
rs936580370 Chr.11: 1564100-1564100 on Build GRCh38
rs774444869 Chr.11: 1563874-1563874 on Build GRCh38
rs958987369 Chr.11: 1564022-1564022 on Build GRCh38
rs919413041 Chr.11: 1563995-1563995 on Build GRCh38
rs1048992704 Chr.11: 1564110-1564110 on Build GRCh38
rs570328703 Chr.11: 1563764-1563764 on Build GRCh38
rs3740620 Chr.11: 1564047-1564047 on Build GRCh38
rs556837017 Chr.11: 1564021-1564021 on Build GRCh38
rs902319432 Chr.11: 1564090-1564090 on Build GRCh38
rs908057318 Chr.11: 1563750-1563750 on Build GRCh38
rs141702492 Chr.11: 1563942-1563942 on Build GRCh38
rs984977530 Chr.11: 1564009-1564009 on Build GRCh38
rs752027810 Chr.11: 1563826-1563826 on Build GRCh38
rs765941225 Chr.11: 1563809-1563809 on Build GRCh38
rs552194759 Chr.11: 1563756-1563756 on Build GRCh38
rs750795908 Chr.11: 1563978-1563978 on Build GRCh38
rs927530991 Chr.11: 1563763-1563763 on Build GRCh38
rs772949165 Chr.11: 1563892-1563892 on Build GRCh38
rs760643314 Chr.11: 1563906-1563906 on Build GRCh38
rs929548863 Chr.11: 1563785-1563785 on Build GRCh38
rs548562221 Chr.11: 1564000-1564000 on Build GRCh38
rs573172258 Chr.11: 1564106-1564106 on Build GRCh38
rs764736268 Chr.11: 1563940-1563940 on Build GRCh38
rs779004993 Chr.11: 1564025-1564025 on Build GRCh38
rs377453113 Chr.11: 1564029-1564029 on Build GRCh38
rs1049070929 Chr.11: 1563803-1563803 on Build GRCh38
rs1019266149 Chr.11: 1564171-1564171 on Build GRCh38
rs757699108 Chr.11: 1563833-1563833 on Build GRCh38
rs749666980 Chr.11: 1563818-1563829 on Build GRCh38
rs535708769 Chr.11: 1564096-1564097 on Build GRCh38
rs966572589 Chr.11: 1563755-1563755 on Build GRCh38
rs753639082 Chr.11: 1564031-1564031 on Build GRCh38
rs1004516170 Chr.11: 1564126-1564126 on Build GRCh38
rs1042212535 Chr.11: 1564069-1564069 on Build GRCh38
rs778416111 Chr.11: 1563825-1563825 on Build GRCh38
rs1055583022 Chr.11: 1563843-1563843 on Build GRCh38
rs371561348 Chr.11: 1564106-1564107 on Build GRCh38
rs754704972 Chr.11: 1563812-1563812 on Build GRCh38
rs768994001 Chr.11: 1563872-1563872 on Build GRCh38
rs758073690 Chr.11: 1563873-1563873 on Build GRCh38
rs1006597278 Chr.11: 1564161-1564161 on Build GRCh38
rs946159438 Chr.11: 1563795-1563795 on Build GRCh38
rs746409767 Chr.11: 1563845-1563845 on Build GRCh38
rs373484439 Chr.11: 1563867-1563867 on Build GRCh38
rs760914489 Chr.11: 1563876-1563876 on Build GRCh38
rs896322169 Chr.11: 1563846-1563846 on Build GRCh38
rs755029968 Chr.11: 1564020-1564020 on Build GRCh38
rs758085629 Chr.11: 1563960-1563960 on Build GRCh38
rs944544373 Chr.11: 1564092-1564092 on Build GRCh38
rs900332656 Chr.11: 1564099-1564099 on Build GRCh38
rs1042631931 Chr.11: 1564058-1564058 on Build GRCh38
rs981602337 Chr.11: 1564010-1564010 on Build GRCh38
rs889706005 Chr.11: 1564125-1564125 on Build GRCh38
rs912608209 Chr.11: 1564074-1564074 on Build GRCh38

View Full Product Details