Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1757197-1757704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSD
Entrez Gene ID
1509
Gene Name
cathepsin D
Gene Aliases
CLN10, CPSD, HEL-S-130P
UniGene
Hs.654447
Gene Chromosome Location
Chr.11: 1752752-1763992 on Build GRCh38
Transcripts
NM_001909.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4031859 c.665A>G p.K222R Chr.11: 1757363-1757363 on Build GRCh38
7178636 c.572T>C p.I191T Chr.11: 1757456-1757456 on Build GRCh38
7143866 c.693C>G p.F231L Chr.11: 1757335-1757335 on Build GRCh38
1746158 c.699G>C p.L233L Chr.11: 1757329-1757329 on Build GRCh38
4919010 c.614G>A p.R205H Chr.11: 1757414-1757414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11555045 Chr.11: 1757491-1757491 on Build GRCh38
rs746048174 Chr.11: 1757608-1757608 on Build GRCh38
rs955799601 Chr.11: 1757477-1757477 on Build GRCh38
rs1029438730 Chr.11: 1757633-1757633 on Build GRCh38
rs375282504 Chr.11: 1757398-1757398 on Build GRCh38
rs572376280 Chr.11: 1757493-1757493 on Build GRCh38
rs767282477 Chr.11: 1757454-1757454 on Build GRCh38
rs934263804 Chr.11: 1757624-1757624 on Build GRCh38
rs777479328 Chr.11: 1757295-1757295 on Build GRCh38
rs767476700 Chr.11: 1757539-1757539 on Build GRCh38
rs762797920 Chr.11: 1757368-1757368 on Build GRCh38
rs796052405 Chr.11: 1757467-1757467 on Build GRCh38
rs1031954177 Chr.11: 1757290-1757290 on Build GRCh38
rs200471178 Chr.11: 1757495-1757495 on Build GRCh38
rs138267882 Chr.11: 1757294-1757294 on Build GRCh38
rs764967062 Chr.11: 1757284-1757284 on Build GRCh38
rs577842448 Chr.11: 1757536-1757536 on Build GRCh38
rs567625495 Chr.11: 1757392-1757392 on Build GRCh38
rs371522391 Chr.11: 1757414-1757414 on Build GRCh38
rs1039984157 Chr.11: 1757304-1757304 on Build GRCh38
rs889418730 Chr.11: 1757652-1757652 on Build GRCh38
rs142769806 Chr.11: 1757380-1757380 on Build GRCh38
rs367978332 Chr.11: 1757407-1757407 on Build GRCh38
rs142330967 Chr.11: 1757331-1757331 on Build GRCh38
rs754943635 Chr.11: 1757282-1757282 on Build GRCh38
rs779469223 Chr.11: 1757397-1757397 on Build GRCh38
rs759415169 Chr.11: 1757346-1757346 on Build GRCh38
rs904430565 Chr.11: 1757520-1757520 on Build GRCh38
rs758852382 Chr.11: 1757494-1757494 on Build GRCh38
rs534216994 Chr.11: 1757335-1757335 on Build GRCh38
rs779084661 Chr.11: 1757313-1757313 on Build GRCh38
rs764584849 Chr.11: 1757573-1757573 on Build GRCh38
rs765865119 Chr.11: 1757562-1757562 on Build GRCh38
rs753302848 Chr.11: 1757567-1757567 on Build GRCh38
rs146073498 Chr.11: 1757389-1757389 on Build GRCh38
rs775478289 Chr.11: 1757422-1757422 on Build GRCh38
rs765228737 Chr.11: 1757347-1757347 on Build GRCh38
rs375802296 Chr.11: 1757553-1757553 on Build GRCh38
rs746140722 Chr.11: 1757510-1757510 on Build GRCh38
rs796052404 Chr.11: 1757526-1757526 on Build GRCh38
rs371260793 Chr.11: 1757306-1757306 on Build GRCh38
rs886048066 Chr.11: 1757369-1757369 on Build GRCh38
rs146434428 Chr.11: 1757583-1757583 on Build GRCh38
rs895179132 Chr.11: 1757250-1757250 on Build GRCh38
rs765849046 Chr.11: 1757476-1757476 on Build GRCh38
rs768487717 Chr.11: 1757523-1757523 on Build GRCh38
rs750166391 Chr.11: 1757457-1757457 on Build GRCh38
rs370847523 Chr.11: 1757318-1757318 on Build GRCh38
rs943016868 Chr.11: 1757528-1757528 on Build GRCh38
rs557849768 Chr.11: 1757589-1757589 on Build GRCh38
rs887185230 Chr.11: 1757357-1757357 on Build GRCh38
rs752646929 Chr.11: 1757353-1757353 on Build GRCh38
rs749772016 Chr.11: 1757302-1757302 on Build GRCh38
rs752122454 Chr.11: 1757579-1757579 on Build GRCh38
rs796052396 Chr.11: 1757531-1757531 on Build GRCh38
rs751231176 Chr.11: 1757296-1757296 on Build GRCh38
rs887291391 Chr.11: 1757670-1757670 on Build GRCh38
rs374359096 Chr.11: 1757301-1757301 on Build GRCh38
rs948059177 Chr.11: 1757447-1757447 on Build GRCh38
rs772976217 Chr.11: 1757540-1757540 on Build GRCh38
rs121912789 Chr.11: 1757343-1757343 on Build GRCh38
rs556334193 Chr.11: 1757436-1757436 on Build GRCh38
rs989820816 Chr.11: 1757277-1757277 on Build GRCh38
rs1017047658 Chr.11: 1757667-1757675 on Build GRCh38
rs377613022 Chr.11: 1757297-1757297 on Build GRCh38
rs1320294 Chr.11: 1757443-1757443 on Build GRCh38
rs370750959 Chr.11: 1757276-1757276 on Build GRCh38
rs748547852 Chr.11: 1757317-1757317 on Build GRCh38
rs962992416 Chr.11: 1757678-1757678 on Build GRCh38
rs751418119 Chr.11: 1757440-1757440 on Build GRCh38
rs772453588 Chr.11: 1757401-1757401 on Build GRCh38
rs758874267 Chr.11: 1757571-1757571 on Build GRCh38
rs769825646 Chr.11: 1757415-1757415 on Build GRCh38
rs757688598 Chr.11: 1757501-1757501 on Build GRCh38
rs56363318 Chr.11: 1757266-1757266 on Build GRCh38
rs753209485 Chr.11: 1757486-1757486 on Build GRCh38
rs749196696 Chr.11: 1757518-1757518 on Build GRCh38
rs752549485 Chr.11: 1757289-1757289 on Build GRCh38
rs778031733 Chr.11: 1757406-1757406 on Build GRCh38
rs781382056 Chr.11: 1757509-1757509 on Build GRCh38
rs755670512 Chr.11: 1757470-1757470 on Build GRCh38
rs760929484 Chr.11: 1757323-1757323 on Build GRCh38
rs878877318 Chr.11: 1757322-1757322 on Build GRCh38
rs1057519591 Chr.11: 1757340-1757342 on Build GRCh38
rs72850956 Chr.11: 1757655-1757655 on Build GRCh38
rs765214304 Chr.11: 1757355-1757355 on Build GRCh38
rs763906263 Chr.11: 1757438-1757438 on Build GRCh38
rs924384587 Chr.11: 1757275-1757275 on Build GRCh38
rs113444155 Chr.11: 1757322-1757322 on Build GRCh38
rs775460488 Chr.11: 1757517-1757517 on Build GRCh38
rs201551100 Chr.11: 1757374-1757374 on Build GRCh38
rs774222715 Chr.11: 1757525-1757525 on Build GRCh38
rs756998360 Chr.11: 1757388-1757388 on Build GRCh38

View Full Product Details