Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2377178-2377675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD81
Entrez Gene ID
975
Gene Name
CD81 molecule
Gene Aliases
CVID6, S5.7, TAPA1, TSPAN28
UniGene
Hs.54457
Gene Chromosome Location
Chr.11: 2376177-2397419 on Build GRCh38
Transcripts
NM_001297649.1 NM_004356.3 NR_108080.1

Gene Symbol
CD81-AS1
Entrez Gene ID
101927682
Gene Name
CD81 antisense RNA 1
Gene Aliases
-
UniGene
Hs.675920
Gene Chromosome Location
Chr.11: 2328749-2377992 on Build GRCh38
Transcripts
NM_001297649.1 NM_004356.3 NR_108080.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5838354 c.7G>A p.V3M Chr.11: 2377556-2377556 on Build GRCh38
6461723 c.55T>A p.F19I Chr.11: 2377604-2377604 on Build GRCh38
6132210 c.42C>T p.L14L Chr.11: 2377591-2377591 on Build GRCh38
4441417 c.11A>G p.E4G Chr.11: 2377560-2377560 on Build GRCh38
5999911 c.46G>T p.V16F Chr.11: 2377595-2377595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958113638 Chr.11: 2377234-2377234 on Build GRCh38
rs999507563 Chr.11: 2377197-2377197 on Build GRCh38
rs969707792 Chr.11: 2377350-2377350 on Build GRCh38
rs1025415746 Chr.11: 2377325-2377325 on Build GRCh38
rs1051254776 Chr.11: 2377248-2377248 on Build GRCh38
rs551479968 Chr.11: 2377279-2377279 on Build GRCh38
rs770684347 Chr.11: 2377529-2377529 on Build GRCh38
rs745367425 Chr.11: 2377542-2377542 on Build GRCh38
rs776912412 Chr.11: 2377586-2377586 on Build GRCh38
rs1023517563 Chr.11: 2377348-2377349 on Build GRCh38
rs760244239 Chr.11: 2377219-2377219 on Build GRCh38
rs898689999 Chr.11: 2377310-2377310 on Build GRCh38
rs746838922 Chr.11: 2377523-2377523 on Build GRCh38
rs11546014 Chr.11: 2377380-2377380 on Build GRCh38
rs943007875 Chr.11: 2377264-2377264 on Build GRCh38
rs766578698 Chr.11: 2377576-2377576 on Build GRCh38
rs956364964 Chr.11: 2377403-2377403 on Build GRCh38
rs113952014 Chr.11: 2377616-2377616 on Build GRCh38
rs1013265075 Chr.11: 2377347-2377347 on Build GRCh38
rs533229202 Chr.11: 2377221-2377221 on Build GRCh38
rs896240258 Chr.11: 2377629-2377629 on Build GRCh38
rs942930444 Chr.11: 2377527-2377528 on Build GRCh38
rs570277983 Chr.11: 2377324-2377324 on Build GRCh38
rs868817598 Chr.11: 2377560-2377560 on Build GRCh38
rs183863782 Chr.11: 2377335-2377335 on Build GRCh38
rs769351886 Chr.11: 2377546-2377546 on Build GRCh38
rs764174461 Chr.11: 2377637-2377637 on Build GRCh38
rs376883566 Chr.11: 2377547-2377547 on Build GRCh38
rs988935081 Chr.11: 2377201-2377201 on Build GRCh38
rs956585506 Chr.11: 2377404-2377404 on Build GRCh38
rs781047891 Chr.11: 2377538-2377538 on Build GRCh38
rs913212375 Chr.11: 2377217-2377217 on Build GRCh38
rs761080989 Chr.11: 2377572-2377572 on Build GRCh38
rs755533264 Chr.11: 2377650-2377650 on Build GRCh38
rs890040531 Chr.11: 2377254-2377254 on Build GRCh38
rs757184180 Chr.11: 2377642-2377642 on Build GRCh38
rs532332389 Chr.11: 2377558-2377558 on Build GRCh38
rs969214297 Chr.11: 2377195-2377195 on Build GRCh38
rs751472722 Chr.11: 2377646-2377646 on Build GRCh38
rs759471923 Chr.11: 2377589-2377589 on Build GRCh38
rs751387081 Chr.11: 2377639-2377639 on Build GRCh38
rs986762173 Chr.11: 2377519-2377519 on Build GRCh38
rs988828588 Chr.11: 2377422-2377422 on Build GRCh38
rs979407035 Chr.11: 2377351-2377351 on Build GRCh38
rs568262977 Chr.11: 2377531-2377531 on Build GRCh38
rs562574803 Chr.11: 2377198-2377198 on Build GRCh38
rs777743175 Chr.11: 2377630-2377630 on Build GRCh38
rs752534348 Chr.11: 2377609-2377609 on Build GRCh38
rs1038937894 Chr.11: 2377297-2377297 on Build GRCh38
rs865925799 Chr.11: 2377401-2377401 on Build GRCh38
rs913328510 Chr.11: 2377440-2377440 on Build GRCh38
rs758323650 Chr.11: 2377633-2377633 on Build GRCh38
rs745547670 Chr.11: 2377645-2377645 on Build GRCh38
rs772547927 Chr.11: 2377656-2377656 on Build GRCh38
rs1033047881 Chr.11: 2377487-2377508 on Build GRCh38
rs748792416 Chr.11: 2377653-2377653 on Build GRCh38
rs1046814935 Chr.11: 2377598-2377598 on Build GRCh38
rs773538306 Chr.11: 2377567-2377567 on Build GRCh38
rs1032571810 Chr.11: 2377376-2377376 on Build GRCh38
rs748713554 Chr.11: 2377549-2377549 on Build GRCh38
rs987830496 Chr.11: 2377550-2377550 on Build GRCh38
rs765378101 Chr.11: 2377591-2377591 on Build GRCh38

View Full Product Details