Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 4572065-4572464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf40
Entrez Gene ID
143501
Gene Name
chromosome 11 open reading frame 40
Gene Aliases
NOV1
UniGene
Hs.350556
Gene Chromosome Location
Chr.11: 4571423-4577820 on Build GRCh38
Transcripts
NM_144663.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5904943 c.422C>T p.T141M Chr.11: 4572181-4572181 on Build GRCh38
1298049 c.344C>G p.S115C Chr.11: 4572259-4572259 on Build GRCh38
1246384 c.309-2A>C p.? Chr.11: 4572296-4572296 on Build GRCh38
4619959 c.427T>G p.S143A Chr.11: 4572176-4572176 on Build GRCh38
1703369 c.371C>A p.P124H Chr.11: 4572232-4572232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776452190 Chr.11: 4572218-4572218 on Build GRCh38
rs560276940 Chr.11: 4572347-4572347 on Build GRCh38
rs147079995 Chr.11: 4572302-4572302 on Build GRCh38
rs770688761 Chr.11: 4572256-4572257 on Build GRCh38
rs368693067 Chr.11: 4572343-4572343 on Build GRCh38
rs538808326 Chr.11: 4572171-4572171 on Build GRCh38
rs773161983 Chr.11: 4572240-4572241 on Build GRCh38
rs780565366 Chr.11: 4572314-4572315 on Build GRCh38
rs199833753 Chr.11: 4572214-4572214 on Build GRCh38
rs780143565 Chr.11: 4572133-4572133 on Build GRCh38
rs766155176 Chr.11: 4572123-4572123 on Build GRCh38
rs762402556 Chr.11: 4572298-4572303 on Build GRCh38
rs929992179 Chr.11: 4572420-4572420 on Build GRCh38
rs749520788 Chr.11: 4572135-4572135 on Build GRCh38
rs758976826 Chr.11: 4572124-4572124 on Build GRCh38
rs774221852 Chr.11: 4572107-4572107 on Build GRCh38
rs537491545 Chr.11: 4572137-4572137 on Build GRCh38
rs116461440 Chr.11: 4572344-4572344 on Build GRCh38
rs760562855 Chr.11: 4572161-4572161 on Build GRCh38
rs74052438 Chr.11: 4572270-4572270 on Build GRCh38
rs571488877 Chr.11: 4572169-4572169 on Build GRCh38
rs147840296 Chr.11: 4572191-4572191 on Build GRCh38
rs754262122 Chr.11: 4572254-4572254 on Build GRCh38
rs770759449 Chr.11: 4572246-4572246 on Build GRCh38
rs777860286 Chr.11: 4572207-4572207 on Build GRCh38
rs780939837 Chr.11: 4572277-4572277 on Build GRCh38
rs755250998 Chr.11: 4572256-4572256 on Build GRCh38
rs368340095 Chr.11: 4572249-4572249 on Build GRCh38
rs755198534 Chr.11: 4572193-4572193 on Build GRCh38
rs778964103 Chr.11: 4572202-4572202 on Build GRCh38
rs756727411 Chr.11: 4572312-4572313 on Build GRCh38
rs781765780 Chr.11: 4572133-4572137 on Build GRCh38
rs780712274 Chr.11: 4572127-4572127 on Build GRCh38
rs143495598 Chr.11: 4572131-4572131 on Build GRCh38
rs201154177 Chr.11: 4572221-4572221 on Build GRCh38
rs759057061 Chr.11: 4572258-4572259 on Build GRCh38
rs201392052 Chr.11: 4572122-4572122 on Build GRCh38
rs779040197 Chr.11: 4572206-4572206 on Build GRCh38
rs1047624896 Chr.11: 4572421-4572421 on Build GRCh38
rs745692906 Chr.11: 4572399-4572399 on Build GRCh38
rs771975177 Chr.11: 4572230-4572231 on Build GRCh38
rs769507078 Chr.11: 4572233-4572233 on Build GRCh38
rs767470496 Chr.11: 4572113-4572113 on Build GRCh38
rs950103534 Chr.11: 4572304-4572304 on Build GRCh38
rs779793730 Chr.11: 4572346-4572346 on Build GRCh38
rs867512390 Chr.11: 4572271-4572271 on Build GRCh38
rs770696113 Chr.11: 4572163-4572163 on Build GRCh38
rs752185330 Chr.11: 4572294-4572295 on Build GRCh38
rs765680341 Chr.11: 4572305-4572305 on Build GRCh38
rs374235805 Chr.11: 4572140-4572140 on Build GRCh38
rs375331291 Chr.11: 4572117-4572117 on Build GRCh38
rs370834667 Chr.11: 4572112-4572112 on Build GRCh38
rs756571197 Chr.11: 4572135-4572136 on Build GRCh38
rs772169929 Chr.11: 4572160-4572160 on Build GRCh38
rs190317123 Chr.11: 4572212-4572212 on Build GRCh38
rs754217513 Chr.11: 4572321-4572325 on Build GRCh38
rs551592858 Chr.11: 4572101-4572101 on Build GRCh38
rs375290812 Chr.11: 4572240-4572240 on Build GRCh38
rs757219876 Chr.11: 4572333-4572333 on Build GRCh38
rs757727303 Chr.11: 4572129-4572129 on Build GRCh38
rs775189134 Chr.11: 4572278-4572278 on Build GRCh38
rs549543059 Chr.11: 4572403-4572403 on Build GRCh38
rs757724865 Chr.11: 4572132-4572133 on Build GRCh38
rs772223493 Chr.11: 4572206-4572206 on Build GRCh38
rs1019116463 Chr.11: 4572360-4572360 on Build GRCh38
rs776217370 Chr.11: 4572164-4572164 on Build GRCh38
rs750779496 Chr.11: 4572133-4572134 on Build GRCh38
rs759337115 Chr.11: 4572273-4572273 on Build GRCh38
rs367892257 Chr.11: 4572130-4572130 on Build GRCh38
rs139274381 Chr.11: 4572147-4572147 on Build GRCh38
rs752232833 Chr.11: 4572176-4572176 on Build GRCh38
rs751348805 Chr.11: 4572204-4572204 on Build GRCh38
rs993608109 Chr.11: 4572367-4572367 on Build GRCh38
rs766695586 Chr.11: 4572253-4572253 on Build GRCh38
rs755166150 Chr.11: 4572327-4572328 on Build GRCh38
rs756632416 Chr.11: 4572120-4572120 on Build GRCh38
rs768714323 Chr.11: 4572106-4572106 on Build GRCh38
rs748138676 Chr.11: 4572156-4572156 on Build GRCh38
rs141736352 Chr.11: 4572248-4572248 on Build GRCh38
rs142163926 Chr.11: 4572185-4572185 on Build GRCh38
rs185019050 Chr.11: 4572168-4572168 on Build GRCh38
rs937819428 Chr.11: 4572288-4572288 on Build GRCh38
rs752052081 Chr.11: 4572128-4572128 on Build GRCh38
rs895820447 Chr.11: 4572172-4572172 on Build GRCh38
rs780616664 Chr.11: 4572139-4572140 on Build GRCh38
rs949919905 Chr.11: 4572439-4572439 on Build GRCh38
rs768195807 Chr.11: 4572284-4572284 on Build GRCh38
rs759158453 Chr.11: 4572117-4572118 on Build GRCh38
rs768831329 Chr.11: 4572195-4572196 on Build GRCh38
rs779002853 Chr.11: 4572329-4572330 on Build GRCh38
rs749497906 Chr.11: 4572176-4572177 on Build GRCh38
rs911530114 Chr.11: 4572096-4572096 on Build GRCh38
rs748935092 Chr.11: 4572281-4572281 on Build GRCh38
rs757246008 Chr.11: 4572272-4572272 on Build GRCh38
rs560860451 Chr.11: 4572397-4572397 on Build GRCh38
rs764786671 Chr.11: 4572125-4572125 on Build GRCh38
rs531321363 Chr.11: 4572400-4572400 on Build GRCh38
rs773828736 Chr.11: 4572286-4572286 on Build GRCh38
rs895761931 Chr.11: 4572438-4572438 on Build GRCh38
rs527884383 Chr.11: 4572355-4572355 on Build GRCh38
rs770676875 Chr.11: 4572217-4572217 on Build GRCh38
rs539471342 Chr.11: 4572366-4572366 on Build GRCh38
rs141444146 Chr.11: 4572259-4572259 on Build GRCh38
rs752215365 Chr.11: 4572120-4572121 on Build GRCh38
rs560762655 Chr.11: 4572257-4572257 on Build GRCh38
rs1025960840 Chr.11: 4572368-4572368 on Build GRCh38
rs760410431 Chr.11: 4572242-4572242 on Build GRCh38
rs777990974 Chr.11: 4572261-4572261 on Build GRCh38
rs746684882 Chr.11: 4572239-4572239 on Build GRCh38
rs918566143 Chr.11: 4572419-4572419 on Build GRCh38
rs750861917 Chr.11: 4572184-4572184 on Build GRCh38
rs766939374 Chr.11: 4572292-4572292 on Build GRCh38
rs117631969 Chr.11: 4572320-4572320 on Build GRCh38
rs764810916 Chr.11: 4572290-4572293 on Build GRCh38
rs759221796 Chr.11: 4572286-4572287 on Build GRCh38
rs201728077 Chr.11: 4572224-4572224 on Build GRCh38
rs146027716 Chr.11: 4572188-4572188 on Build GRCh38
rs34097396 Chr.11: 4572181-4572181 on Build GRCh38
rs375834537 Chr.11: 4572327-4572327 on Build GRCh38
rs142861848 Chr.11: 4572180-4572180 on Build GRCh38
rs768090211 Chr.11: 4572305-4572306 on Build GRCh38
rs753030729 Chr.11: 4572313-4572313 on Build GRCh38
rs796502297 Chr.11: 4572117-4572118 on Build GRCh38
rs369393694 Chr.11: 4572321-4572321 on Build GRCh38
rs34520159 Chr.11: 4572104-4572104 on Build GRCh38
rs372723586 Chr.11: 4572326-4572326 on Build GRCh38
rs376669062 Chr.11: 4572345-4572345 on Build GRCh38
rs16915110 Chr.11: 4572359-4572359 on Build GRCh38
rs571246272 Chr.11: 4572432-4572432 on Build GRCh38
rs368139062 Chr.11: 4572330-4572330 on Build GRCh38
rs748187138 Chr.11: 4572223-4572224 on Build GRCh38
rs750946786 Chr.11: 4572309-4572311 on Build GRCh38
rs1041711772 Chr.11: 4572318-4572318 on Build GRCh38
rs951990554 Chr.11: 4572389-4572389 on Build GRCh38

View Full Product Details