Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6211256-6211766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf42
Entrez Gene ID
160298
Gene Name
chromosome 11 open reading frame 42
Gene Aliases
-
UniGene
Hs.278221
Gene Chromosome Location
Chr.11: 6202997-6226783 on Build GRCh38
Transcripts
XM_011519926.2 NM_001098794.1 NM_032127.3 XM_006718343.2 XM_011520397.2

Gene Symbol
FAM160A2
Entrez Gene ID
84067
Gene Name
family with sequence similarity 160 member A2
Gene Aliases
C11orf56
UniGene
Hs.501793
Gene Chromosome Location
Chr.11: 6211334-6234711 on Build GRCh38
Transcripts
XM_011519926.2 NM_001098794.1 NM_032127.3 XM_006718343.2 XM_011520397.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
929686 c.2783G>A p.R928H Chr.11: 6211684-6211684 on Build GRCh38
929683 c.2950C>A p.L984I Chr.11: 6211517-6211517 on Build GRCh38
3739182 c.2881G>A p.V961I Chr.11: 6211586-6211586 on Build GRCh38
5373082 c.2733C>T p.S911S Chr.11: 6211734-6211734 on Build GRCh38
5368946 c.2950C>T p.L984F Chr.11: 6211517-6211517 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049434702 Chr.11: 6211650-6211650 on Build GRCh38
rs764004096 Chr.11: 6211461-6211461 on Build GRCh38
rs143871240 Chr.11: 6211699-6211699 on Build GRCh38
rs375426902 Chr.11: 6211469-6211469 on Build GRCh38
rs553286788 Chr.11: 6211347-6211347 on Build GRCh38
rs777129200 Chr.11: 6211296-6211296 on Build GRCh38
rs763280190 Chr.11: 6211719-6211719 on Build GRCh38
rs555544898 Chr.11: 6211323-6211323 on Build GRCh38
rs373859617 Chr.11: 6211720-6211720 on Build GRCh38
rs751914391 Chr.11: 6211647-6211647 on Build GRCh38
rs755544760 Chr.11: 6211679-6211679 on Build GRCh38
rs755451478 Chr.11: 6211732-6211732 on Build GRCh38
rs887721015 Chr.11: 6211652-6211652 on Build GRCh38
rs750468481 Chr.11: 6211707-6211707 on Build GRCh38
rs370304342 Chr.11: 6211738-6211738 on Build GRCh38
rs139429864 Chr.11: 6211703-6211703 on Build GRCh38
rs780536778 Chr.11: 6211505-6211505 on Build GRCh38
rs757507679 Chr.11: 6211489-6211489 on Build GRCh38
rs761780365 Chr.11: 6211704-6211704 on Build GRCh38
rs544117611 Chr.11: 6211735-6211735 on Build GRCh38
rs202217305 Chr.11: 6211637-6211637 on Build GRCh38
rs751827058 Chr.11: 6211725-6211725 on Build GRCh38
rs140136348 Chr.11: 6211697-6211697 on Build GRCh38
rs557522108 Chr.11: 6211461-6211461 on Build GRCh38
rs575055797 Chr.11: 6211385-6211385 on Build GRCh38
rs753774271 Chr.11: 6211578-6211578 on Build GRCh38
rs200445220 Chr.11: 6211737-6211737 on Build GRCh38
rs962602514 Chr.11: 6211443-6211443 on Build GRCh38
rs375970911 Chr.11: 6211569-6211569 on Build GRCh38
rs202015594 Chr.11: 6211742-6211742 on Build GRCh38
rs910932360 Chr.11: 6211297-6211297 on Build GRCh38
rs986482905 Chr.11: 6211283-6211283 on Build GRCh38
rs750647959 Chr.11: 6211498-6211498 on Build GRCh38
rs79511779 Chr.11: 6211462-6211462 on Build GRCh38
rs759201179 Chr.11: 6211541-6211541 on Build GRCh38
rs778451280 Chr.11: 6211698-6211698 on Build GRCh38
rs1002791588 Chr.11: 6211397-6211397 on Build GRCh38
rs750652228 Chr.11: 6211599-6211599 on Build GRCh38
rs201893041 Chr.11: 6211522-6211522 on Build GRCh38
rs185826903 Chr.11: 6211311-6211311 on Build GRCh38
rs1041946429 Chr.11: 6211328-6211328 on Build GRCh38
rs767327639 Chr.11: 6211467-6211467 on Build GRCh38
rs540613056 Chr.11: 6211620-6211620 on Build GRCh38
rs758739849 Chr.11: 6211502-6211502 on Build GRCh38
rs201012137 Chr.11: 6211685-6211685 on Build GRCh38
rs761860531 Chr.11: 6211587-6211587 on Build GRCh38
rs867323964 Chr.11: 6211531-6211531 on Build GRCh38
rs1017161820 Chr.11: 6211437-6211437 on Build GRCh38
rs9659 Chr.11: 6211423-6211423 on Build GRCh38
rs546272121 Chr.11: 6211520-6211520 on Build GRCh38
rs535004569 Chr.11: 6211342-6211342 on Build GRCh38
rs377652633 Chr.11: 6211700-6211700 on Build GRCh38
rs769236235 Chr.11: 6211513-6211513 on Build GRCh38
rs994419840 Chr.11: 6211453-6211453 on Build GRCh38
rs13899 Chr.11: 6211561-6211561 on Build GRCh38
rs897707967 Chr.11: 6211282-6211282 on Build GRCh38
rs369707658 Chr.11: 6211470-6211470 on Build GRCh38
rs1006345731 Chr.11: 6211424-6211424 on Build GRCh38
rs752536693 Chr.11: 6211308-6211308 on Build GRCh38
rs893340897 Chr.11: 6211318-6211318 on Build GRCh38
rs765338857 Chr.11: 6211590-6211590 on Build GRCh38
rs867024680 Chr.11: 6211445-6211445 on Build GRCh38
rs370074542 Chr.11: 6211680-6211680 on Build GRCh38
rs1003000198 Chr.11: 6211393-6211393 on Build GRCh38
rs946280348 Chr.11: 6211320-6211320 on Build GRCh38
rs200174554 Chr.11: 6211667-6211667 on Build GRCh38
rs910922546 Chr.11: 6211472-6211472 on Build GRCh38
rs779076100 Chr.11: 6211494-6211494 on Build GRCh38
rs762829247 Chr.11: 6211407-6211407 on Build GRCh38
rs765264764 Chr.11: 6211705-6211705 on Build GRCh38
rs146871349 Chr.11: 6211702-6211702 on Build GRCh38
rs993373925 Chr.11: 6211288-6211288 on Build GRCh38
rs573240563 Chr.11: 6211558-6211558 on Build GRCh38
rs989937217 Chr.11: 6211523-6211523 on Build GRCh38
rs759314570 Chr.11: 6211466-6211466 on Build GRCh38
rs564494787 Chr.11: 6211528-6211528 on Build GRCh38
rs753859976 Chr.11: 6211477-6211477 on Build GRCh38
rs771892412 Chr.11: 6211554-6211554 on Build GRCh38
rs61876694 Chr.11: 6211364-6211364 on Build GRCh38
rs747552046 Chr.11: 6211510-6211510 on Build GRCh38

View Full Product Details