Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6218478-6218985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf42
Entrez Gene ID
160298
Gene Name
chromosome 11 open reading frame 42
Gene Aliases
-
UniGene
Hs.278221
Gene Chromosome Location
Chr.11: 6202997-6226783 on Build GRCh38
Transcripts
XM_011519926.2 NM_001098794.1 NM_032127.3 XM_006718343.2 XM_011520397.2 XM_011520398.2 XM_011520399.2

Gene Symbol
FAM160A2
Entrez Gene ID
84067
Gene Name
family with sequence similarity 160 member A2
Gene Aliases
C11orf56
UniGene
Hs.501793
Gene Chromosome Location
Chr.11: 6211334-6234711 on Build GRCh38
Transcripts
XM_011519926.2 NM_001098794.1 NM_032127.3 XM_006718343.2 XM_011520397.2 XM_011520398.2 XM_011520399.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1355328 c.1460G>A p.R487K Chr.11: 6218575-6218575 on Build GRCh38
3700112 c.1294G>T p.V432F Chr.11: 6218741-6218741 on Build GRCh38
6658716 c.1410T>A p.R470R Chr.11: 6218625-6218625 on Build GRCh38
4995016 c.1372C>T p.P458S Chr.11: 6218663-6218663 on Build GRCh38
4943817 c.1283C>T p.P428L Chr.11: 6218752-6218752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372584975 Chr.11: 6218941-6218941 on Build GRCh38
rs147077635 Chr.11: 6218642-6218642 on Build GRCh38
rs934448829 Chr.11: 6218520-6218520 on Build GRCh38
rs397794265 Chr.11: 6218511-6218511 on Build GRCh38
rs771051126 Chr.11: 6218522-6218522 on Build GRCh38
rs756132827 Chr.11: 6218943-6218943 on Build GRCh38
rs3833754 Chr.11: 6218510-6218510 on Build GRCh38
rs114329486 Chr.11: 6218722-6218722 on Build GRCh38
rs770210129 Chr.11: 6218510-6218510 on Build GRCh38
rs35932378 Chr.11: 6218651-6218651 on Build GRCh38
rs762522136 Chr.11: 6218688-6218688 on Build GRCh38
rs376903758 Chr.11: 6218514-6218514 on Build GRCh38
rs777821303 Chr.11: 6218640-6218640 on Build GRCh38
rs144402782 Chr.11: 6218610-6218610 on Build GRCh38
rs544785370 Chr.11: 6218631-6218631 on Build GRCh38
rs899422799 Chr.11: 6218586-6218586 on Build GRCh38
rs191657277 Chr.11: 6218584-6218584 on Build GRCh38
rs551678500 Chr.11: 6218896-6218896 on Build GRCh38
rs761121887 Chr.11: 6218664-6218664 on Build GRCh38
rs899412933 Chr.11: 6218745-6218745 on Build GRCh38
rs746982734 Chr.11: 6218573-6218573 on Build GRCh38
rs780339823 Chr.11: 6218776-6218776 on Build GRCh38
rs749392430 Chr.11: 6218724-6218724 on Build GRCh38
rs968714301 Chr.11: 6218669-6218669 on Build GRCh38
rs761335206 Chr.11: 6218692-6218693 on Build GRCh38
rs201798905 Chr.11: 6218956-6218956 on Build GRCh38
rs750055025 Chr.11: 6218559-6218559 on Build GRCh38
rs772531889 Chr.11: 6218682-6218682 on Build GRCh38
rs1019466784 Chr.11: 6218638-6218638 on Build GRCh38
rs769220491 Chr.11: 6218797-6218797 on Build GRCh38
rs983055654 Chr.11: 6218900-6218900 on Build GRCh38
rs768077599 Chr.11: 6218614-6218614 on Build GRCh38
rs367742077 Chr.11: 6218646-6218646 on Build GRCh38
rs533593944 Chr.11: 6218841-6218841 on Build GRCh38
rs749535032 Chr.11: 6218650-6218650 on Build GRCh38
rs766008071 Chr.11: 6218709-6218709 on Build GRCh38
rs756094983 Chr.11: 6218713-6218713 on Build GRCh38
rs747520526 Chr.11: 6218795-6218795 on Build GRCh38
rs781274931 Chr.11: 6218593-6218593 on Build GRCh38
rs758371519 Chr.11: 6218617-6218617 on Build GRCh38
rs529633386 Chr.11: 6218714-6218714 on Build GRCh38
rs756754195 Chr.11: 6218548-6218548 on Build GRCh38
rs527738340 Chr.11: 6218939-6218939 on Build GRCh38
rs149631608 Chr.11: 6218598-6218598 on Build GRCh38
rs775962246 Chr.11: 6218619-6218619 on Build GRCh38
rs759909208 Chr.11: 6218539-6218539 on Build GRCh38
rs34767365 Chr.11: 6218673-6218673 on Build GRCh38
rs965204786 Chr.11: 6218639-6218639 on Build GRCh38
rs779666278 Chr.11: 6218570-6218570 on Build GRCh38
rs760540227 Chr.11: 6218957-6218957 on Build GRCh38
rs762422743 Chr.11: 6218808-6218808 on Build GRCh38
rs772902652 Chr.11: 6218924-6218925 on Build GRCh38
rs138279096 Chr.11: 6218560-6218560 on Build GRCh38
rs746070487 Chr.11: 6218753-6218753 on Build GRCh38
rs746274335 Chr.11: 6218658-6218658 on Build GRCh38
rs1056532375 Chr.11: 6218659-6218659 on Build GRCh38
rs1048461312 Chr.11: 6218793-6218793 on Build GRCh38
rs12363184 Chr.11: 6218924-6218924 on Build GRCh38
rs766203455 Chr.11: 6218634-6218634 on Build GRCh38
rs777185828 Chr.11: 6218806-6218806 on Build GRCh38
rs922797096 Chr.11: 6218860-6218860 on Build GRCh38
rs886419539 Chr.11: 6218831-6218831 on Build GRCh38
rs996635541 Chr.11: 6218893-6218893 on Build GRCh38
rs752701282 Chr.11: 6218717-6218717 on Build GRCh38
rs767429737 Chr.11: 6218944-6218944 on Build GRCh38
rs745993384 Chr.11: 6218913-6218913 on Build GRCh38
rs566994676 Chr.11: 6218898-6218898 on Build GRCh38
rs751096154 Chr.11: 6218580-6218580 on Build GRCh38
rs1018997577 Chr.11: 6218855-6218855 on Build GRCh38
rs995116704 Chr.11: 6218771-6218771 on Build GRCh38
rs368609593 Chr.11: 6218742-6218742 on Build GRCh38
rs749427194 Chr.11: 6218603-6218603 on Build GRCh38
rs368281359 Chr.11: 6218951-6218951 on Build GRCh38
rs762133260 Chr.11: 6218509-6218509 on Build GRCh38
rs753277158 Chr.11: 6218543-6218543 on Build GRCh38
rs375612041 Chr.11: 6218721-6218721 on Build GRCh38
rs761361147 Chr.11: 6218626-6218626 on Build GRCh38
rs370073421 Chr.11: 6218780-6218780 on Build GRCh38
rs774229199 Chr.11: 6218592-6218593 on Build GRCh38
rs373113703 Chr.11: 6218533-6218533 on Build GRCh38
rs772448189 Chr.11: 6218766-6218766 on Build GRCh38
rs113571470 Chr.11: 6218534-6218534 on Build GRCh38
rs757345885 Chr.11: 6218741-6218741 on Build GRCh38
rs770448371 Chr.11: 6218921-6218921 on Build GRCh38
rs771345955 Chr.11: 6218606-6218606 on Build GRCh38
rs771728032 Chr.11: 6218791-6218792 on Build GRCh38
rs778593475 Chr.11: 6218553-6218553 on Build GRCh38
rs1009317369 Chr.11: 6218840-6218840 on Build GRCh38
rs760091862 Chr.11: 6218613-6218613 on Build GRCh38
rs763605148 Chr.11: 6218540-6218540 on Build GRCh38
rs773604505 Chr.11: 6218601-6218601 on Build GRCh38
rs964800541 Chr.11: 6218876-6218876 on Build GRCh38
rs77726581 Chr.11: 6218660-6218660 on Build GRCh38
rs748300245 Chr.11: 6218595-6218595 on Build GRCh38
rs763645071 Chr.11: 6218575-6218575 on Build GRCh38
rs199767800 Chr.11: 6218648-6218648 on Build GRCh38
rs375084452 Chr.11: 6218535-6218535 on Build GRCh38
rs560672879 Chr.11: 6218670-6218670 on Build GRCh38
rs773559494 Chr.11: 6218516-6218516 on Build GRCh38
rs754810687 Chr.11: 6218643-6218643 on Build GRCh38
rs994831489 Chr.11: 6218602-6218602 on Build GRCh38
rs753569407 Chr.11: 6218894-6218894 on Build GRCh38
rs141051752 Chr.11: 6218627-6218627 on Build GRCh38
rs948830987 Chr.11: 6218733-6218733 on Build GRCh38
rs144607645 Chr.11: 6218649-6218649 on Build GRCh38

View Full Product Details