Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6239596-6240087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGA4
Entrez Gene ID
1262
Gene Name
cyclic nucleotide gated channel alpha 4
Gene Aliases
CNCA2, CNG-4, CNG4, CNG5, CNGB2, OCNC2, OCNCBETA, OCNCb
UniGene
Hs.434618
Gene Chromosome Location
Chr.11: 6234774-6245924 on Build GRCh38
Transcripts
NM_001037329.3 XM_017017217.1 XM_017017218.1 XM_017017219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3809889 c.190G>A p.G64S Chr.11: 6239709-6239709 on Build GRCh38
5477743 c.263T>C p.F88S Chr.11: 6239782-6239782 on Build GRCh38
5492743 c.217G>A p.D73N Chr.11: 6239736-6239736 on Build GRCh38
5739155 c.248A>G p.D83G Chr.11: 6239767-6239767 on Build GRCh38
1248536 c.234G>A p.L78L Chr.11: 6239753-6239753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042052556 Chr.11: 6239983-6239983 on Build GRCh38
rs773230575 Chr.11: 6240015-6240015 on Build GRCh38
rs753395347 Chr.11: 6239708-6239708 on Build GRCh38
rs911038935 Chr.11: 6239662-6239662 on Build GRCh38
rs367747927 Chr.11: 6240039-6240039 on Build GRCh38
rs759955658 Chr.11: 6239709-6239709 on Build GRCh38
rs547829254 Chr.11: 6239630-6239630 on Build GRCh38
rs868732203 Chr.11: 6239782-6239782 on Build GRCh38
rs773492976 Chr.11: 6239696-6239696 on Build GRCh38
rs780893905 Chr.11: 6240061-6240061 on Build GRCh38
rs781760773 Chr.11: 6239637-6239637 on Build GRCh38
rs758057217 Chr.11: 6239773-6239773 on Build GRCh38
rs887430611 Chr.11: 6239863-6239863 on Build GRCh38
rs958895332 Chr.11: 6239872-6239872 on Build GRCh38
rs771106065 Chr.11: 6240023-6240023 on Build GRCh38
rs769812039 Chr.11: 6239684-6239684 on Build GRCh38
rs558605371 Chr.11: 6239932-6239932 on Build GRCh38
rs1016441301 Chr.11: 6239882-6239882 on Build GRCh38
rs757906465 Chr.11: 6240055-6240055 on Build GRCh38
rs752626383 Chr.11: 6239904-6239904 on Build GRCh38
rs375093341 Chr.11: 6240020-6240020 on Build GRCh38
rs761908936 Chr.11: 6239681-6239681 on Build GRCh38
rs776515867 Chr.11: 6239679-6239679 on Build GRCh38
rs746858122 Chr.11: 6239778-6239778 on Build GRCh38
rs906866901 Chr.11: 6239995-6239995 on Build GRCh38
rs936761433 Chr.11: 6239931-6239931 on Build GRCh38
rs914594085 Chr.11: 6239654-6239654 on Build GRCh38
rs764594438 Chr.11: 6239743-6239743 on Build GRCh38
rs746805196 Chr.11: 6239663-6239663 on Build GRCh38
rs1054225493 Chr.11: 6239730-6239730 on Build GRCh38
rs201459029 Chr.11: 6239635-6239635 on Build GRCh38
rs1031924719 Chr.11: 6239901-6239902 on Build GRCh38
rs146113879 Chr.11: 6239846-6239846 on Build GRCh38
rs923283916 Chr.11: 6239895-6239895 on Build GRCh38
rs370908460 Chr.11: 6239638-6239638 on Build GRCh38
rs749963340 Chr.11: 6240054-6240054 on Build GRCh38
rs558890020 Chr.11: 6240021-6240021 on Build GRCh38
rs769921335 Chr.11: 6239837-6239837 on Build GRCh38
rs370277883 Chr.11: 6239697-6239697 on Build GRCh38
rs370207927 Chr.11: 6240064-6240064 on Build GRCh38
rs751160947 Chr.11: 6240060-6240060 on Build GRCh38
rs538838063 Chr.11: 6239893-6239893 on Build GRCh38
rs954026550 Chr.11: 6239943-6239943 on Build GRCh38
rs746311816 Chr.11: 6240005-6240005 on Build GRCh38
rs781126794 Chr.11: 6239789-6239789 on Build GRCh38
rs538955568 Chr.11: 6239799-6239799 on Build GRCh38
rs111621831 Chr.11: 6239987-6239987 on Build GRCh38
rs1034554219 Chr.11: 6239797-6239797 on Build GRCh38
rs536669512 Chr.11: 6240003-6240003 on Build GRCh38
rs972322069 Chr.11: 6239894-6239894 on Build GRCh38
rs538115869 Chr.11: 6239814-6239814 on Build GRCh38
rs114651071 Chr.11: 6239879-6239879 on Build GRCh38
rs760011098 Chr.11: 6240035-6240035 on Build GRCh38
rs1022721239 Chr.11: 6239891-6239891 on Build GRCh38
rs756851204 Chr.11: 6239731-6239731 on Build GRCh38
rs112963161 Chr.11: 6240010-6240010 on Build GRCh38
rs750099398 Chr.11: 6239744-6239744 on Build GRCh38
rs181952007 Chr.11: 6239928-6239928 on Build GRCh38
rs370664809 Chr.11: 6239775-6239775 on Build GRCh38
rs765243118 Chr.11: 6239892-6239892 on Build GRCh38
rs1041355168 Chr.11: 6239748-6239748 on Build GRCh38
rs945956870 Chr.11: 6239955-6239955 on Build GRCh38
rs906894297 Chr.11: 6239769-6239769 on Build GRCh38
rs745462263 Chr.11: 6239648-6239648 on Build GRCh38
rs753216603 Chr.11: 6239720-6239720 on Build GRCh38
rs776154096 Chr.11: 6240040-6240040 on Build GRCh38
rs923281179 Chr.11: 6239629-6239629 on Build GRCh38
rs758929746 Chr.11: 6239860-6239860 on Build GRCh38
rs771329884 Chr.11: 6239620-6239620 on Build GRCh38
rs577153328 Chr.11: 6240030-6240030 on Build GRCh38
rs756798204 Chr.11: 6239642-6239642 on Build GRCh38
rs554631077 Chr.11: 6240007-6240007 on Build GRCh38
rs575434638 Chr.11: 6239888-6239888 on Build GRCh38
rs766591273 Chr.11: 6239634-6239634 on Build GRCh38
rs768640369 Chr.11: 6239676-6239676 on Build GRCh38
rs771918312 Chr.11: 6239651-6239651 on Build GRCh38
rs986229009 Chr.11: 6239911-6239911 on Build GRCh38
rs779650308 Chr.11: 6239652-6239652 on Build GRCh38
rs761201112 Chr.11: 6240046-6240046 on Build GRCh38
rs778659850 Chr.11: 6239645-6239645 on Build GRCh38
rs558059573 Chr.11: 6239940-6239940 on Build GRCh38
rs768180337 Chr.11: 6239715-6239715 on Build GRCh38
rs754725951 Chr.11: 6239779-6239779 on Build GRCh38
rs764661626 Chr.11: 6240048-6240048 on Build GRCh38
rs765977093 Chr.11: 6240056-6240056 on Build GRCh38
rs776140163 Chr.11: 6240058-6240059 on Build GRCh38

View Full Product Details