Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6508789-6509293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNHD1
Entrez Gene ID
144132
Gene Name
dynein heavy chain domain 1
Gene Aliases
C11orf47, CCDC35, DHCD1, DNHD1L
UniGene
Hs.720080
Gene Chromosome Location
Chr.11: 6497296-6572025 on Build GRCh38
Transcripts
NM_144666.2 NM_173589.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
388700 c.1109G>C p.R370P Chr.11: 6509068-6509068 on Build GRCh38
7280234 c.990G>A p.G330G Chr.11: 6508949-6508949 on Build GRCh38
74410 c.988G>C p.G330R Chr.11: 6508947-6508947 on Build GRCh38
1355928 c.1160G>A p.R387Q Chr.11: 6509197-6509197 on Build GRCh38
4678807 c.993C>A p.I331I Chr.11: 6508952-6508952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746938233 Chr.11: 6508901-6508901 on Build GRCh38
rs758258758 Chr.11: 6508886-6508886 on Build GRCh38
rs746295368 Chr.11: 6509027-6509027 on Build GRCh38
rs771040802 Chr.11: 6509209-6509210 on Build GRCh38
rs201933727 Chr.11: 6509162-6509162 on Build GRCh38
rs750084260 Chr.11: 6508967-6508967 on Build GRCh38
rs764725469 Chr.11: 6508966-6508966 on Build GRCh38
rs191737842 Chr.11: 6508830-6508830 on Build GRCh38
rs990290378 Chr.11: 6509132-6509132 on Build GRCh38
rs534034133 Chr.11: 6508958-6508958 on Build GRCh38
rs147108806 Chr.11: 6509197-6509197 on Build GRCh38
rs918737675 Chr.11: 6508837-6508837 on Build GRCh38
rs747540896 Chr.11: 6509210-6509210 on Build GRCh38
rs141561327 Chr.11: 6509068-6509068 on Build GRCh38
rs752685203 Chr.11: 6509110-6509110 on Build GRCh38
rs773534881 Chr.11: 6508859-6508859 on Build GRCh38
rs374340852 Chr.11: 6508979-6508979 on Build GRCh38
rs370686709 Chr.11: 6508965-6508965 on Build GRCh38
rs772518108 Chr.11: 6508861-6508862 on Build GRCh38
rs778972137 Chr.11: 6509023-6509023 on Build GRCh38
rs368524005 Chr.11: 6508980-6508980 on Build GRCh38
rs371864238 Chr.11: 6508839-6508839 on Build GRCh38
rs11605607 Chr.11: 6508876-6508876 on Build GRCh38
rs764176673 Chr.11: 6509269-6509269 on Build GRCh38
rs138424831 Chr.11: 6508909-6508909 on Build GRCh38
rs766751413 Chr.11: 6508865-6508865 on Build GRCh38
rs774117627 Chr.11: 6509238-6509238 on Build GRCh38
rs772312325 Chr.11: 6509044-6509044 on Build GRCh38
rs764920159 Chr.11: 6508877-6508877 on Build GRCh38
rs368146755 Chr.11: 6508957-6508957 on Build GRCh38
rs77615924 Chr.11: 6509045-6509045 on Build GRCh38
rs868725621 Chr.11: 6509187-6509187 on Build GRCh38
rs1012035182 Chr.11: 6509016-6509016 on Build GRCh38
rs773625042 Chr.11: 6508923-6508923 on Build GRCh38
rs372813987 Chr.11: 6508983-6508983 on Build GRCh38
rs748083515 Chr.11: 6508990-6508990 on Build GRCh38
rs759430513 Chr.11: 6509096-6509096 on Build GRCh38
rs773745280 Chr.11: 6509165-6509167 on Build GRCh38
rs372485903 Chr.11: 6508840-6508840 on Build GRCh38
rs200198471 Chr.11: 6509012-6509012 on Build GRCh38
rs150652653 Chr.11: 6509171-6509171 on Build GRCh38
rs757262891 Chr.11: 6509156-6509156 on Build GRCh38
rs761142617 Chr.11: 6509207-6509216 on Build GRCh38
rs752491082 Chr.11: 6509258-6509258 on Build GRCh38
rs760515750 Chr.11: 6509263-6509263 on Build GRCh38
rs1017154198 Chr.11: 6508821-6508821 on Build GRCh38
rs200579735 Chr.11: 6508882-6508882 on Build GRCh38
rs766092670 Chr.11: 6509088-6509088 on Build GRCh38
rs59666671 Chr.11: 6509117-6509117 on Build GRCh38
rs772664290 Chr.11: 6509067-6509067 on Build GRCh38
rs963283157 Chr.11: 6508827-6508827 on Build GRCh38
rs11040904 Chr.11: 6509246-6509246 on Build GRCh38
rs955502320 Chr.11: 6509095-6509095 on Build GRCh38
rs754891117 Chr.11: 6508907-6508907 on Build GRCh38
rs559911004 Chr.11: 6508868-6508868 on Build GRCh38
rs770097644 Chr.11: 6508854-6508854 on Build GRCh38
rs562468372 Chr.11: 6508894-6508894 on Build GRCh38
rs560475031 Chr.11: 6508846-6508846 on Build GRCh38
rs770331471 Chr.11: 6509233-6509233 on Build GRCh38
rs767433471 Chr.11: 6509251-6509251 on Build GRCh38
rs759042132 Chr.11: 6509250-6509250 on Build GRCh38
rs753990936 Chr.11: 6509137-6509137 on Build GRCh38
rs767413450 Chr.11: 6509104-6509104 on Build GRCh38
rs148851238 Chr.11: 6508950-6508950 on Build GRCh38
rs777088299 Chr.11: 6509225-6509225 on Build GRCh38
rs547953838 Chr.11: 6509000-6509000 on Build GRCh38
rs376990274 Chr.11: 6508875-6508875 on Build GRCh38
rs758015047 Chr.11: 6508977-6508977 on Build GRCh38
rs756327896 Chr.11: 6508996-6508996 on Build GRCh38
rs138581667 Chr.11: 6509221-6509221 on Build GRCh38
rs371529610 Chr.11: 6509123-6509123 on Build GRCh38
rs1003351058 Chr.11: 6508981-6508981 on Build GRCh38
rs763974639 Chr.11: 6509076-6509076 on Build GRCh38
rs775956138 Chr.11: 6508964-6508964 on Build GRCh38
rs374766435 Chr.11: 6509061-6509061 on Build GRCh38
rs781237261 Chr.11: 6508984-6508984 on Build GRCh38
rs201807447 Chr.11: 6508922-6508922 on Build GRCh38
rs763457297 Chr.11: 6508861-6508861 on Build GRCh38
rs202036632 Chr.11: 6509203-6509203 on Build GRCh38
rs934040552 Chr.11: 6509196-6509196 on Build GRCh38
rs762570786 Chr.11: 6509226-6509226 on Build GRCh38
rs142816700 Chr.11: 6509011-6509011 on Build GRCh38
rs774729558 Chr.11: 6508953-6508953 on Build GRCh38
rs10839568 Chr.11: 6509048-6509048 on Build GRCh38
rs2555152 Chr.11: 6508908-6508908 on Build GRCh38
rs749517252 Chr.11: 6508925-6508925 on Build GRCh38
rs946063054 Chr.11: 6509194-6509194 on Build GRCh38
rs925886708 Chr.11: 6508868-6508869 on Build GRCh38

View Full Product Details