Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6519846-6520348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNHD1
Entrez Gene ID
144132
Gene Name
dynein heavy chain domain 1
Gene Aliases
C11orf47, CCDC35, DHCD1, DNHD1L
UniGene
Hs.720080
Gene Chromosome Location
Chr.11: 6497296-6572025 on Build GRCh38
Transcripts
NM_144666.2 NM_173589.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6134251 c.1655G>A p.R552K Chr.11: 6519972-6519972 on Build GRCh38
4146116 c.1678C>G p.Q560E Chr.11: 6519995-6519995 on Build GRCh38
6134252 c.1655G>A p.R552K Chr.11: 6519972-6519972 on Build GRCh38
369246 c.1701T>C p.G567G Chr.11: 6520018-6520018 on Build GRCh38
3953665 c.1717C>A p.P573T Chr.11: 6520034-6520034 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758313222 Chr.11: 6519885-6519885 on Build GRCh38
rs200084337 Chr.11: 6519916-6519916 on Build GRCh38
rs781456460 Chr.11: 6520057-6520057 on Build GRCh38
rs150005725 Chr.11: 6519928-6519928 on Build GRCh38
rs774250477 Chr.11: 6520117-6520117 on Build GRCh38
rs369968188 Chr.11: 6519920-6519920 on Build GRCh38
rs531838027 Chr.11: 6520309-6520309 on Build GRCh38
rs747446302 Chr.11: 6520228-6520228 on Build GRCh38
rs554735967 Chr.11: 6520179-6520179 on Build GRCh38
rs776078547 Chr.11: 6520029-6520029 on Build GRCh38
rs180925806 Chr.11: 6520291-6520291 on Build GRCh38
rs763833968 Chr.11: 6519960-6519961 on Build GRCh38
rs372824250 Chr.11: 6520059-6520059 on Build GRCh38
rs769363993 Chr.11: 6520229-6520229 on Build GRCh38
rs764015324 Chr.11: 6520144-6520144 on Build GRCh38
rs758104167 Chr.11: 6519963-6519963 on Build GRCh38
rs1055736063 Chr.11: 6519904-6519904 on Build GRCh38
rs765228863 Chr.11: 6519931-6519932 on Build GRCh38
rs1007218548 Chr.11: 6520281-6520281 on Build GRCh38
rs759464478 Chr.11: 6520298-6520298 on Build GRCh38
rs140575681 Chr.11: 6519869-6519869 on Build GRCh38
rs747158555 Chr.11: 6519914-6519914 on Build GRCh38
rs761403490 Chr.11: 6519952-6519952 on Build GRCh38
rs762869811 Chr.11: 6520048-6520048 on Build GRCh38
rs766024913 Chr.11: 6520054-6520054 on Build GRCh38
rs1008185209 Chr.11: 6519988-6519988 on Build GRCh38
rs770103720 Chr.11: 6520013-6520013 on Build GRCh38
rs777828412 Chr.11: 6520016-6520016 on Build GRCh38
rs376610634 Chr.11: 6520089-6520089 on Build GRCh38
rs2555144 Chr.11: 6519876-6519876 on Build GRCh38
rs749713520 Chr.11: 6520017-6520017 on Build GRCh38
rs537598476 Chr.11: 6519981-6519981 on Build GRCh38
rs766223945 Chr.11: 6520113-6520113 on Build GRCh38
rs756242298 Chr.11: 6520073-6520073 on Build GRCh38
rs1046414141 Chr.11: 6519868-6519868 on Build GRCh38
rs11603869 Chr.11: 6519995-6519995 on Build GRCh38
rs751105412 Chr.11: 6520161-6520161 on Build GRCh38
rs562793982 Chr.11: 6520278-6520278 on Build GRCh38
rs751560800 Chr.11: 6520056-6520056 on Build GRCh38
rs890082715 Chr.11: 6520270-6520270 on Build GRCh38
rs756283836 Chr.11: 6520143-6520143 on Build GRCh38
rs374350205 Chr.11: 6520122-6520122 on Build GRCh38
rs749003821 Chr.11: 6520243-6520243 on Build GRCh38
rs369082978 Chr.11: 6519974-6519974 on Build GRCh38
rs779073850 Chr.11: 6520205-6520205 on Build GRCh38
rs200207984 Chr.11: 6520023-6520023 on Build GRCh38
rs773932014 Chr.11: 6519968-6519968 on Build GRCh38
rs772552317 Chr.11: 6520230-6520230 on Build GRCh38
rs748318667 Chr.11: 6519918-6519918 on Build GRCh38
rs1000469363 Chr.11: 6520283-6520283 on Build GRCh38
rs750269175 Chr.11: 6519957-6519957 on Build GRCh38
rs770274244 Chr.11: 6519960-6519960 on Build GRCh38
rs762897900 Chr.11: 6519953-6519953 on Build GRCh38
rs749467852 Chr.11: 6520078-6520078 on Build GRCh38
rs746277595 Chr.11: 6520214-6520214 on Build GRCh38
rs775951835 Chr.11: 6520096-6520096 on Build GRCh38
rs752670743 Chr.11: 6520133-6520133 on Build GRCh38
rs759977241 Chr.11: 6520024-6520024 on Build GRCh38
rs763526268 Chr.11: 6519939-6519939 on Build GRCh38
rs751276530 Chr.11: 6520153-6520154 on Build GRCh38
rs555131212 Chr.11: 6519941-6519941 on Build GRCh38
rs774699427 Chr.11: 6519933-6519933 on Build GRCh38
rs771339647 Chr.11: 6520080-6520080 on Build GRCh38
rs779821997 Chr.11: 6519903-6519903 on Build GRCh38
rs777064772 Chr.11: 6519917-6519917 on Build GRCh38
rs371116329 Chr.11: 6520108-6520108 on Build GRCh38
rs775346894 Chr.11: 6519952-6519953 on Build GRCh38
rs772606882 Chr.11: 6520095-6520095 on Build GRCh38
rs771187718 Chr.11: 6520018-6520018 on Build GRCh38
rs746402650 Chr.11: 6520093-6520093 on Build GRCh38
rs147700957 Chr.11: 6519953-6519953 on Build GRCh38
rs770298039 Chr.11: 6519919-6519919 on Build GRCh38
rs747712194 Chr.11: 6520098-6520098 on Build GRCh38
rs754112651 Chr.11: 6520203-6520203 on Build GRCh38
rs755116584 Chr.11: 6519991-6519991 on Build GRCh38
rs551945734 Chr.11: 6520293-6520293 on Build GRCh38
rs16914748 Chr.11: 6520075-6520075 on Build GRCh38
rs142458347 Chr.11: 6520174-6520174 on Build GRCh38
rs897675186 Chr.11: 6520226-6520226 on Build GRCh38
rs780601353 Chr.11: 6520220-6520220 on Build GRCh38
rs183492473 Chr.11: 6520312-6520312 on Build GRCh38
rs868669609 Chr.11: 6520008-6520008 on Build GRCh38
rs560416128 Chr.11: 6520119-6520119 on Build GRCh38
rs35232075 Chr.11: 6520277-6520278 on Build GRCh38
rs769178794 Chr.11: 6520100-6520100 on Build GRCh38
rs762459458 Chr.11: 6520112-6520112 on Build GRCh38
rs201456556 Chr.11: 6519940-6519940 on Build GRCh38
rs770532037 Chr.11: 6520263-6520263 on Build GRCh38
rs763402834 Chr.11: 6519925-6519925 on Build GRCh38
rs535448591 Chr.11: 6519937-6519937 on Build GRCh38
rs750026869 Chr.11: 6520035-6520035 on Build GRCh38
rs1021158823 Chr.11: 6520069-6520069 on Build GRCh38
rs761279028 Chr.11: 6520030-6520030 on Build GRCh38
rs757125046 Chr.11: 6520241-6520241 on Build GRCh38
rs73404907 Chr.11: 6520204-6520204 on Build GRCh38
rs140159867 Chr.11: 6520031-6520031 on Build GRCh38
rs758746595 Chr.11: 6520215-6520215 on Build GRCh38
rs189469214 Chr.11: 6520028-6520028 on Build GRCh38

View Full Product Details