Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 31290550-31291054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCDC1
Entrez Gene ID
341019
Gene Name
doublecortin domain containing 1
Gene Aliases
-
UniGene
Hs.127405
Gene Chromosome Location
Chr.11: 31262624-31369810 on Build GRCh38
Transcripts
NM_181807.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4886683 c.960G>T p.K320N Chr.11: 31290647-31290647 on Build GRCh38
687546 c.827G>T p.R276L Chr.11: 31290780-31290780 on Build GRCh38
6132541 c.810T>A p.P270P Chr.11: 31290797-31290797 on Build GRCh38
6990278 c.945A>C p.K315N Chr.11: 31290662-31290662 on Build GRCh38
6132542 c.920A>T p.D307V Chr.11: 31290687-31290687 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375102072 Chr.11: 31290772-31290772 on Build GRCh38
rs1054451977 Chr.11: 31291023-31291023 on Build GRCh38
rs753012643 Chr.11: 31290888-31290888 on Build GRCh38
rs781524276 Chr.11: 31290705-31290705 on Build GRCh38
rs868553033 Chr.11: 31290799-31290799 on Build GRCh38
rs777916661 Chr.11: 31290893-31290893 on Build GRCh38
rs756842589 Chr.11: 31290683-31290683 on Build GRCh38
rs368433945 Chr.11: 31290789-31290789 on Build GRCh38
rs200169786 Chr.11: 31290687-31290687 on Build GRCh38
rs780593249 Chr.11: 31290770-31290770 on Build GRCh38
rs550364554 Chr.11: 31290721-31290721 on Build GRCh38
rs369862087 Chr.11: 31290646-31290646 on Build GRCh38
rs148157215 Chr.11: 31290778-31290778 on Build GRCh38
rs902758879 Chr.11: 31290962-31290962 on Build GRCh38
rs957566567 Chr.11: 31290788-31290788 on Build GRCh38
rs546144417 Chr.11: 31290606-31290606 on Build GRCh38
rs972037014 Chr.11: 31290843-31290843 on Build GRCh38
rs370896971 Chr.11: 31290720-31290720 on Build GRCh38
rs760605601 Chr.11: 31290742-31290742 on Build GRCh38
rs765841446 Chr.11: 31290869-31290869 on Build GRCh38
rs761602412 Chr.11: 31290853-31290853 on Build GRCh38
rs201970490 Chr.11: 31290634-31290634 on Build GRCh38
rs764863960 Chr.11: 31290855-31290855 on Build GRCh38
rs142025656 Chr.11: 31290781-31290781 on Build GRCh38
rs748382083 Chr.11: 31290713-31290713 on Build GRCh38
rs755465517 Chr.11: 31290873-31290873 on Build GRCh38
rs755906212 Chr.11: 31290661-31290661 on Build GRCh38
rs201768545 Chr.11: 31290820-31290820 on Build GRCh38
rs377395170 Chr.11: 31290690-31290690 on Build GRCh38
rs781684227 Chr.11: 31290882-31290882 on Build GRCh38
rs143766770 Chr.11: 31290753-31290753 on Build GRCh38
rs930507370 Chr.11: 31290984-31290984 on Build GRCh38
rs769923964 Chr.11: 31290782-31290782 on Build GRCh38
rs777989162 Chr.11: 31290783-31290783 on Build GRCh38
rs920007996 Chr.11: 31290973-31290973 on Build GRCh38
rs780472612 Chr.11: 31290691-31290691 on Build GRCh38
rs778316009 Chr.11: 31290685-31290685 on Build GRCh38
rs988968496 Chr.11: 31290805-31290805 on Build GRCh38
rs775853715 Chr.11: 31290624-31290624 on Build GRCh38
rs918029060 Chr.11: 31290849-31290849 on Build GRCh38
rs985028397 Chr.11: 31290779-31290779 on Build GRCh38
rs776132836 Chr.11: 31290646-31290646 on Build GRCh38
rs763777697 Chr.11: 31290747-31290747 on Build GRCh38
rs537908701 Chr.11: 31291021-31291021 on Build GRCh38
rs765205939 Chr.11: 31290642-31290642 on Build GRCh38
rs756717181 Chr.11: 31290598-31290598 on Build GRCh38
rs775620147 Chr.11: 31290804-31290804 on Build GRCh38
rs528453045 Chr.11: 31290652-31290652 on Build GRCh38
rs747315761 Chr.11: 31290629-31290629 on Build GRCh38
rs762494038 Chr.11: 31290859-31290859 on Build GRCh38
rs913513340 Chr.11: 31290838-31290838 on Build GRCh38
rs566493478 Chr.11: 31290872-31290872 on Build GRCh38
rs748836247 Chr.11: 31290596-31290596 on Build GRCh38
rs981483007 Chr.11: 31290864-31290864 on Build GRCh38
rs772143702 Chr.11: 31290737-31290737 on Build GRCh38
rs936896078 Chr.11: 31290996-31290996 on Build GRCh38
rs781437821 Chr.11: 31290780-31290780 on Build GRCh38
rs188130177 Chr.11: 31290960-31290960 on Build GRCh38
rs756500358 Chr.11: 31290889-31290889 on Build GRCh38
rs753568409 Chr.11: 31290749-31290749 on Build GRCh38
rs750060523 Chr.11: 31290856-31290856 on Build GRCh38
rs768587567 Chr.11: 31290837-31290837 on Build GRCh38
rs533541855 Chr.11: 31290916-31290916 on Build GRCh38
rs377121072 Chr.11: 31290763-31290763 on Build GRCh38
rs937448894 Chr.11: 31290943-31290943 on Build GRCh38
rs201457983 Chr.11: 31290670-31290670 on Build GRCh38
rs762007144 Chr.11: 31290636-31290636 on Build GRCh38
rs760659009 Chr.11: 31290834-31290834 on Build GRCh38
rs759581018 Chr.11: 31290648-31290648 on Build GRCh38
rs370579706 Chr.11: 31290768-31290768 on Build GRCh38
rs746339656 Chr.11: 31290608-31290608 on Build GRCh38
rs374215197 Chr.11: 31290617-31290617 on Build GRCh38
rs768869205 Chr.11: 31290631-31290631 on Build GRCh38
rs776301174 Chr.11: 31290847-31290847 on Build GRCh38
rs555544870 Chr.11: 31290937-31290937 on Build GRCh38
rs759172244 Chr.11: 31290758-31290761 on Build GRCh38
rs764884735 Chr.11: 31290756-31290756 on Build GRCh38
rs371688726 Chr.11: 31290796-31290796 on Build GRCh38
rs201039841 Chr.11: 31290658-31290658 on Build GRCh38
rs1054897157 Chr.11: 31290645-31290645 on Build GRCh38
rs374620942 Chr.11: 31290671-31290671 on Build GRCh38
rs529307605 Chr.11: 31290699-31290699 on Build GRCh38
rs568684214 Chr.11: 31290738-31290738 on Build GRCh38
rs747437986 Chr.11: 31290697-31290697 on Build GRCh38

View Full Product Details