Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 32589061-32589577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3M
Entrez Gene ID
10480
Gene Name
eukaryotic translation initiation factor 3 subunit M
Gene Aliases
B5, GA17, PCID1, TANGO7, hfl-B5
UniGene
Hs.502244
Gene Chromosome Location
Chr.11: 32583767-32602873 on Build GRCh38
Transcripts
NM_001307929.1 NM_006360.5 XM_011519840.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5626038 c.381T>G p.L127L Chr.11: 32589078-32589078 on Build GRCh38
926369 c.462G>T p.W154C Chr.11: 32589570-32589570 on Build GRCh38
310834 c.439-2A>T p.? Chr.11: 32589545-32589545 on Build GRCh38
1746205 c.429G>T p.E143D Chr.11: 32589126-32589126 on Build GRCh38
4974888 c.450G>T p.W150C Chr.11: 32589558-32589558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037147354 Chr.11: 32589217-32589217 on Build GRCh38
rs532471144 Chr.11: 32589270-32589270 on Build GRCh38
rs527363716 Chr.11: 32589198-32589198 on Build GRCh38
rs769933891 Chr.11: 32589148-32589148 on Build GRCh38
rs767400024 Chr.11: 32589158-32589158 on Build GRCh38
rs763035659 Chr.11: 32589153-32589153 on Build GRCh38
rs748385317 Chr.11: 32589147-32589147 on Build GRCh38
rs537615119 Chr.11: 32589365-32589365 on Build GRCh38
rs760453112 Chr.11: 32589179-32589179 on Build GRCh38
rs75907554 Chr.11: 32589193-32589193 on Build GRCh38
rs560620221 Chr.11: 32589468-32589468 on Build GRCh38
rs753627173 Chr.11: 32589508-32589508 on Build GRCh38
rs556010748 Chr.11: 32589492-32589496 on Build GRCh38
rs184975960 Chr.11: 32589238-32589238 on Build GRCh38
rs1055266107 Chr.11: 32589294-32589294 on Build GRCh38
rs938174765 Chr.11: 32589284-32589284 on Build GRCh38
rs911799761 Chr.11: 32589405-32589405 on Build GRCh38
rs375604117 Chr.11: 32589180-32589181 on Build GRCh38
rs754809465 Chr.11: 32589434-32589434 on Build GRCh38
rs1028786788 Chr.11: 32589377-32589377 on Build GRCh38
rs967134820 Chr.11: 32589431-32589431 on Build GRCh38
rs944387804 Chr.11: 32589499-32589499 on Build GRCh38
rs974522376 Chr.11: 32589222-32589222 on Build GRCh38
rs540636725 Chr.11: 32589465-32589465 on Build GRCh38
rs912541284 Chr.11: 32589209-32589209 on Build GRCh38
rs972728677 Chr.11: 32589425-32589425 on Build GRCh38
rs895512642 Chr.11: 32589323-32589323 on Build GRCh38
rs181709087 Chr.11: 32589304-32589304 on Build GRCh38
rs776929097 Chr.11: 32589181-32589181 on Build GRCh38
rs367920217 Chr.11: 32589538-32589538 on Build GRCh38
rs558265467 Chr.11: 32589429-32589429 on Build GRCh38
rs568573241 Chr.11: 32589345-32589345 on Build GRCh38
rs540154931 Chr.11: 32589426-32589426 on Build GRCh38
rs1009221019 Chr.11: 32589160-32589160 on Build GRCh38
rs1047917156 Chr.11: 32589469-32589469 on Build GRCh38
rs374411010 Chr.11: 32589535-32589535 on Build GRCh38
rs780588966 Chr.11: 32589121-32589121 on Build GRCh38
rs775466054 Chr.11: 32589159-32589159 on Build GRCh38
rs747377742 Chr.11: 32589138-32589138 on Build GRCh38
rs749951116 Chr.11: 32589526-32589526 on Build GRCh38
rs995971241 Chr.11: 32589384-32589384 on Build GRCh38
rs781539131 Chr.11: 32589543-32589543 on Build GRCh38
rs755385731 Chr.11: 32589539-32589539 on Build GRCh38
rs79383227 Chr.11: 32589194-32589194 on Build GRCh38
rs188596977 Chr.11: 32589262-32589262 on Build GRCh38
rs921877779 Chr.11: 32589156-32589156 on Build GRCh38
rs149778891 Chr.11: 32589457-32589457 on Build GRCh38
rs775755884 Chr.11: 32589181-32589182 on Build GRCh38
rs928210047 Chr.11: 32589454-32589454 on Build GRCh38
rs1057401228 Chr.11: 32589491-32589491 on Build GRCh38
rs900397024 Chr.11: 32589366-32589366 on Build GRCh38
rs549969116 Chr.11: 32589257-32589257 on Build GRCh38
rs527977049 Chr.11: 32589267-32589267 on Build GRCh38
rs1018836056 Chr.11: 32589364-32589364 on Build GRCh38
rs906118727 Chr.11: 32589264-32589264 on Build GRCh38
rs529800100 Chr.11: 32589241-32589241 on Build GRCh38
rs918712862 Chr.11: 32589432-32589432 on Build GRCh38
rs764936657 Chr.11: 32589516-32589516 on Build GRCh38
rs763965988 Chr.11: 32589500-32589500 on Build GRCh38
rs763074334 Chr.11: 32589181-32589183 on Build GRCh38
rs1008839163 Chr.11: 32589344-32589344 on Build GRCh38
rs886090071 Chr.11: 32589334-32589334 on Build GRCh38
rs937490076 Chr.11: 32589478-32589478 on Build GRCh38
rs771497291 Chr.11: 32589140-32589141 on Build GRCh38
rs540774808 Chr.11: 32589237-32589237 on Build GRCh38
rs1047423745 Chr.11: 32589333-32589333 on Build GRCh38
rs958185817 Chr.11: 32589404-32589404 on Build GRCh38
rs757894482 Chr.11: 32589530-32589530 on Build GRCh38
rs907592907 Chr.11: 32589477-32589477 on Build GRCh38
rs1028374300 Chr.11: 32589226-32589226 on Build GRCh38
rs555058796 Chr.11: 32589337-32589337 on Build GRCh38
rs748469156 Chr.11: 32589546-32589546 on Build GRCh38
rs201876 Chr.11: 32589422-32589422 on Build GRCh38
rs113960713 Chr.11: 32589326-32589326 on Build GRCh38
rs184499491 Chr.11: 32589403-32589403 on Build GRCh38
rs931883360 Chr.11: 32589182-32589182 on Build GRCh38
rs945678528 Chr.11: 32589253-32589253 on Build GRCh38
rs867775948 Chr.11: 32589199-32589199 on Build GRCh38
rs991027060 Chr.11: 32589459-32589459 on Build GRCh38
rs769065555 Chr.11: 32589143-32589143 on Build GRCh38
rs1009898670 Chr.11: 32589501-32589501 on Build GRCh38
rs898919024 Chr.11: 32589317-32589317 on Build GRCh38
rs1000444654 Chr.11: 32589374-32589374 on Build GRCh38
rs773309074 Chr.11: 32589151-32589151 on Build GRCh38
rs142224866 Chr.11: 32589278-32589278 on Build GRCh38
rs757002593 Chr.11: 32589515-32589515 on Build GRCh38
rs776705413 Chr.11: 32589344-32589346 on Build GRCh38
rs564931655 Chr.11: 32589146-32589146 on Build GRCh38

View Full Product Details