Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 32601995-32602498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC73
Entrez Gene ID
493860
Gene Name
coiled-coil domain containing 73
Gene Aliases
NY-SAR-79
UniGene
Hs.706808
Gene Chromosome Location
Chr.11: 32602080-32794658 on Build GRCh38
Transcripts
NM_001008391.3 XM_011520139.2 NM_001307929.1 NM_006360.5 XM_011519840.2

Gene Symbol
EIF3M
Entrez Gene ID
10480
Gene Name
eukaryotic translation initiation factor 3 subunit M
Gene Aliases
B5, GA17, PCID1, TANGO7, hfl-B5
UniGene
Hs.502244
Gene Chromosome Location
Chr.11: 32583767-32602873 on Build GRCh38
Transcripts
NM_001008391.3 XM_011520139.2 NM_001307929.1 NM_006360.5 XM_011519840.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3986128 c.1078A>T p.N360Y Chr.11: 32602352-32602352 on Build GRCh38
4032393 c.1055A>G p.D352G Chr.11: 32602329-32602329 on Build GRCh38
3935440 c.1037A>G p.Q346R Chr.11: 32602311-32602311 on Build GRCh38
6652891 c.1114T>C p.S372P Chr.11: 32602388-32602388 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766205374 Chr.11: 32602259-32602263 on Build GRCh38
rs200744266 Chr.11: 32602377-32602377 on Build GRCh38
rs753447122 Chr.11: 32602400-32602400 on Build GRCh38
rs755768294 Chr.11: 32602128-32602128 on Build GRCh38
rs780500967 Chr.11: 32602445-32602445 on Build GRCh38
rs200108153 Chr.11: 32602349-32602349 on Build GRCh38
rs930633659 Chr.11: 32602130-32602130 on Build GRCh38
rs764987409 Chr.11: 32602429-32602429 on Build GRCh38
rs779346728 Chr.11: 32602424-32602424 on Build GRCh38
rs139971850 Chr.11: 32602354-32602354 on Build GRCh38
rs763789706 Chr.11: 32602382-32602382 on Build GRCh38
rs920106958 Chr.11: 32602081-32602081 on Build GRCh38
rs549328943 Chr.11: 32602252-32602252 on Build GRCh38
rs772471774 Chr.11: 32602232-32602232 on Build GRCh38
rs757860370 Chr.11: 32602145-32602145 on Build GRCh38
rs890875743 Chr.11: 32602144-32602144 on Build GRCh38
rs756760747 Chr.11: 32602407-32602407 on Build GRCh38
rs750456612 Chr.11: 32602249-32602250 on Build GRCh38
rs767499449 Chr.11: 32602218-32602218 on Build GRCh38
rs937582315 Chr.11: 32602053-32602053 on Build GRCh38
rs151094802 Chr.11: 32602351-32602351 on Build GRCh38
rs762005099 Chr.11: 32602121-32602122 on Build GRCh38
rs774248775 Chr.11: 32602122-32602122 on Build GRCh38
rs899771794 Chr.11: 32602174-32602174 on Build GRCh38
rs768828123 Chr.11: 32602285-32602285 on Build GRCh38
rs767533888 Chr.11: 32602248-32602249 on Build GRCh38
rs74673753 Chr.11: 32602075-32602075 on Build GRCh38
rs12581 Chr.11: 32602398-32602398 on Build GRCh38
rs761861749 Chr.11: 32602309-32602309 on Build GRCh38
rs533897916 Chr.11: 32602120-32602120 on Build GRCh38
rs766272074 Chr.11: 32602336-32602336 on Build GRCh38
rs138423523 Chr.11: 32602168-32602168 on Build GRCh38
rs755860947 Chr.11: 32602375-32602375 on Build GRCh38
rs1028081653 Chr.11: 32602374-32602374 on Build GRCh38
rs186745785 Chr.11: 32602129-32602129 on Build GRCh38
rs776717920 Chr.11: 32602290-32602290 on Build GRCh38
rs753694073 Chr.11: 32602400-32602401 on Build GRCh38
rs374239887 Chr.11: 32602345-32602345 on Build GRCh38
rs754534624 Chr.11: 32602428-32602429 on Build GRCh38
rs1028482777 Chr.11: 32602073-32602073 on Build GRCh38
rs532912439 Chr.11: 32602116-32602117 on Build GRCh38
rs747238061 Chr.11: 32602255-32602255 on Build GRCh38
rs775660528 Chr.11: 32602238-32602238 on Build GRCh38
rs750954431 Chr.11: 32602442-32602442 on Build GRCh38
rs757136622 Chr.11: 32602151-32602151 on Build GRCh38
rs116006893 Chr.11: 32602179-32602179 on Build GRCh38
rs961255180 Chr.11: 32602121-32602121 on Build GRCh38
rs1008081588 Chr.11: 32602466-32602467 on Build GRCh38
rs1049240 Chr.11: 32602355-32602355 on Build GRCh38
rs746257830 Chr.11: 32602157-32602157 on Build GRCh38
rs1802362 Chr.11: 32602368-32602368 on Build GRCh38
rs117038056 Chr.11: 32602428-32602428 on Build GRCh38
rs747327870 Chr.11: 32602450-32602450 on Build GRCh38
rs760538055 Chr.11: 32602255-32602256 on Build GRCh38
rs956755541 Chr.11: 32602233-32602233 on Build GRCh38
rs1802363 Chr.11: 32602311-32602311 on Build GRCh38
rs748766241 Chr.11: 32602132-32602132 on Build GRCh38
rs758766576 Chr.11: 32602444-32602444 on Build GRCh38

View Full Product Details