Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33559671-33560165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1549L
Entrez Gene ID
25758
Gene Name
KIAA1549 like
Gene Aliases
C11orf41, C11orf69, G2
UniGene
Hs.502266
Gene Chromosome Location
Chr.11: 33376569-33674102 on Build GRCh38
Transcripts
NM_012194.2 XM_005252847.3 XM_005252848.3 XM_011519969.2 XM_011519970.2 XM_017017484.1 XM_017017485.1 XM_017017486.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5999979 c.3058C>T p.L1020F Chr.11: 33559842-33559842 on Build GRCh38
6518844 c.3077C>A p.A1026D Chr.11: 33559861-33559861 on Build GRCh38
2115165 c.3009C>T p.Y1003Y Chr.11: 33559793-33559793 on Build GRCh38
3710169 c.3066C>T p.V1022V Chr.11: 33559832-33559832 on Build GRCh38
6694272 c.3026A>G p.Y1009C Chr.11: 33559792-33559792 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751160839 Chr.11: 33559776-33559776 on Build GRCh38
rs955270764 Chr.11: 33560031-33560031 on Build GRCh38
rs765763365 Chr.11: 33559830-33559830 on Build GRCh38
rs377212326 Chr.11: 33559926-33559926 on Build GRCh38
rs535097988 Chr.11: 33559695-33559695 on Build GRCh38
rs747075827 Chr.11: 33559891-33559891 on Build GRCh38
rs866361540 Chr.11: 33560103-33560103 on Build GRCh38
rs760407126 Chr.11: 33559935-33559935 on Build GRCh38
rs370615392 Chr.11: 33559892-33559892 on Build GRCh38
rs776686339 Chr.11: 33559898-33559898 on Build GRCh38
rs376877164 Chr.11: 33559824-33559824 on Build GRCh38
rs201351687 Chr.11: 33559813-33559813 on Build GRCh38
rs757060464 Chr.11: 33559953-33559953 on Build GRCh38
rs767891016 Chr.11: 33559991-33559992 on Build GRCh38
rs375574453 Chr.11: 33559832-33559832 on Build GRCh38
rs1008106121 Chr.11: 33560035-33560035 on Build GRCh38
rs752298292 Chr.11: 33559930-33559930 on Build GRCh38
rs748425161 Chr.11: 33559903-33559903 on Build GRCh38
rs200341089 Chr.11: 33560001-33560001 on Build GRCh38
rs974374071 Chr.11: 33559869-33559869 on Build GRCh38
rs778993358 Chr.11: 33559955-33559955 on Build GRCh38
rs777054653 Chr.11: 33559814-33559814 on Build GRCh38
rs1015682557 Chr.11: 33560054-33560054 on Build GRCh38
rs767446780 Chr.11: 33559998-33560003 on Build GRCh38
rs775222228 Chr.11: 33560118-33560118 on Build GRCh38
rs187248502 Chr.11: 33560014-33560014 on Build GRCh38
rs565290499 Chr.11: 33560010-33560010 on Build GRCh38
rs754261322 Chr.11: 33559758-33559758 on Build GRCh38
rs750430092 Chr.11: 33559957-33559957 on Build GRCh38
rs373132227 Chr.11: 33559833-33559833 on Build GRCh38
rs372052625 Chr.11: 33559765-33559765 on Build GRCh38
rs939395656 Chr.11: 33559943-33559943 on Build GRCh38
rs569636292 Chr.11: 33559870-33559870 on Build GRCh38
rs921794423 Chr.11: 33560003-33560003 on Build GRCh38
rs773566329 Chr.11: 33559827-33559827 on Build GRCh38
rs369439478 Chr.11: 33559797-33559797 on Build GRCh38
rs373858902 Chr.11: 33559894-33559894 on Build GRCh38
rs370357009 Chr.11: 33560119-33560119 on Build GRCh38
rs765959298 Chr.11: 33559769-33559769 on Build GRCh38
rs67485241 Chr.11: 33559996-33560005 on Build GRCh38
rs2281380 Chr.11: 33559760-33559760 on Build GRCh38
rs777566871 Chr.11: 33559796-33559796 on Build GRCh38
rs891814480 Chr.11: 33559730-33559730 on Build GRCh38
rs377470417 Chr.11: 33559859-33559859 on Build GRCh38
rs749141038 Chr.11: 33559861-33559861 on Build GRCh38
rs764515847 Chr.11: 33559757-33559757 on Build GRCh38
rs761002526 Chr.11: 33559728-33559728 on Build GRCh38
rs767054764 Chr.11: 33559929-33559929 on Build GRCh38
rs762355801 Chr.11: 33559759-33559759 on Build GRCh38
rs548862565 Chr.11: 33559721-33559721 on Build GRCh38
rs775672416 Chr.11: 33559800-33559800 on Build GRCh38
rs2281381 Chr.11: 33559716-33559716 on Build GRCh38
rs747163814 Chr.11: 33559801-33559801 on Build GRCh38
rs770001779 Chr.11: 33559911-33559911 on Build GRCh38
rs774431585 Chr.11: 33559707-33559707 on Build GRCh38
rs1028277288 Chr.11: 33560017-33560017 on Build GRCh38
rs554406748 Chr.11: 33560056-33560056 on Build GRCh38
rs780896033 Chr.11: 33559783-33559783 on Build GRCh38
rs892435463 Chr.11: 33559977-33559977 on Build GRCh38
rs376171809 Chr.11: 33559793-33559793 on Build GRCh38
rs35812670 Chr.11: 33559901-33559902 on Build GRCh38
rs907279404 Chr.11: 33560109-33560109 on Build GRCh38
rs559746494 Chr.11: 33559703-33559703 on Build GRCh38
rs774648717 Chr.11: 33559994-33560003 on Build GRCh38
rs775554287 Chr.11: 33559722-33559722 on Build GRCh38
rs752412913 Chr.11: 33559842-33559842 on Build GRCh38
rs763535300 Chr.11: 33559829-33559829 on Build GRCh38
rs1054393544 Chr.11: 33560045-33560045 on Build GRCh38
rs201089001 Chr.11: 33559996-33559996 on Build GRCh38
rs754654431 Chr.11: 33559777-33559777 on Build GRCh38
rs202009436 Chr.11: 33560000-33560000 on Build GRCh38
rs201608678 Chr.11: 33559823-33559823 on Build GRCh38
rs3830851 Chr.11: 33559992-33560001 on Build GRCh38
rs755959751 Chr.11: 33559794-33559794 on Build GRCh38
rs568033302 Chr.11: 33560108-33560108 on Build GRCh38
rs773654513 Chr.11: 33559924-33559924 on Build GRCh38
rs538467920 Chr.11: 33559941-33559941 on Build GRCh38
rs199908701 Chr.11: 33559942-33559942 on Build GRCh38
rs757811833 Chr.11: 33559768-33559768 on Build GRCh38
rs763299881 Chr.11: 33559925-33559925 on Build GRCh38
rs1056588308 Chr.11: 33559709-33559709 on Build GRCh38
rs764876123 Chr.11: 33560095-33560095 on Build GRCh38
rs759041196 Chr.11: 33559927-33559927 on Build GRCh38
rs755842537 Chr.11: 33559858-33559858 on Build GRCh38
rs779942373 Chr.11: 33559873-33559873 on Build GRCh38

View Full Product Details