Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33698310-33698769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf91
Entrez Gene ID
100131378
Gene Name
chromosome 11 open reading frame 91
Gene Aliases
-
UniGene
Hs.709466
Gene Chromosome Location
Chr.11: 33698108-33703026 on Build GRCh38
Transcripts
NM_001166692.1 XM_017017053.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1197943 c.582A>G p.*194W Chr.11: 33698429-33698429 on Build GRCh38
4145662 c.505C>A p.L169M Chr.11: 33698506-33698506 on Build GRCh38
6854861 c.522C>T p.D174D Chr.11: 33698489-33698489 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs190991415 Chr.11: 33698521-33698521 on Build GRCh38
rs772877435 Chr.11: 33698399-33698399 on Build GRCh38
rs150735832 Chr.11: 33698697-33698697 on Build GRCh38
rs1027722425 Chr.11: 33698375-33698375 on Build GRCh38
rs936449098 Chr.11: 33698696-33698696 on Build GRCh38
rs75967993 Chr.11: 33698461-33698461 on Build GRCh38
rs557468474 Chr.11: 33698628-33698628 on Build GRCh38
rs925151627 Chr.11: 33698542-33698542 on Build GRCh38
rs537910499 Chr.11: 33698611-33698611 on Build GRCh38
rs776196410 Chr.11: 33698381-33698381 on Build GRCh38
rs181771728 Chr.11: 33698452-33698452 on Build GRCh38
rs186192966 Chr.11: 33698462-33698462 on Build GRCh38
rs556775570 Chr.11: 33698573-33698573 on Build GRCh38
rs536895939 Chr.11: 33698553-33698553 on Build GRCh38
rs117079787 Chr.11: 33698676-33698676 on Build GRCh38
rs762847183 Chr.11: 33698401-33698401 on Build GRCh38
rs952204534 Chr.11: 33698390-33698390 on Build GRCh38
rs890795244 Chr.11: 33698576-33698576 on Build GRCh38
rs370336358 Chr.11: 33698499-33698499 on Build GRCh38
rs111357194 Chr.11: 33698541-33698541 on Build GRCh38
rs777214236 Chr.11: 33698534-33698546 on Build GRCh38
rs542409915 Chr.11: 33698402-33698402 on Build GRCh38
rs181704992 Chr.11: 33698577-33698577 on Build GRCh38
rs905153301 Chr.11: 33698407-33698407 on Build GRCh38
rs898103990 Chr.11: 33698658-33698658 on Build GRCh38
rs372577465 Chr.11: 33698633-33698633 on Build GRCh38
rs1026511873 Chr.11: 33698586-33698586 on Build GRCh38
rs963714655 Chr.11: 33698641-33698641 on Build GRCh38
rs1002696609 Chr.11: 33698711-33698711 on Build GRCh38
rs539836305 Chr.11: 33698691-33698691 on Build GRCh38
rs771548822 Chr.11: 33698398-33698398 on Build GRCh38
rs779265881 Chr.11: 33698523-33698523 on Build GRCh38
rs932360882 Chr.11: 33698571-33698571 on Build GRCh38
rs955056100 Chr.11: 33698609-33698609 on Build GRCh38
rs773200220 Chr.11: 33698726-33698726 on Build GRCh38
rs761466657 Chr.11: 33698396-33698396 on Build GRCh38
rs527981527 Chr.11: 33698498-33698498 on Build GRCh38
rs963493926 Chr.11: 33698488-33698488 on Build GRCh38
rs960733478 Chr.11: 33698445-33698445 on Build GRCh38
rs751565270 Chr.11: 33698430-33698430 on Build GRCh38
rs987714346 Chr.11: 33698623-33698623 on Build GRCh38
rs913709971 Chr.11: 33698718-33698718 on Build GRCh38
rs141161902 Chr.11: 33698444-33698444 on Build GRCh38
rs573577302 Chr.11: 33698704-33698704 on Build GRCh38

View Full Product Details