Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33704923-33705429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD59
Entrez Gene ID
966
Gene Name
CD59 molecule
Gene Aliases
16.3A5, 1F5, EJ16, EJ30, EL32, G344, HRF-20, HRF20, MAC-IP, MACIF, MEM43, MIC11, MIN1, MIN2, MIN3, MIRL, MSK21, p18-20
UniGene
Hs.278573
Gene Chromosome Location
Chr.11: 33703010-33736479 on Build GRCh38
Transcripts
NM_000611.5 NM_001127223.1 NM_001127225.1 NM_001127226.1 NM_001127227.1 NM_203329.2 NM_203330.2 NM_203331.2

SNP Detail

SNP ID Chromosome Location
rs568945852 Chr.11: 33705086-33705086 on Build GRCh38
rs938105000 Chr.11: 33705037-33705037 on Build GRCh38
rs146297407 Chr.11: 33705354-33705354 on Build GRCh38
rs565086120 Chr.11: 33704947-33704947 on Build GRCh38
rs976351621 Chr.11: 33704956-33704956 on Build GRCh38
rs531263241 Chr.11: 33705087-33705087 on Build GRCh38
rs764904585 Chr.11: 33705068-33705068 on Build GRCh38
rs958104758 Chr.11: 33705137-33705137 on Build GRCh38
rs558923209 Chr.11: 33705329-33705329 on Build GRCh38
rs1042409422 Chr.11: 33705055-33705056 on Build GRCh38
rs996961798 Chr.11: 33705117-33705117 on Build GRCh38
rs1048444858 Chr.11: 33705050-33705050 on Build GRCh38
rs766820752 Chr.11: 33704981-33704982 on Build GRCh38
rs1037791711 Chr.11: 33704991-33704991 on Build GRCh38
rs561611705 Chr.11: 33705386-33705386 on Build GRCh38
rs985046985 Chr.11: 33705246-33705246 on Build GRCh38
rs561546156 Chr.11: 33705036-33705036 on Build GRCh38
rs1009080059 Chr.11: 33705372-33705372 on Build GRCh38
rs972779096 Chr.11: 33705179-33705179 on Build GRCh38
rs929444963 Chr.11: 33705328-33705328 on Build GRCh38
rs1019212087 Chr.11: 33705114-33705114 on Build GRCh38
rs781723742 Chr.11: 33705247-33705247 on Build GRCh38
rs149287724 Chr.11: 33704945-33704945 on Build GRCh38
rs919484280 Chr.11: 33704961-33704961 on Build GRCh38
rs950928386 Chr.11: 33705210-33705210 on Build GRCh38
rs113743057 Chr.11: 33705198-33705198 on Build GRCh38
rs80074245 Chr.11: 33705038-33705038 on Build GRCh38
rs138742737 Chr.11: 33704944-33704944 on Build GRCh38
rs3204747 Chr.11: 33705346-33705346 on Build GRCh38
rs1032102432 Chr.11: 33705123-33705123 on Build GRCh38
rs965173798 Chr.11: 33705168-33705168 on Build GRCh38
rs1039264276 Chr.11: 33705395-33705395 on Build GRCh38
rs576484897 Chr.11: 33705293-33705293 on Build GRCh38
rs567220391 Chr.11: 33705180-33705180 on Build GRCh38
rs12364970 Chr.11: 33704998-33704998 on Build GRCh38
rs573222618 Chr.11: 33705304-33705304 on Build GRCh38
rs902402990 Chr.11: 33705056-33705056 on Build GRCh38
rs1047922942 Chr.11: 33705349-33705349 on Build GRCh38
rs758162895 Chr.11: 33705116-33705116 on Build GRCh38
rs184127621 Chr.11: 33705222-33705222 on Build GRCh38
rs567350305 Chr.11: 33704977-33704977 on Build GRCh38
rs77695144 Chr.11: 33705143-33705143 on Build GRCh38
rs920728624 Chr.11: 33705314-33705314 on Build GRCh38
rs917971498 Chr.11: 33705339-33705339 on Build GRCh38
rs1042378951 Chr.11: 33705403-33705403 on Build GRCh38
rs7123070 Chr.11: 33705055-33705055 on Build GRCh38
rs572504355 Chr.11: 33705332-33705332 on Build GRCh38
rs570877708 Chr.11: 33705142-33705142 on Build GRCh38
rs1000793588 Chr.11: 33705069-33705069 on Build GRCh38
rs575522601 Chr.11: 33704970-33704970 on Build GRCh38
rs896472425 Chr.11: 33705042-33705042 on Build GRCh38
rs141810136 Chr.11: 33705088-33705088 on Build GRCh38
rs752512293 Chr.11: 33705083-33705083 on Build GRCh38

View Full Product Details