Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33707998-33708544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD59
Entrez Gene ID
966
Gene Name
CD59 molecule
Gene Aliases
16.3A5, 1F5, EJ16, EJ30, EL32, G344, HRF-20, HRF20, MAC-IP, MACIF, MEM43, MIC11, MIN1, MIN2, MIN3, MIRL, MSK21, p18-20
UniGene
Hs.278573
Gene Chromosome Location
Chr.11: 33703010-33736479 on Build GRCh38
Transcripts
NM_000611.5 NM_001127223.1 NM_001127225.1 NM_001127226.1 NM_001127227.1 NM_203329.2 NM_203330.2 NM_203331.2

SNP Detail

SNP ID Chromosome Location
rs566817566 Chr.11: 33708150-33708150 on Build GRCh38
rs150347783 Chr.11: 33708273-33708273 on Build GRCh38
rs774328201 Chr.11: 33708337-33708337 on Build GRCh38
rs181537191 Chr.11: 33708149-33708149 on Build GRCh38
rs767134414 Chr.11: 33708368-33708368 on Build GRCh38
rs571382566 Chr.11: 33708380-33708380 on Build GRCh38
rs11032349 Chr.11: 33708126-33708126 on Build GRCh38
rs11032350 Chr.11: 33708514-33708514 on Build GRCh38
rs915908632 Chr.11: 33708443-33708443 on Build GRCh38
rs948701879 Chr.11: 33708140-33708140 on Build GRCh38
rs997177722 Chr.11: 33708211-33708211 on Build GRCh38
rs955742113 Chr.11: 33708047-33708047 on Build GRCh38
rs975677660 Chr.11: 33708081-33708081 on Build GRCh38
rs974788331 Chr.11: 33708315-33708315 on Build GRCh38
rs908699867 Chr.11: 33708447-33708447 on Build GRCh38
rs140246936 Chr.11: 33708233-33708233 on Build GRCh38
rs1002798975 Chr.11: 33708185-33708185 on Build GRCh38
rs377307129 Chr.11: 33708464-33708464 on Build GRCh38
rs138053542 Chr.11: 33708355-33708355 on Build GRCh38
rs986006547 Chr.11: 33708027-33708027 on Build GRCh38
rs919984048 Chr.11: 33708465-33708465 on Build GRCh38
rs1048524484 Chr.11: 33708129-33708129 on Build GRCh38
rs7130936 Chr.11: 33708459-33708459 on Build GRCh38
rs370661440 Chr.11: 33708423-33708423 on Build GRCh38
rs761664023 Chr.11: 33708353-33708353 on Build GRCh38
rs964756054 Chr.11: 33708068-33708068 on Build GRCh38
rs921485368 Chr.11: 33708087-33708087 on Build GRCh38
rs960007798 Chr.11: 33708404-33708404 on Build GRCh38
rs775836895 Chr.11: 33708398-33708399 on Build GRCh38
rs868527937 Chr.11: 33708449-33708449 on Build GRCh38
rs138698272 Chr.11: 33708090-33708090 on Build GRCh38
rs888629771 Chr.11: 33708131-33708131 on Build GRCh38
rs968595617 Chr.11: 33708049-33708049 on Build GRCh38
rs977304706 Chr.11: 33708352-33708352 on Build GRCh38
rs914462643 Chr.11: 33708042-33708042 on Build GRCh38
rs924570250 Chr.11: 33708088-33708088 on Build GRCh38
rs1044757512 Chr.11: 33708469-33708469 on Build GRCh38
rs74352141 Chr.11: 33708293-33708293 on Build GRCh38
rs528355916 Chr.11: 33708468-33708468 on Build GRCh38
rs964505209 Chr.11: 33708314-33708314 on Build GRCh38
rs908722193 Chr.11: 33708105-33708105 on Build GRCh38
rs866497908 Chr.11: 33708174-33708174 on Build GRCh38
rs977356910 Chr.11: 33708061-33708061 on Build GRCh38
rs1011311989 Chr.11: 33708280-33708280 on Build GRCh38
rs1057232863 Chr.11: 33708113-33708113 on Build GRCh38
rs932163337 Chr.11: 33708466-33708466 on Build GRCh38
rs186279624 Chr.11: 33708215-33708215 on Build GRCh38
rs574471862 Chr.11: 33708253-33708253 on Build GRCh38
rs940223696 Chr.11: 33708461-33708461 on Build GRCh38
rs1050539726 Chr.11: 33708472-33708472 on Build GRCh38
rs190535673 Chr.11: 33708116-33708116 on Build GRCh38
rs1031512352 Chr.11: 33708038-33708038 on Build GRCh38
rs185756495 Chr.11: 33708093-33708093 on Build GRCh38
rs973460898 Chr.11: 33708458-33708458 on Build GRCh38
rs948835089 Chr.11: 33708467-33708467 on Build GRCh38

View Full Product Details