Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33709241-33709742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD59
Entrez Gene ID
966
Gene Name
CD59 molecule
Gene Aliases
16.3A5, 1F5, EJ16, EJ30, EL32, G344, HRF-20, HRF20, MAC-IP, MACIF, MEM43, MIC11, MIN1, MIN2, MIN3, MIRL, MSK21, p18-20
UniGene
Hs.278573
Gene Chromosome Location
Chr.11: 33703010-33736479 on Build GRCh38
Transcripts
NM_000611.5 NM_001127223.1 NM_001127225.1 NM_001127226.1 NM_001127227.1 NM_203329.2 NM_203330.2 NM_203331.2

SNP Detail

SNP ID Chromosome Location
rs941257155 Chr.11: 33709603-33709603 on Build GRCh38
rs979681464 Chr.11: 33709493-33709493 on Build GRCh38
rs569450159 Chr.11: 33709555-33709555 on Build GRCh38
rs564278886 Chr.11: 33709339-33709339 on Build GRCh38
rs897147953 Chr.11: 33709641-33709641 on Build GRCh38
rs994180381 Chr.11: 33709417-33709417 on Build GRCh38
rs533211533 Chr.11: 33709402-33709402 on Build GRCh38
rs1044589118 Chr.11: 33709568-33709568 on Build GRCh38
rs1005481781 Chr.11: 33709393-33709393 on Build GRCh38
rs911404737 Chr.11: 33709450-33709450 on Build GRCh38
rs1044302046 Chr.11: 33709343-33709343 on Build GRCh38
rs560083033 Chr.11: 33709451-33709451 on Build GRCh38
rs529236235 Chr.11: 33709472-33709472 on Build GRCh38
rs142715542 Chr.11: 33709272-33709272 on Build GRCh38
rs1014542627 Chr.11: 33709310-33709310 on Build GRCh38
rs1029737672 Chr.11: 33709424-33709424 on Build GRCh38
rs1052862509 Chr.11: 33709274-33709274 on Build GRCh38
rs979186125 Chr.11: 33709667-33709667 on Build GRCh38
rs1035861805 Chr.11: 33709498-33709498 on Build GRCh38
rs895641978 Chr.11: 33709283-33709283 on Build GRCh38
rs921850877 Chr.11: 33709708-33709708 on Build GRCh38
rs1027266909 Chr.11: 33709416-33709417 on Build GRCh38
rs185837701 Chr.11: 33709435-33709435 on Build GRCh38
rs905804839 Chr.11: 33709362-33709362 on Build GRCh38
rs181309215 Chr.11: 33709282-33709282 on Build GRCh38
rs538472463 Chr.11: 33709560-33709560 on Build GRCh38
rs1035979594 Chr.11: 33709285-33709285 on Build GRCh38
rs955566499 Chr.11: 33709432-33709432 on Build GRCh38
rs934358008 Chr.11: 33709269-33709269 on Build GRCh38
rs898870122 Chr.11: 33709331-33709331 on Build GRCh38
rs934348184 Chr.11: 33709549-33709549 on Build GRCh38
rs996143907 Chr.11: 33709374-33709374 on Build GRCh38
rs917026197 Chr.11: 33709558-33709558 on Build GRCh38
rs769451757 Chr.11: 33709608-33709608 on Build GRCh38
rs1016867424 Chr.11: 33709395-33709395 on Build GRCh38
rs960075504 Chr.11: 33709525-33709525 on Build GRCh38
rs116176729 Chr.11: 33709541-33709541 on Build GRCh38
rs1029741879 Chr.11: 33709683-33709683 on Build GRCh38
rs3209285 Chr.11: 33709438-33709438 on Build GRCh38
rs191085190 Chr.11: 33709661-33709661 on Build GRCh38
rs905698218 Chr.11: 33709600-33709600 on Build GRCh38
rs765228692 Chr.11: 33709564-33709564 on Build GRCh38

View Full Product Details