Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34453659-34454157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAT
Entrez Gene ID
847
Gene Name
catalase
Gene Aliases
-
UniGene
Hs.502302
Gene Chromosome Location
Chr.11: 34438925-34472060 on Build GRCh38
Transcripts
NM_001752.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4612678 c.711_711+1delGG p.? Chr.11: 34453926-34453927 on Build GRCh38
3446966 c.703C>T p.H235Y Chr.11: 34453918-34453918 on Build GRCh38
3446965 c.616C>T p.P206S Chr.11: 34453831-34453831 on Build GRCh38
7168564 c.679G>A p.G227R Chr.11: 34453894-34453894 on Build GRCh38
5477560 c.657T>G p.T219T Chr.11: 34453872-34453872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965287892 Chr.11: 34453978-34453978 on Build GRCh38
rs757152645 Chr.11: 34453836-34453836 on Build GRCh38
rs752283917 Chr.11: 34453786-34453786 on Build GRCh38
rs991517680 Chr.11: 34454037-34454037 on Build GRCh38
rs369304814 Chr.11: 34453710-34453710 on Build GRCh38
rs937566735 Chr.11: 34453820-34453820 on Build GRCh38
rs889556144 Chr.11: 34454092-34454092 on Build GRCh38
rs199514112 Chr.11: 34453752-34453752 on Build GRCh38
rs1053958723 Chr.11: 34454117-34454117 on Build GRCh38
rs187490567 Chr.11: 34453972-34453972 on Build GRCh38
rs755722836 Chr.11: 34454096-34454096 on Build GRCh38
rs537291766 Chr.11: 34453739-34453739 on Build GRCh38
rs753516945 Chr.11: 34453929-34453929 on Build GRCh38
rs999525761 Chr.11: 34453919-34453919 on Build GRCh38
rs74906502 Chr.11: 34454101-34454101 on Build GRCh38
rs1006492374 Chr.11: 34454125-34454125 on Build GRCh38
rs539218654 Chr.11: 34454068-34454068 on Build GRCh38
rs976550888 Chr.11: 34453979-34453979 on Build GRCh38
rs766677570 Chr.11: 34453869-34453869 on Build GRCh38
rs922268689 Chr.11: 34454035-34454035 on Build GRCh38
rs530197904 Chr.11: 34453687-34453687 on Build GRCh38
rs749134062 Chr.11: 34453830-34453830 on Build GRCh38
rs759966400 Chr.11: 34453906-34453906 on Build GRCh38
rs61733252 Chr.11: 34453896-34453896 on Build GRCh38
rs757024376 Chr.11: 34453931-34453931 on Build GRCh38
rs755744483 Chr.11: 34453924-34453924 on Build GRCh38
rs748286212 Chr.11: 34453964-34453964 on Build GRCh38
rs574208281 Chr.11: 34453920-34453920 on Build GRCh38
rs1018133229 Chr.11: 34453976-34453976 on Build GRCh38
rs960785468 Chr.11: 34453954-34453954 on Build GRCh38
rs556723718 Chr.11: 34454011-34454011 on Build GRCh38
rs763370887 Chr.11: 34453855-34453855 on Build GRCh38
rs781318238 Chr.11: 34453951-34453951 on Build GRCh38
rs766950326 Chr.11: 34453784-34453784 on Build GRCh38
rs763779127 Chr.11: 34453927-34453927 on Build GRCh38
rs778583616 Chr.11: 34453935-34453935 on Build GRCh38
rs533575957 Chr.11: 34453853-34453853 on Build GRCh38
rs767190535 Chr.11: 34453909-34453909 on Build GRCh38
rs199892309 Chr.11: 34453806-34453806 on Build GRCh38
rs183080448 Chr.11: 34454112-34454112 on Build GRCh38
rs200989008 Chr.11: 34453839-34453839 on Build GRCh38
rs1036658994 Chr.11: 34454032-34454032 on Build GRCh38
rs17886248 Chr.11: 34453692-34453692 on Build GRCh38
rs779036767 Chr.11: 34453823-34453823 on Build GRCh38
rs889964408 Chr.11: 34453874-34453874 on Build GRCh38
rs374654105 Chr.11: 34453854-34453854 on Build GRCh38
rs147148220 Chr.11: 34453840-34453840 on Build GRCh38
rs748380856 Chr.11: 34453977-34453979 on Build GRCh38
rs137989905 Chr.11: 34453940-34453940 on Build GRCh38
rs752039722 Chr.11: 34453751-34453751 on Build GRCh38
rs201484998 Chr.11: 34453823-34453823 on Build GRCh38
rs902762844 Chr.11: 34454107-34454107 on Build GRCh38
rs939687415 Chr.11: 34453693-34453693 on Build GRCh38
rs372830902 Chr.11: 34454065-34454065 on Build GRCh38
rs767171579 Chr.11: 34454118-34454118 on Build GRCh38
rs183360403 Chr.11: 34453968-34453968 on Build GRCh38
rs371809582 Chr.11: 34453843-34453843 on Build GRCh38
rs755836244 Chr.11: 34453816-34453816 on Build GRCh38
rs915720823 Chr.11: 34453997-34453997 on Build GRCh38
rs768858587 Chr.11: 34453772-34453773 on Build GRCh38
rs752265868 Chr.11: 34453923-34453923 on Build GRCh38
rs969899886 Chr.11: 34454006-34454006 on Build GRCh38
rs748376031 Chr.11: 34453845-34453845 on Build GRCh38
rs758333161 Chr.11: 34453944-34453944 on Build GRCh38
rs939970615 Chr.11: 34454031-34454031 on Build GRCh38
rs192738550 Chr.11: 34454039-34454039 on Build GRCh38
rs576686491 Chr.11: 34453727-34453727 on Build GRCh38
rs149450955 Chr.11: 34453822-34453822 on Build GRCh38
rs750035465 Chr.11: 34453989-34453989 on Build GRCh38
rs909791407 Chr.11: 34454030-34454030 on Build GRCh38
rs754462351 Chr.11: 34453771-34453771 on Build GRCh38
rs778092269 Chr.11: 34453970-34453970 on Build GRCh38
rs2266628 Chr.11: 34453745-34453745 on Build GRCh38
rs1000189640 Chr.11: 34454111-34454111 on Build GRCh38
rs559462171 Chr.11: 34453948-34453948 on Build GRCh38
rs893584217 Chr.11: 34454125-34454127 on Build GRCh38

View Full Product Details