Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34478706-34479215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELF5
Entrez Gene ID
2001
Gene Name
E74 like ETS transcription factor 5
Gene Aliases
ESE2
UniGene
Hs.11713
Gene Chromosome Location
Chr.11: 34478791-34513800 on Build GRCh38
Transcripts
NM_001243080.1 NM_001243081.1 NM_001422.3 NM_198381.1 XM_017017308.1 XM_017017309.1

SNP Detail

SNP ID Chromosome Location
rs562421935 Chr.11: 34478739-34478739 on Build GRCh38
rs949702299 Chr.11: 34478769-34478769 on Build GRCh38
rs1006328183 Chr.11: 34478851-34478851 on Build GRCh38
rs986292278 Chr.11: 34479183-34479183 on Build GRCh38
rs72556400 Chr.11: 34479003-34479003 on Build GRCh38
rs901810661 Chr.11: 34478881-34478881 on Build GRCh38
rs542167150 Chr.11: 34478870-34478870 on Build GRCh38
rs770391964 Chr.11: 34478869-34478869 on Build GRCh38
rs997637521 Chr.11: 34479153-34479153 on Build GRCh38
rs888979323 Chr.11: 34478843-34478843 on Build GRCh38
rs375649297 Chr.11: 34479150-34479150 on Build GRCh38
rs72556401 Chr.11: 34478928-34478928 on Build GRCh38
rs72556402 Chr.11: 34478784-34478784 on Build GRCh38
rs917010525 Chr.11: 34479043-34479043 on Build GRCh38
rs952414771 Chr.11: 34478937-34478937 on Build GRCh38
rs896326382 Chr.11: 34478772-34478772 on Build GRCh38
rs951684874 Chr.11: 34478831-34478831 on Build GRCh38
rs1034250103 Chr.11: 34479000-34479000 on Build GRCh38
rs977511487 Chr.11: 34479101-34479101 on Build GRCh38
rs762535926 Chr.11: 34478900-34478900 on Build GRCh38
rs947902555 Chr.11: 34479076-34479076 on Build GRCh38
rs896316569 Chr.11: 34479067-34479067 on Build GRCh38
rs10312 Chr.11: 34478806-34478806 on Build GRCh38
rs1535719 Chr.11: 34479019-34479019 on Build GRCh38
rs924646054 Chr.11: 34479010-34479010 on Build GRCh38
rs897498368 Chr.11: 34479152-34479152 on Build GRCh38
rs147166108 Chr.11: 34478816-34478816 on Build GRCh38
rs763761051 Chr.11: 34478948-34478948 on Build GRCh38
rs1323690 Chr.11: 34479140-34479140 on Build GRCh38
rs766027879 Chr.11: 34478971-34478971 on Build GRCh38
rs947398006 Chr.11: 34478991-34478991 on Build GRCh38
rs184022036 Chr.11: 34479182-34479182 on Build GRCh38
rs544560536 Chr.11: 34478940-34478940 on Build GRCh38
rs986344889 Chr.11: 34478946-34478946 on Build GRCh38
rs1054678919 Chr.11: 34478817-34478817 on Build GRCh38
rs539665408 Chr.11: 34478791-34478791 on Build GRCh38
rs1030078795 Chr.11: 34479159-34479159 on Build GRCh38
rs1044990484 Chr.11: 34479080-34479080 on Build GRCh38
rs949672666 Chr.11: 34479063-34479063 on Build GRCh38
rs1057218997 Chr.11: 34479036-34479036 on Build GRCh38
rs573094916 Chr.11: 34478852-34478852 on Build GRCh38
rs140445350 Chr.11: 34479139-34479139 on Build GRCh38
rs975871811 Chr.11: 34478915-34478915 on Build GRCh38
rs953551926 Chr.11: 34478929-34478929 on Build GRCh38
rs1535720 Chr.11: 34478884-34478884 on Build GRCh38

View Full Product Details