Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 36273842-36274340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD9
Entrez Gene ID
29099
Gene Name
COMM domain containing 9
Gene Aliases
HSPC166
UniGene
Hs.279836
Gene Chromosome Location
Chr.11: 36272292-36289449 on Build GRCh38
Transcripts
NM_001101653.1 NM_001307932.1 NM_001307937.1 NM_014186.3 XM_017017625.1

SNP Detail

SNP ID Chromosome Location
rs752449001 Chr.11: 36274143-36274143 on Build GRCh38
rs539326907 Chr.11: 36274208-36274208 on Build GRCh38
rs962631830 Chr.11: 36274058-36274058 on Build GRCh38
rs936637445 Chr.11: 36274317-36274317 on Build GRCh38
rs185733066 Chr.11: 36273881-36273881 on Build GRCh38
rs1001153700 Chr.11: 36273967-36273967 on Build GRCh38
rs1050815773 Chr.11: 36274218-36274218 on Build GRCh38
rs576515054 Chr.11: 36274123-36274123 on Build GRCh38
rs147014879 Chr.11: 36274262-36274262 on Build GRCh38
rs995434818 Chr.11: 36274258-36274258 on Build GRCh38
rs542196750 Chr.11: 36273987-36273987 on Build GRCh38
rs890858736 Chr.11: 36274233-36274233 on Build GRCh38
rs914147323 Chr.11: 36274155-36274155 on Build GRCh38
rs1052617933 Chr.11: 36273934-36273934 on Build GRCh38
rs930095028 Chr.11: 36273884-36273884 on Build GRCh38
rs996625528 Chr.11: 36274215-36274215 on Build GRCh38
rs969485683 Chr.11: 36274095-36274095 on Build GRCh38
rs1052418517 Chr.11: 36274146-36274146 on Build GRCh38
rs1013363155 Chr.11: 36274031-36274031 on Build GRCh38
rs560834860 Chr.11: 36274087-36274087 on Build GRCh38
rs764788404 Chr.11: 36274096-36274096 on Build GRCh38
rs879704934 Chr.11: 36274207-36274207 on Build GRCh38
rs138172093 Chr.11: 36274277-36274277 on Build GRCh38
rs1044320975 Chr.11: 36274180-36274180 on Build GRCh38
rs960744591 Chr.11: 36274318-36274318 on Build GRCh38
rs1015946834 Chr.11: 36274032-36274032 on Build GRCh38
rs1029366295 Chr.11: 36274017-36274017 on Build GRCh38
rs955013177 Chr.11: 36274022-36274022 on Build GRCh38
rs980313682 Chr.11: 36274108-36274108 on Build GRCh38
rs868699837 Chr.11: 36274069-36274069 on Build GRCh38
rs903790111 Chr.11: 36273965-36273965 on Build GRCh38
rs879586489 Chr.11: 36274097-36274097 on Build GRCh38
rs946948597 Chr.11: 36274178-36274178 on Build GRCh38
rs927991293 Chr.11: 36274142-36274142 on Build GRCh38
rs974722554 Chr.11: 36274186-36274186 on Build GRCh38
rs962950483 Chr.11: 36274251-36274251 on Build GRCh38
rs757997874 Chr.11: 36274189-36274189 on Build GRCh38
rs903651698 Chr.11: 36274297-36274297 on Build GRCh38
rs939633399 Chr.11: 36273887-36273887 on Build GRCh38
rs942535730 Chr.11: 36273941-36273941 on Build GRCh38
rs1039913812 Chr.11: 36273952-36273952 on Build GRCh38
rs915707026 Chr.11: 36274091-36274091 on Build GRCh38
rs556360963 Chr.11: 36274045-36274045 on Build GRCh38
rs758577042 Chr.11: 36273930-36273930 on Build GRCh38
rs986875964 Chr.11: 36274219-36274219 on Build GRCh38
rs912659538 Chr.11: 36274252-36274252 on Build GRCh38
rs1003896995 Chr.11: 36274242-36274243 on Build GRCh38
rs552625646 Chr.11: 36274222-36274222 on Build GRCh38
rs969513164 Chr.11: 36274086-36274086 on Build GRCh38
rs899580923 Chr.11: 36273973-36273973 on Build GRCh38
rs890983601 Chr.11: 36273937-36273937 on Build GRCh38
rs1015225759 Chr.11: 36274247-36274247 on Build GRCh38
rs1051123256 Chr.11: 36273915-36273915 on Build GRCh38
rs929961067 Chr.11: 36274197-36274197 on Build GRCh38
rs969845112 Chr.11: 36274273-36274273 on Build GRCh38
rs547515077 Chr.11: 36273880-36273880 on Build GRCh38
rs6484833 Chr.11: 36273982-36273982 on Build GRCh38
rs376886745 Chr.11: 36274282-36274282 on Build GRCh38
rs759998299 Chr.11: 36274076-36274076 on Build GRCh38
rs962999297 Chr.11: 36274145-36274145 on Build GRCh38
rs1030682743 Chr.11: 36273981-36273981 on Build GRCh38

View Full Product Details