Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 36597920-36598439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf74
Entrez Gene ID
119710
Gene Name
chromosome 11 open reading frame 74
Gene Aliases
HEPIS
UniGene
Hs.159376
Gene Chromosome Location
Chr.11: 36594245-36659291 on Build GRCh38
Transcripts
NM_001276722.1 NM_001276723.1 NM_001276724.1 NM_001276725.1 NM_001276726.1 NM_001276727.1 NM_138787.3 NR_077243.1 XM_006718138.3 XM_011519885.2 NM_000536.3 NM_001243785.1 NM_001243786.1

Gene Symbol
RAG2
Entrez Gene ID
5897
Gene Name
recombination activating gene 2
Gene Aliases
RAG-2
UniGene
Hs.714519
Gene Chromosome Location
Chr.11: 36591943-36598279 on Build GRCh38
Transcripts
NM_001276722.1 NM_001276723.1 NM_001276724.1 NM_001276725.1 NM_001276726.1 NM_001276727.1 NM_138787.3 NR_077243.1 XM_006718138.3 XM_011519885.2 NM_000536.3 NM_001243785.1 NM_001243786.1

SNP Detail

SNP ID Chromosome Location
rs1035897449 Chr.11: 36598011-36598011 on Build GRCh38
rs1015641905 Chr.11: 36598181-36598181 on Build GRCh38
rs945172392 Chr.11: 36598343-36598343 on Build GRCh38
rs959823782 Chr.11: 36598335-36598335 on Build GRCh38
rs1013048316 Chr.11: 36598032-36598032 on Build GRCh38
rs1057130622 Chr.11: 36598125-36598125 on Build GRCh38
rs893530110 Chr.11: 36597978-36597978 on Build GRCh38
rs569611522 Chr.11: 36598319-36598319 on Build GRCh38
rs1003119954 Chr.11: 36598010-36598010 on Build GRCh38
rs989297357 Chr.11: 36598286-36598286 on Build GRCh38
rs906244893 Chr.11: 36598369-36598369 on Build GRCh38
rs1042357300 Chr.11: 36598167-36598167 on Build GRCh38
rs938180613 Chr.11: 36598063-36598064 on Build GRCh38
rs185359195 Chr.11: 36598279-36598279 on Build GRCh38
rs1019954487 Chr.11: 36598243-36598243 on Build GRCh38
rs753211137 Chr.11: 36598389-36598389 on Build GRCh38
rs952798055 Chr.11: 36598147-36598147 on Build GRCh38
rs1048578801 Chr.11: 36598408-36598408 on Build GRCh38
rs759799357 Chr.11: 36598238-36598238 on Build GRCh38
rs994485238 Chr.11: 36598273-36598273 on Build GRCh38
rs536995581 Chr.11: 36598126-36598126 on Build GRCh38
rs547663028 Chr.11: 36598172-36598172 on Build GRCh38
rs1000988323 Chr.11: 36598197-36598197 on Build GRCh38
rs906192677 Chr.11: 36597996-36597996 on Build GRCh38
rs945681758 Chr.11: 36598375-36598375 on Build GRCh38
rs567981266 Chr.11: 36598206-36598206 on Build GRCh38
rs927017760 Chr.11: 36598307-36598307 on Build GRCh38
rs965362659 Chr.11: 36598192-36598192 on Build GRCh38
rs925294750 Chr.11: 36598403-36598403 on Build GRCh38
rs961355375 Chr.11: 36597979-36597979 on Build GRCh38
rs527948817 Chr.11: 36598166-36598166 on Build GRCh38
rs886048273 Chr.11: 36598225-36598225 on Build GRCh38
rs73455545 Chr.11: 36598211-36598211 on Build GRCh38
rs770944720 Chr.11: 36597984-36597984 on Build GRCh38
rs199509458 Chr.11: 36598300-36598300 on Build GRCh38
rs982846165 Chr.11: 36598171-36598171 on Build GRCh38
rs564950586 Chr.11: 36598050-36598050 on Build GRCh38
rs894644853 Chr.11: 36598028-36598028 on Build GRCh38
rs894703484 Chr.11: 36598405-36598405 on Build GRCh38
rs945047490 Chr.11: 36597980-36597980 on Build GRCh38
rs932493947 Chr.11: 36598225-36598225 on Build GRCh38
rs765337106 Chr.11: 36598383-36598383 on Build GRCh38
rs538742576 Chr.11: 36598336-36598336 on Build GRCh38
rs1013515837 Chr.11: 36598407-36598407 on Build GRCh38
rs961286531 Chr.11: 36598271-36598271 on Build GRCh38
rs1027502310 Chr.11: 36598145-36598145 on Build GRCh38
rs926817305 Chr.11: 36598051-36598051 on Build GRCh38
rs919687527 Chr.11: 36598008-36598008 on Build GRCh38
rs373459658 Chr.11: 36597943-36597943 on Build GRCh38
rs952364448 Chr.11: 36598299-36598300 on Build GRCh38
rs573719595 Chr.11: 36598359-36598359 on Build GRCh38
rs1042069006 Chr.11: 36597985-36597985 on Build GRCh38

View Full Product Details