Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 36609948-36610456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf74
Entrez Gene ID
119710
Gene Name
chromosome 11 open reading frame 74
Gene Aliases
HEPIS
UniGene
Hs.159376
Gene Chromosome Location
Chr.11: 36594245-36659291 on Build GRCh38
Transcripts
NM_001276722.1 NM_001276723.1 NM_001276724.1 NM_001276725.1 NM_001276726.1 NM_001276727.1 NM_138787.3 NR_077243.1 XM_006718138.3 XM_011519885.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4911617 c.89A>G p.Q30R Chr.11: 36610192-36610192 on Build GRCh38
3447104 c.72C>T p.F24F Chr.11: 36610175-36610175 on Build GRCh38
4793213 c.133C>T p.Q45* Chr.11: 36610236-36610236 on Build GRCh38
4807202 c.58G>A p.V20I Chr.11: 36610161-36610161 on Build GRCh38
3368358 c.77A>G p.N26S Chr.11: 36610180-36610180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897625961 Chr.11: 36610417-36610417 on Build GRCh38
rs769162533 Chr.11: 36610254-36610259 on Build GRCh38
rs544704143 Chr.11: 36610351-36610351 on Build GRCh38
rs138838269 Chr.11: 36610235-36610235 on Build GRCh38
rs774873017 Chr.11: 36610134-36610134 on Build GRCh38
rs763820764 Chr.11: 36610154-36610154 on Build GRCh38
rs913376917 Chr.11: 36610326-36610329 on Build GRCh38
rs374022577 Chr.11: 36610140-36610140 on Build GRCh38
rs781404218 Chr.11: 36610110-36610110 on Build GRCh38
rs113645801 Chr.11: 36610275-36610275 on Build GRCh38
rs370463806 Chr.11: 36610137-36610137 on Build GRCh38
rs752758339 Chr.11: 36610176-36610176 on Build GRCh38
rs928399657 Chr.11: 36610359-36610359 on Build GRCh38
rs12294126 Chr.11: 36610160-36610160 on Build GRCh38
rs148923154 Chr.11: 36610350-36610350 on Build GRCh38
rs377590377 Chr.11: 36610111-36610111 on Build GRCh38
rs927664867 Chr.11: 36610240-36610240 on Build GRCh38
rs974742735 Chr.11: 36610228-36610228 on Build GRCh38
rs749630393 Chr.11: 36610120-36610120 on Build GRCh38
rs755033051 Chr.11: 36610108-36610108 on Build GRCh38
rs181253019 Chr.11: 36610114-36610114 on Build GRCh38
rs757583767 Chr.11: 36610236-36610236 on Build GRCh38
rs768458231 Chr.11: 36610276-36610276 on Build GRCh38
rs760796969 Chr.11: 36610390-36610390 on Build GRCh38
rs537523655 Chr.11: 36610123-36610123 on Build GRCh38
rs776100252 Chr.11: 36610064-36610065 on Build GRCh38
rs937067328 Chr.11: 36610386-36610386 on Build GRCh38
rs12281063 Chr.11: 36610327-36610327 on Build GRCh38
rs755013993 Chr.11: 36610166-36610166 on Build GRCh38
rs1045480980 Chr.11: 36609990-36609990 on Build GRCh38
rs773782641 Chr.11: 36610060-36610060 on Build GRCh38
rs756376720 Chr.11: 36610186-36610186 on Build GRCh38
rs767099351 Chr.11: 36610070-36610070 on Build GRCh38
rs776370146 Chr.11: 36610291-36610291 on Build GRCh38
rs374843985 Chr.11: 36610098-36610098 on Build GRCh38
rs776611331 Chr.11: 36610158-36610158 on Build GRCh38
rs375243176 Chr.11: 36610250-36610250 on Build GRCh38
rs771456163 Chr.11: 36610059-36610059 on Build GRCh38
rs779293690 Chr.11: 36610239-36610239 on Build GRCh38
rs906888434 Chr.11: 36610015-36610015 on Build GRCh38
rs61745970 Chr.11: 36610161-36610161 on Build GRCh38
rs868456534 Chr.11: 36610426-36610426 on Build GRCh38
rs531626972 Chr.11: 36610242-36610242 on Build GRCh38
rs535628899 Chr.11: 36610092-36610092 on Build GRCh38
rs1018960619 Chr.11: 36610207-36610207 on Build GRCh38
rs77576907 Chr.11: 36610317-36610317 on Build GRCh38
rs140413313 Chr.11: 36610116-36610116 on Build GRCh38
rs765193976 Chr.11: 36610093-36610093 on Build GRCh38
rs950914644 Chr.11: 36610181-36610181 on Build GRCh38
rs966080613 Chr.11: 36610224-36610224 on Build GRCh38
rs753675107 Chr.11: 36610086-36610086 on Build GRCh38
rs764049950 Chr.11: 36610081-36610081 on Build GRCh38
rs1014790849 Chr.11: 36610397-36610397 on Build GRCh38
rs186405840 Chr.11: 36610395-36610395 on Build GRCh38
rs567200799 Chr.11: 36610077-36610077 on Build GRCh38
rs779761721 Chr.11: 36610258-36610258 on Build GRCh38
rs777940591 Chr.11: 36610201-36610201 on Build GRCh38
rs759047637 Chr.11: 36610061-36610061 on Build GRCh38
rs770865857 Chr.11: 36610065-36610065 on Build GRCh38
rs892081526 Chr.11: 36610044-36610044 on Build GRCh38
rs1050375850 Chr.11: 36610380-36610380 on Build GRCh38
rs1049749897 Chr.11: 36610430-36610430 on Build GRCh38
rs749858622 Chr.11: 36610055-36610055 on Build GRCh38
rs746455184 Chr.11: 36610247-36610247 on Build GRCh38
rs1008763586 Chr.11: 36610135-36610135 on Build GRCh38
rs540268800 Chr.11: 36610171-36610171 on Build GRCh38

View Full Product Details