Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 43943328-43943835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACCSL
Entrez Gene ID
390110
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive) like
Gene Aliases
-
UniGene
Hs.558851
Gene Chromosome Location
Chr.11: 43920987-44060667 on Build GRCh38
Transcripts
XM_017017714.1 NM_001145033.1

Gene Symbol
C11orf96
Entrez Gene ID
387763
Gene Name
chromosome 11 open reading frame 96
Gene Aliases
AG2
UniGene
Hs.530443
Gene Chromosome Location
Chr.11: 43942556-43943883 on Build GRCh38
Transcripts
XM_017017714.1 NM_001145033.1

SNP Detail

SNP ID Chromosome Location
rs977968561 Chr.11: 43943745-43943745 on Build GRCh38
rs11545442 Chr.11: 43943509-43943509 on Build GRCh38
rs754488008 Chr.11: 43943741-43943741 on Build GRCh38
rs1038653006 Chr.11: 43943497-43943497 on Build GRCh38
rs959237631 Chr.11: 43943688-43943688 on Build GRCh38
rs571100467 Chr.11: 43943760-43943760 on Build GRCh38
rs546141426 Chr.11: 43943429-43943429 on Build GRCh38
rs750934442 Chr.11: 43943399-43943399 on Build GRCh38
rs994655485 Chr.11: 43943381-43943381 on Build GRCh38
rs969161963 Chr.11: 43943548-43943548 on Build GRCh38
rs745460318 Chr.11: 43943486-43943486 on Build GRCh38
rs941621587 Chr.11: 43943622-43943622 on Build GRCh38
rs761992412 Chr.11: 43943607-43943607 on Build GRCh38
rs999374094 Chr.11: 43943472-43943472 on Build GRCh38
rs928161991 Chr.11: 43943396-43943396 on Build GRCh38
rs9846 Chr.11: 43943458-43943458 on Build GRCh38
rs934079597 Chr.11: 43943678-43943678 on Build GRCh38
rs982292220 Chr.11: 43943560-43943560 on Build GRCh38
rs1050730873 Chr.11: 43943474-43943474 on Build GRCh38
rs184122792 Chr.11: 43943385-43943385 on Build GRCh38
rs538529722 Chr.11: 43943771-43943771 on Build GRCh38
rs7524 Chr.11: 43943348-43943348 on Build GRCh38
rs1051588917 Chr.11: 43943713-43943713 on Build GRCh38
rs764325387 Chr.11: 43943567-43943567 on Build GRCh38
rs969488784 Chr.11: 43943799-43943799 on Build GRCh38
rs542323765 Chr.11: 43943379-43943379 on Build GRCh38
rs982929049 Chr.11: 43943574-43943574 on Build GRCh38
rs758844284 Chr.11: 43943663-43943663 on Build GRCh38
rs930209757 Chr.11: 43943406-43943406 on Build GRCh38
rs572237123 Chr.11: 43943413-43943413 on Build GRCh38
rs994376932 Chr.11: 43943511-43943511 on Build GRCh38
rs893720232 Chr.11: 43943780-43943780 on Build GRCh38
rs749051169 Chr.11: 43943784-43943784 on Build GRCh38
rs999321557 Chr.11: 43943753-43943753 on Build GRCh38
rs898650192 Chr.11: 43943500-43943500 on Build GRCh38
rs531347119 Chr.11: 43943446-43943446 on Build GRCh38
rs556313454 Chr.11: 43943790-43943790 on Build GRCh38
rs1039698712 Chr.11: 43943641-43943641 on Build GRCh38
rs760708884 Chr.11: 43943563-43943563 on Build GRCh38
rs758941167 Chr.11: 43943402-43943402 on Build GRCh38
rs780826961 Chr.11: 43943742-43943742 on Build GRCh38
rs7118458 Chr.11: 43943448-43943448 on Build GRCh38
rs773024634 Chr.11: 43943482-43943482 on Build GRCh38
rs766841240 Chr.11: 43943682-43943682 on Build GRCh38
rs971793327 Chr.11: 43943387-43943387 on Build GRCh38
rs535373119 Chr.11: 43943436-43943436 on Build GRCh38
rs1011280384 Chr.11: 43943787-43943787 on Build GRCh38
rs761709566 Chr.11: 43943663-43943664 on Build GRCh38
rs1030661491 Chr.11: 43943349-43943349 on Build GRCh38
rs768425726 Chr.11: 43943444-43943444 on Build GRCh38
rs898538313 Chr.11: 43943380-43943380 on Build GRCh38
rs933634214 Chr.11: 43943469-43943469 on Build GRCh38
rs574884322 Chr.11: 43943366-43943366 on Build GRCh38
rs951519778 Chr.11: 43943568-43943568 on Build GRCh38
rs888709378 Chr.11: 43943430-43943430 on Build GRCh38
rs552452517 Chr.11: 43943744-43943744 on Build GRCh38
rs189499465 Chr.11: 43943492-43943492 on Build GRCh38
rs774945097 Chr.11: 43943577-43943577 on Build GRCh38
rs578128835 Chr.11: 43943767-43943767 on Build GRCh38
rs764092105 Chr.11: 43943415-43943415 on Build GRCh38
rs11545443 Chr.11: 43943781-43943781 on Build GRCh38
rs148586972 Chr.11: 43943519-43943519 on Build GRCh38
rs760077700 Chr.11: 43943603-43943603 on Build GRCh38
rs1023755825 Chr.11: 43943544-43943544 on Build GRCh38
rs971481394 Chr.11: 43943398-43943398 on Build GRCh38
rs534097055 Chr.11: 43943644-43943644 on Build GRCh38
rs959291175 Chr.11: 43943418-43943418 on Build GRCh38
rs769522036 Chr.11: 43943462-43943462 on Build GRCh38
rs529007038 Chr.11: 43943490-43943490 on Build GRCh38
rs770885909 Chr.11: 43943528-43943528 on Build GRCh38
rs754112448 Chr.11: 43943601-43943601 on Build GRCh38
rs957945348 Chr.11: 43943483-43943483 on Build GRCh38
rs990619690 Chr.11: 43943422-43943422 on Build GRCh38
rs991142215 Chr.11: 43943715-43943715 on Build GRCh38
rs910096290 Chr.11: 43943609-43943609 on Build GRCh38
rs749632773 Chr.11: 43943481-43943481 on Build GRCh38
rs771571091 Chr.11: 43943526-43943526 on Build GRCh38
rs751144177 Chr.11: 43943731-43943731 on Build GRCh38
rs765588765 Chr.11: 43943620-43943620 on Build GRCh38
rs907307260 Chr.11: 43943602-43943602 on Build GRCh38
rs774226782 Chr.11: 43943793-43943793 on Build GRCh38
rs775553621 Chr.11: 43943531-43943531 on Build GRCh38
rs890324271 Chr.11: 43943470-43943470 on Build GRCh38
rs957665641 Chr.11: 43943362-43943362 on Build GRCh38

View Full Product Details