Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44618521-44619041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD82
Entrez Gene ID
3732
Gene Name
CD82 molecule
Gene Aliases
4F9, C33, GR15, IA4, KAI1, R2, SAR2, ST6, TSPAN27
UniGene
Hs.527778
Gene Chromosome Location
Chr.11: 44565591-44620363 on Build GRCh38
Transcripts
NM_001024844.1 NM_002231.3 XM_006718223.2 XM_011520067.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1979353 c.686G>T p.G229V Chr.11: 44618683-44618683 on Build GRCh38
3967505 c.654G>A p.E218E Chr.11: 44618651-44618651 on Build GRCh38
926998 c.672G>A p.L224L Chr.11: 44618669-44618669 on Build GRCh38
7360312 c.709G>T p.G237C Chr.11: 44618706-44618706 on Build GRCh38
3752367 c.721A>G p.I241V Chr.11: 44618718-44618718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778562920 Chr.11: 44618646-44618646 on Build GRCh38
rs1047951966 Chr.11: 44618857-44618857 on Build GRCh38
rs374096200 Chr.11: 44618601-44618601 on Build GRCh38
rs371497237 Chr.11: 44618706-44618706 on Build GRCh38
rs779543798 Chr.11: 44618593-44618593 on Build GRCh38
rs911970440 Chr.11: 44618771-44618771 on Build GRCh38
rs376406578 Chr.11: 44619006-44619006 on Build GRCh38
rs115163271 Chr.11: 44618909-44618909 on Build GRCh38
rs768215987 Chr.11: 44618599-44618599 on Build GRCh38
rs758381473 Chr.11: 44618756-44618756 on Build GRCh38
rs141512107 Chr.11: 44619007-44619007 on Build GRCh38
rs201290700 Chr.11: 44618662-44618662 on Build GRCh38
rs899428880 Chr.11: 44618639-44618639 on Build GRCh38
rs776567252 Chr.11: 44618679-44618679 on Build GRCh38
rs990647138 Chr.11: 44618551-44618551 on Build GRCh38
rs765384989 Chr.11: 44618687-44618687 on Build GRCh38
rs200441128 Chr.11: 44618622-44618622 on Build GRCh38
rs762323541 Chr.11: 44618932-44618932 on Build GRCh38
rs114663838 Chr.11: 44618969-44618969 on Build GRCh38
rs74375347 Chr.11: 44618675-44618675 on Build GRCh38
rs780823917 Chr.11: 44618699-44618699 on Build GRCh38
rs748913627 Chr.11: 44618704-44618704 on Build GRCh38
rs991701933 Chr.11: 44618885-44618885 on Build GRCh38
rs375271738 Chr.11: 44618997-44618997 on Build GRCh38
rs554846063 Chr.11: 44618655-44618655 on Build GRCh38
rs575112343 Chr.11: 44618623-44618623 on Build GRCh38
rs375058488 Chr.11: 44618732-44618732 on Build GRCh38
rs542337864 Chr.11: 44618657-44618657 on Build GRCh38
rs924596603 Chr.11: 44618580-44618580 on Build GRCh38
rs751061937 Chr.11: 44618631-44618631 on Build GRCh38
rs915176387 Chr.11: 44618567-44618567 on Build GRCh38
rs1003635058 Chr.11: 44618905-44618905 on Build GRCh38
rs200610627 Chr.11: 44618760-44618760 on Build GRCh38
rs76669660 Chr.11: 44618741-44618741 on Build GRCh38
rs79095986 Chr.11: 44618696-44618696 on Build GRCh38
rs11038081 Chr.11: 44618927-44618927 on Build GRCh38
rs767858307 Chr.11: 44618736-44618736 on Build GRCh38
rs763141770 Chr.11: 44618729-44618729 on Build GRCh38
rs375815891 Chr.11: 44618589-44618589 on Build GRCh38
rs375606462 Chr.11: 44618746-44618746 on Build GRCh38
rs190517526 Chr.11: 44618702-44618702 on Build GRCh38
rs763656690 Chr.11: 44618903-44618903 on Build GRCh38
rs755430626 Chr.11: 44618697-44618697 on Build GRCh38
rs769912129 Chr.11: 44618768-44618768 on Build GRCh38
rs199858417 Chr.11: 44618717-44618717 on Build GRCh38
rs201974709 Chr.11: 44618609-44618609 on Build GRCh38
rs777719140 Chr.11: 44618658-44618658 on Build GRCh38
rs373528762 Chr.11: 44619004-44619004 on Build GRCh38
rs762027127 Chr.11: 44618684-44618684 on Build GRCh38
rs745591831 Chr.11: 44618755-44618755 on Build GRCh38
rs918994654 Chr.11: 44618908-44618908 on Build GRCh38
rs926144594 Chr.11: 44618806-44618806 on Build GRCh38
rs955282682 Chr.11: 44618996-44618996 on Build GRCh38
rs1039218154 Chr.11: 44618790-44618790 on Build GRCh38
rs367797052 Chr.11: 44618703-44618703 on Build GRCh38
rs761247903 Chr.11: 44619015-44619015 on Build GRCh38
rs946233956 Chr.11: 44618612-44618612 on Build GRCh38
rs113281135 Chr.11: 44618572-44618572 on Build GRCh38
rs201733300 Chr.11: 44618621-44618621 on Build GRCh38
rs375672218 Chr.11: 44618744-44618744 on Build GRCh38
rs957592072 Chr.11: 44618843-44618843 on Build GRCh38
rs772029006 Chr.11: 44618731-44618731 on Build GRCh38
rs746471269 Chr.11: 44618597-44618597 on Build GRCh38
rs750718911 Chr.11: 44618625-44618625 on Build GRCh38
rs553479075 Chr.11: 44618562-44618562 on Build GRCh38
rs760763762 Chr.11: 44618616-44618616 on Build GRCh38
rs773463121 Chr.11: 44618727-44618727 on Build GRCh38
rs879077881 Chr.11: 44619021-44619021 on Build GRCh38
rs1024522572 Chr.11: 44618559-44618559 on Build GRCh38
rs756853651 Chr.11: 44618752-44618752 on Build GRCh38
rs746917689 Chr.11: 44618762-44618762 on Build GRCh38
rs967853115 Chr.11: 44618948-44618948 on Build GRCh38
rs752282276 Chr.11: 44618738-44618738 on Build GRCh38
rs774382622 Chr.11: 44618663-44618663 on Build GRCh38
rs142423874 Chr.11: 44618693-44618693 on Build GRCh38
rs978794503 Chr.11: 44618577-44618577 on Build GRCh38
rs367958042 Chr.11: 44618608-44618608 on Build GRCh38
rs778556850 Chr.11: 44618712-44618712 on Build GRCh38
rs1139971 Chr.11: 44618718-44618718 on Build GRCh38
rs750810564 Chr.11: 44618692-44618692 on Build GRCh38
rs545079990 Chr.11: 44618794-44618794 on Build GRCh38
rs769763430 Chr.11: 44618606-44618606 on Build GRCh38
rs372966901 Chr.11: 44618588-44618588 on Build GRCh38
rs930925817 Chr.11: 44618801-44618801 on Build GRCh38
rs372946885 Chr.11: 44618664-44618664 on Build GRCh38
rs773268231 Chr.11: 44618770-44618770 on Build GRCh38
rs1055366914 Chr.11: 44618669-44618669 on Build GRCh38
rs776308413 Chr.11: 44618600-44618600 on Build GRCh38
rs539179773 Chr.11: 44618828-44618828 on Build GRCh38
rs1055417301 Chr.11: 44618545-44618545 on Build GRCh38
rs372446671 Chr.11: 44619014-44619014 on Build GRCh38
rs745383395 Chr.11: 44618702-44618703 on Build GRCh38
rs1045836192 Chr.11: 44618907-44618907 on Build GRCh38
rs967466894 Chr.11: 44618777-44618777 on Build GRCh38
rs369767006 Chr.11: 44618753-44618753 on Build GRCh38
rs564776657 Chr.11: 44618895-44618895 on Build GRCh38
rs754558079 Chr.11: 44618635-44618635 on Build GRCh38
rs920757031 Chr.11: 44618798-44618798 on Build GRCh38
rs1007443593 Chr.11: 44618944-44618944 on Build GRCh38
rs998186141 Chr.11: 44618995-44618995 on Build GRCh38
rs770993959 Chr.11: 44619005-44619005 on Build GRCh38
rs759367396 Chr.11: 44618674-44618674 on Build GRCh38
rs764046473 Chr.11: 44618620-44618620 on Build GRCh38
rs12421108 Chr.11: 44618550-44618550 on Build GRCh38
rs565388979 Chr.11: 44618785-44618785 on Build GRCh38
rs1030936072 Chr.11: 44618993-44618993 on Build GRCh38
rs146220817 Chr.11: 44618650-44618650 on Build GRCh38
rs767888414 Chr.11: 44619008-44619008 on Build GRCh38
rs150062713 Chr.11: 44618720-44618720 on Build GRCh38
rs780491115 Chr.11: 44618630-44618630 on Build GRCh38
rs1022163598 Chr.11: 44618933-44618933 on Build GRCh38
rs769089689 Chr.11: 44618731-44618731 on Build GRCh38
rs775258252 Chr.11: 44618735-44618735 on Build GRCh38

View Full Product Details