Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46310892-46311338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB3L1
Entrez Gene ID
90993
Gene Name
cAMP responsive element binding protein 3 like 1
Gene Aliases
OASIS
UniGene
Hs.405961
Gene Chromosome Location
Chr.11: 46277638-46321422 on Build GRCh38
Transcripts
NM_052854.3 XM_006718380.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1604564 c.753C>A p.H251Q Chr.11: 46311189-46311189 on Build GRCh38
5831749 c.621_622insC p.S210fs*6 Chr.11: 46311057-46311058 on Build GRCh38
4754304 c.631A>G p.S211G Chr.11: 46311067-46311067 on Build GRCh38
5372501 c.714C>T p.S238S Chr.11: 46311150-46311150 on Build GRCh38
4191774 c.648C>T p.S216S Chr.11: 46311084-46311084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375270539 Chr.11: 46311200-46311200 on Build GRCh38
rs774942627 Chr.11: 46311105-46311105 on Build GRCh38
rs942826091 Chr.11: 46311041-46311041 on Build GRCh38
rs528337004 Chr.11: 46311308-46311308 on Build GRCh38
rs376419401 Chr.11: 46311024-46311024 on Build GRCh38
rs746823561 Chr.11: 46311115-46311115 on Build GRCh38
rs946852280 Chr.11: 46310976-46310976 on Build GRCh38
rs745598774 Chr.11: 46311171-46311171 on Build GRCh38
rs756479447 Chr.11: 46311085-46311085 on Build GRCh38
rs775343544 Chr.11: 46311053-46311053 on Build GRCh38
rs536974189 Chr.11: 46310958-46310958 on Build GRCh38
rs751752437 Chr.11: 46311000-46311000 on Build GRCh38
rs192179218 Chr.11: 46311152-46311152 on Build GRCh38
rs867474347 Chr.11: 46311074-46311074 on Build GRCh38
rs748649332 Chr.11: 46311011-46311011 on Build GRCh38
rs375485514 Chr.11: 46311057-46311057 on Build GRCh38
rs757791649 Chr.11: 46311166-46311166 on Build GRCh38
rs184532655 Chr.11: 46311281-46311281 on Build GRCh38
rs148174469 Chr.11: 46311226-46311226 on Build GRCh38
rs770508076 Chr.11: 46311036-46311036 on Build GRCh38
rs886226307 Chr.11: 46311282-46311282 on Build GRCh38
rs573541101 Chr.11: 46311018-46311018 on Build GRCh38
rs777225566 Chr.11: 46311084-46311084 on Build GRCh38
rs990709388 Chr.11: 46310951-46310951 on Build GRCh38
rs2303434 Chr.11: 46310945-46310945 on Build GRCh38
rs372951534 Chr.11: 46311087-46311087 on Build GRCh38
rs756569240 Chr.11: 46311145-46311145 on Build GRCh38
rs763039074 Chr.11: 46311069-46311069 on Build GRCh38
rs149913359 Chr.11: 46311260-46311261 on Build GRCh38
rs754446917 Chr.11: 46311148-46311148 on Build GRCh38
rs755326376 Chr.11: 46311009-46311009 on Build GRCh38
rs767925766 Chr.11: 46310926-46310926 on Build GRCh38
rs2303435 Chr.11: 46310953-46310953 on Build GRCh38
rs761704234 Chr.11: 46311117-46311117 on Build GRCh38
rs548853523 Chr.11: 46310921-46310921 on Build GRCh38
rs774472916 Chr.11: 46311126-46311126 on Build GRCh38
rs559287577 Chr.11: 46311245-46311245 on Build GRCh38
rs778465642 Chr.11: 46311090-46311090 on Build GRCh38
rs751767246 Chr.11: 46311072-46311072 on Build GRCh38
rs773907336 Chr.11: 46311037-46311037 on Build GRCh38
rs376081099 Chr.11: 46311088-46311088 on Build GRCh38
rs1041204424 Chr.11: 46311298-46311298 on Build GRCh38
rs760674300 Chr.11: 46311054-46311054 on Build GRCh38
rs771754544 Chr.11: 46311104-46311104 on Build GRCh38
rs771813222 Chr.11: 46311048-46311048 on Build GRCh38
rs960519202 Chr.11: 46310940-46310940 on Build GRCh38
rs779911363 Chr.11: 46311216-46311216 on Build GRCh38
rs755202953 Chr.11: 46311077-46311077 on Build GRCh38
rs117481046 Chr.11: 46311189-46311189 on Build GRCh38
rs547183036 Chr.11: 46311277-46311277 on Build GRCh38
rs780940667 Chr.11: 46311187-46311187 on Build GRCh38
rs781585862 Chr.11: 46311010-46311010 on Build GRCh38
rs779707746 Chr.11: 46311170-46311170 on Build GRCh38
rs745390642 Chr.11: 46311155-46311155 on Build GRCh38
rs891602843 Chr.11: 46311306-46311306 on Build GRCh38
rs891692942 Chr.11: 46311137-46311137 on Build GRCh38
rs1034780350 Chr.11: 46310927-46310927 on Build GRCh38
rs545047399 Chr.11: 46311141-46311141 on Build GRCh38
rs772954581 Chr.11: 46311206-46311206 on Build GRCh38
rs776390325 Chr.11: 46311057-46311058 on Build GRCh38
rs185752252 Chr.11: 46310923-46310923 on Build GRCh38
rs1027334112 Chr.11: 46310911-46310911 on Build GRCh38
rs374891862 Chr.11: 46310999-46310999 on Build GRCh38
rs367799632 Chr.11: 46311143-46311143 on Build GRCh38
rs368536782 Chr.11: 46311082-46311082 on Build GRCh38
rs371335074 Chr.11: 46311146-46311146 on Build GRCh38
rs753092259 Chr.11: 46310995-46310995 on Build GRCh38
rs372615525 Chr.11: 46311229-46311229 on Build GRCh38
rs745502768 Chr.11: 46311038-46311038 on Build GRCh38
rs565308246 Chr.11: 46311142-46311142 on Build GRCh38
rs371931737 Chr.11: 46311172-46311172 on Build GRCh38
rs551370451 Chr.11: 46310947-46310947 on Build GRCh38
rs930352444 Chr.11: 46311246-46311246 on Build GRCh38
rs776498586 Chr.11: 46311064-46311064 on Build GRCh38
rs759673733 Chr.11: 46311209-46311209 on Build GRCh38
rs766415281 Chr.11: 46311119-46311119 on Build GRCh38
rs986821504 Chr.11: 46311217-46311217 on Build GRCh38
rs776576938 Chr.11: 46311116-46311116 on Build GRCh38
rs771995536 Chr.11: 46311222-46311222 on Build GRCh38
rs956684921 Chr.11: 46311204-46311204 on Build GRCh38
rs912803932 Chr.11: 46311221-46311221 on Build GRCh38
rs766438770 Chr.11: 46311071-46311071 on Build GRCh38
rs187725533 Chr.11: 46311035-46311035 on Build GRCh38
rs369168458 Chr.11: 46311153-46311153 on Build GRCh38
rs536747540 Chr.11: 46311022-46311022 on Build GRCh38
rs750500533 Chr.11: 46310988-46310988 on Build GRCh38
rs745567388 Chr.11: 46311103-46311103 on Build GRCh38
rs923957110 Chr.11: 46311002-46311002 on Build GRCh38
rs919572251 Chr.11: 46311039-46311039 on Build GRCh38

View Full Product Details