Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46645261-46645758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG13
Entrez Gene ID
9776
Gene Name
autophagy related 13
Gene Aliases
KIAA0652, PARATARG8
UniGene
Hs.127403
Gene Chromosome Location
Chr.11: 46617276-46676019 on Build GRCh38
Transcripts
NM_001142673.2 NM_001205119.1 NM_001205120.1 NM_001205121.1 NM_001205122.1 NM_014741.4 XM_005253262.2 XM_005253263.2 XM_005253265.2 XM_005253266.2 XM_005253268.2 XM_006718394.2 XM_006718395.2 XM_006718396.2 XM_011520493.1 XM_011520496.2 XM_011520499.2 XM_011520500.1 XM_011520501.1 XM_011520502.1 XM_011520503.1 XM_017018599.1 XM_017018600.1 XM_017018601.1 XM_017018602.1 XM_017018603.1 XM_017018604.1 XM_017018605.1 XM_017018606.1 XM_017018607.1 XM_017018608.1 XM_017018609.1 XM_017018610.1 XM_017018611.1 XM_017018612.1 XM_017018613.1 XM_017018614.1 XM_017018615.1 XM_017018616.1 XM_017018617.1 XM_017018618.1 XM_017018619.1 XM_017018620.1 XM_017018621.1 XM_017018622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1980716 c.131C>T p.S44F Chr.11: 46645400-46645400 on Build GRCh38
1746256 c.138G>T p.T46T Chr.11: 46645407-46645407 on Build GRCh38
4414847 c.88C>T p.Q30* Chr.11: 46645357-46645357 on Build GRCh38
1980717 c.131C>T p.S44F Chr.11: 46645400-46645400 on Build GRCh38
466830 c.134C>A p.P45Q Chr.11: 46645403-46645403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200722802 Chr.11: 46645441-46645441 on Build GRCh38
rs148226802 Chr.11: 46645404-46645404 on Build GRCh38
rs779760415 Chr.11: 46645383-46645383 on Build GRCh38
rs751319265 Chr.11: 46645444-46645444 on Build GRCh38
rs867822449 Chr.11: 46645533-46645533 on Build GRCh38
rs369218292 Chr.11: 46645416-46645416 on Build GRCh38
rs111544937 Chr.11: 46645565-46645565 on Build GRCh38
rs905030149 Chr.11: 46645323-46645323 on Build GRCh38
rs771571345 Chr.11: 46645300-46645300 on Build GRCh38
rs919214960 Chr.11: 46645302-46645302 on Build GRCh38
rs559285412 Chr.11: 46645666-46645666 on Build GRCh38
rs746417694 Chr.11: 46645400-46645400 on Build GRCh38
rs142536582 Chr.11: 46645388-46645388 on Build GRCh38
rs764764041 Chr.11: 46645431-46645431 on Build GRCh38
rs996680793 Chr.11: 46645647-46645647 on Build GRCh38
rs557821529 Chr.11: 46645443-46645443 on Build GRCh38
rs774802327 Chr.11: 46645309-46645309 on Build GRCh38
rs754181248 Chr.11: 46645468-46645468 on Build GRCh38
rs759967771 Chr.11: 46645319-46645319 on Build GRCh38
rs112388953 Chr.11: 46645541-46645541 on Build GRCh38
rs1044043456 Chr.11: 46645481-46645481 on Build GRCh38
rs765145772 Chr.11: 46645353-46645353 on Build GRCh38
rs1053605488 Chr.11: 46645534-46645534 on Build GRCh38
rs1029006292 Chr.11: 46645684-46645684 on Build GRCh38
rs563245576 Chr.11: 46645555-46645555 on Build GRCh38
rs998117355 Chr.11: 46645682-46645682 on Build GRCh38
rs542902649 Chr.11: 46645637-46645637 on Build GRCh38
rs536521034 Chr.11: 46645428-46645428 on Build GRCh38
rs147563838 Chr.11: 46645392-46645392 on Build GRCh38
rs775740650 Chr.11: 46645407-46645407 on Build GRCh38
rs926965735 Chr.11: 46645490-46645490 on Build GRCh38
rs753290726 Chr.11: 46645333-46645333 on Build GRCh38
rs766570338 Chr.11: 46645323-46645323 on Build GRCh38
rs373254182 Chr.11: 46645430-46645430 on Build GRCh38
rs753962856 Chr.11: 46645429-46645430 on Build GRCh38
rs760295528 Chr.11: 46645331-46645331 on Build GRCh38
rs547443616 Chr.11: 46645474-46645474 on Build GRCh38
rs750423536 Chr.11: 46645363-46645363 on Build GRCh38
rs750798917 Chr.11: 46645312-46645316 on Build GRCh38
rs1002021775 Chr.11: 46645355-46645355 on Build GRCh38
rs758215266 Chr.11: 46645451-46645451 on Build GRCh38
rs752912929 Chr.11: 46645387-46645387 on Build GRCh38
rs377219841 Chr.11: 46645381-46645381 on Build GRCh38
rs772632542 Chr.11: 46645322-46645322 on Build GRCh38
rs1037546811 Chr.11: 46645582-46645582 on Build GRCh38
rs899208758 Chr.11: 46645589-46645589 on Build GRCh38
rs577745912 Chr.11: 46645452-46645452 on Build GRCh38
rs768203975 Chr.11: 46645427-46645427 on Build GRCh38
rs571599311 Chr.11: 46645288-46645288 on Build GRCh38
rs777495055 Chr.11: 46645452-46645452 on Build GRCh38
rs768736724 Chr.11: 46645641-46645641 on Build GRCh38
rs1042416857 Chr.11: 46645657-46645657 on Build GRCh38
rs949276700 Chr.11: 46645321-46645321 on Build GRCh38
rs79005969 Chr.11: 46645518-46645518 on Build GRCh38
rs775179435 Chr.11: 46645325-46645325 on Build GRCh38
rs760948232 Chr.11: 46645322-46645323 on Build GRCh38
rs761043129 Chr.11: 46645432-46645433 on Build GRCh38
rs143855756 Chr.11: 46645398-46645398 on Build GRCh38
rs763618977 Chr.11: 46645332-46645332 on Build GRCh38
rs573348494 Chr.11: 46645440-46645440 on Build GRCh38
rs372958950 Chr.11: 46645485-46645485 on Build GRCh38
rs926534617 Chr.11: 46645525-46645525 on Build GRCh38
rs1054296018 Chr.11: 46645380-46645380 on Build GRCh38
rs529203319 Chr.11: 46645433-46645438 on Build GRCh38
rs938390121 Chr.11: 46645504-46645504 on Build GRCh38
rs901184047 Chr.11: 46645669-46645669 on Build GRCh38
rs948995156 Chr.11: 46645498-46645498 on Build GRCh38
rs918935377 Chr.11: 46645497-46645497 on Build GRCh38
rs756304138 Chr.11: 46645445-46645445 on Build GRCh38

View Full Product Details