Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46648995-46649528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG13
Entrez Gene ID
9776
Gene Name
autophagy related 13
Gene Aliases
KIAA0652, PARATARG8
UniGene
Hs.127403
Gene Chromosome Location
Chr.11: 46617276-46676019 on Build GRCh38
Transcripts
NM_001142673.2 NM_001205119.1 NM_001205120.1 NM_001205121.1 NM_001205122.1 NM_014741.4 XM_005253262.2 XM_005253263.2 XM_005253265.2 XM_005253266.2 XM_005253268.2 XM_006718394.2 XM_006718395.2 XM_006718396.2 XM_011520493.1 XM_011520496.2 XM_011520499.2 XM_011520500.1 XM_011520501.1 XM_011520502.1 XM_011520503.1 XM_017018599.1 XM_017018600.1 XM_017018601.1 XM_017018602.1 XM_017018603.1 XM_017018604.1 XM_017018605.1 XM_017018606.1 XM_017018607.1 XM_017018608.1 XM_017018609.1 XM_017018610.1 XM_017018611.1 XM_017018612.1 XM_017018613.1 XM_017018614.1 XM_017018615.1 XM_017018616.1 XM_017018617.1 XM_017018618.1 XM_017018619.1 XM_017018620.1 XM_017018621.1 XM_017018622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1316751 c.285G>T p.E95D Chr.11: 46649151-46649151 on Build GRCh38
5818654 c.316A>T p.K106* Chr.11: 46649182-46649182 on Build GRCh38
5935678 c.278C>T p.S93F Chr.11: 46649144-46649144 on Build GRCh38
1316753 c.290A>T p.E97V Chr.11: 46649156-46649156 on Build GRCh38
1316752 c.285G>T p.E95D Chr.11: 46649151-46649151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542516191 Chr.11: 46649028-46649031 on Build GRCh38
rs370531802 Chr.11: 46649210-46649210 on Build GRCh38
rs955834619 Chr.11: 46649420-46649420 on Build GRCh38
rs542593975 Chr.11: 46649485-46649485 on Build GRCh38
rs775458708 Chr.11: 46649134-46649134 on Build GRCh38
rs944748133 Chr.11: 46649473-46649473 on Build GRCh38
rs551129544 Chr.11: 46649064-46649064 on Build GRCh38
rs756517897 Chr.11: 46649240-46649240 on Build GRCh38
rs1000468669 Chr.11: 46649482-46649482 on Build GRCh38
rs944681569 Chr.11: 46649104-46649104 on Build GRCh38
rs113203272 Chr.11: 46649306-46649306 on Build GRCh38
rs746274042 Chr.11: 46649440-46649440 on Build GRCh38
rs199850974 Chr.11: 46649154-46649154 on Build GRCh38
rs568471338 Chr.11: 46649191-46649191 on Build GRCh38
rs774128345 Chr.11: 46649098-46649098 on Build GRCh38
rs1023411336 Chr.11: 46649165-46649165 on Build GRCh38
rs757064243 Chr.11: 46649259-46649259 on Build GRCh38
rs569779894 Chr.11: 46649078-46649078 on Build GRCh38
rs925797742 Chr.11: 46649395-46649395 on Build GRCh38
rs1049301722 Chr.11: 46649490-46649490 on Build GRCh38
rs375774792 Chr.11: 46649042-46649042 on Build GRCh38
rs1039788836 Chr.11: 46649384-46649384 on Build GRCh38
rs748737180 Chr.11: 46649089-46649089 on Build GRCh38
rs1039301104 Chr.11: 46649126-46649126 on Build GRCh38
rs911935898 Chr.11: 46649453-46649453 on Build GRCh38
rs770546459 Chr.11: 46649093-46649093 on Build GRCh38
rs556607757 Chr.11: 46649479-46649479 on Build GRCh38
rs1003965864 Chr.11: 46649294-46649294 on Build GRCh38
rs750723551 Chr.11: 46649159-46649159 on Build GRCh38
rs771945457 Chr.11: 46649125-46649125 on Build GRCh38
rs762067700 Chr.11: 46649151-46649151 on Build GRCh38
rs1041703824 Chr.11: 46649452-46649452 on Build GRCh38
rs1014321987 Chr.11: 46649352-46649352 on Build GRCh38
rs759567335 Chr.11: 46649099-46649099 on Build GRCh38
rs1022907094 Chr.11: 46649355-46649355 on Build GRCh38
rs942555852 Chr.11: 46649337-46649337 on Build GRCh38
rs756531838 Chr.11: 46649206-46649206 on Build GRCh38
rs377691475 Chr.11: 46649192-46649192 on Build GRCh38
rs978549203 Chr.11: 46649061-46649061 on Build GRCh38
rs951332315 Chr.11: 46649274-46649274 on Build GRCh38
rs922724592 Chr.11: 46649385-46649385 on Build GRCh38
rs886572092 Chr.11: 46649278-46649278 on Build GRCh38
rs770755752 Chr.11: 46649109-46649109 on Build GRCh38
rs746785843 Chr.11: 46649089-46649089 on Build GRCh38
rs796305978 Chr.11: 46649033-46649033 on Build GRCh38
rs145405423 Chr.11: 46649484-46649484 on Build GRCh38
rs140341222 Chr.11: 46649461-46649461 on Build GRCh38
rs753077047 Chr.11: 46649195-46649195 on Build GRCh38
rs1000732449 Chr.11: 46649032-46649033 on Build GRCh38
rs766526438 Chr.11: 46649190-46649190 on Build GRCh38
rs555778315 Chr.11: 46649501-46649501 on Build GRCh38
rs762909334 Chr.11: 46649175-46649175 on Build GRCh38
rs988874573 Chr.11: 46649427-46649427 on Build GRCh38
rs549588938 Chr.11: 46649476-46649476 on Build GRCh38
rs143736943 Chr.11: 46649307-46649307 on Build GRCh38
rs548075058 Chr.11: 46649245-46649245 on Build GRCh38
rs568285297 Chr.11: 46649297-46649297 on Build GRCh38
rs774692224 Chr.11: 46649366-46649366 on Build GRCh38
rs749489324 Chr.11: 46649255-46649255 on Build GRCh38
rs146814112 Chr.11: 46649373-46649373 on Build GRCh38
rs202223911 Chr.11: 46649194-46649194 on Build GRCh38

View Full Product Details