Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46650038-46650569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG13
Entrez Gene ID
9776
Gene Name
autophagy related 13
Gene Aliases
KIAA0652, PARATARG8
UniGene
Hs.127403
Gene Chromosome Location
Chr.11: 46617276-46676019 on Build GRCh38
Transcripts
NM_001142673.2 NM_001205119.1 NM_001205120.1 NM_001205121.1 NM_001205122.1 NM_014741.4 XM_005253262.2 XM_005253263.2 XM_005253265.2 XM_005253266.2 XM_005253268.2 XM_006718394.2 XM_006718395.2 XM_006718396.2 XM_011520493.1 XM_011520496.2 XM_011520499.2 XM_011520500.1 XM_011520501.1 XM_011520502.1 XM_011520503.1 XM_017018599.1 XM_017018600.1 XM_017018601.1 XM_017018602.1 XM_017018603.1 XM_017018604.1 XM_017018605.1 XM_017018606.1 XM_017018607.1 XM_017018608.1 XM_017018609.1 XM_017018610.1 XM_017018611.1 XM_017018612.1 XM_017018613.1 XM_017018614.1 XM_017018615.1 XM_017018616.1 XM_017018617.1 XM_017018618.1 XM_017018619.1 XM_017018620.1 XM_017018621.1 XM_017018622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3670768 c.398G>T p.R133M Chr.11: 46650257-46650257 on Build GRCh38
5037090 c.400G>T p.V134L Chr.11: 46650259-46650259 on Build GRCh38
1746258 c.382C>G p.L128V Chr.11: 46650241-46650241 on Build GRCh38
4994983 c.397A>G p.R133G Chr.11: 46650256-46650256 on Build GRCh38
3967526 c.348G>A p.T116T Chr.11: 46650207-46650207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778253155 Chr.11: 46650256-46650256 on Build GRCh38
rs763254277 Chr.11: 46650198-46650198 on Build GRCh38
rs145617209 Chr.11: 46650250-46650250 on Build GRCh38
rs769133736 Chr.11: 46650325-46650326 on Build GRCh38
rs138409929 Chr.11: 46650333-46650333 on Build GRCh38
rs149007632 Chr.11: 46650249-46650249 on Build GRCh38
rs940934437 Chr.11: 46650270-46650270 on Build GRCh38
rs756442263 Chr.11: 46650255-46650255 on Build GRCh38
rs769357200 Chr.11: 46650462-46650462 on Build GRCh38
rs776270948 Chr.11: 46650512-46650512 on Build GRCh38
rs768001692 Chr.11: 46650217-46650217 on Build GRCh38
rs935693400 Chr.11: 46650437-46650437 on Build GRCh38
rs556693640 Chr.11: 46650071-46650071 on Build GRCh38
rs955208083 Chr.11: 46650394-46650394 on Build GRCh38
rs768518895 Chr.11: 46650346-46650346 on Build GRCh38
rs192375992 Chr.11: 46650136-46650136 on Build GRCh38
rs981672375 Chr.11: 46650521-46650521 on Build GRCh38
rs772141554 Chr.11: 46650155-46650155 on Build GRCh38
rs1044552479 Chr.11: 46650386-46650386 on Build GRCh38
rs927032653 Chr.11: 46650409-46650409 on Build GRCh38
rs766325449 Chr.11: 46650203-46650203 on Build GRCh38
rs541476833 Chr.11: 46650469-46650469 on Build GRCh38
rs778943166 Chr.11: 46650123-46650123 on Build GRCh38
rs191856916 Chr.11: 46650524-46650524 on Build GRCh38
rs948966026 Chr.11: 46650067-46650067 on Build GRCh38
rs952052519 Chr.11: 46650188-46650188 on Build GRCh38
rs370497611 Chr.11: 46650147-46650147 on Build GRCh38
rs200378608 Chr.11: 46650303-46650303 on Build GRCh38
rs182890678 Chr.11: 46650143-46650143 on Build GRCh38
rs771773631 Chr.11: 46650309-46650309 on Build GRCh38
rs756967277 Chr.11: 46650264-46650264 on Build GRCh38
rs138230726 Chr.11: 46650300-46650300 on Build GRCh38
rs759975268 Chr.11: 46650207-46650207 on Build GRCh38
rs1031719058 Chr.11: 46650424-46650424 on Build GRCh38
rs561695923 Chr.11: 46650513-46650513 on Build GRCh38
rs780955957 Chr.11: 46650165-46650166 on Build GRCh38
rs760540421 Chr.11: 46650168-46650168 on Build GRCh38
rs1029103396 Chr.11: 46650532-46650532 on Build GRCh38
rs559425965 Chr.11: 46650445-46650445 on Build GRCh38
rs187243114 Chr.11: 46650162-46650162 on Build GRCh38
rs564329036 Chr.11: 46650514-46650514 on Build GRCh38
rs879149211 Chr.11: 46650443-46650443 on Build GRCh38
rs769502312 Chr.11: 46650511-46650511 on Build GRCh38
rs562223535 Chr.11: 46650448-46650448 on Build GRCh38
rs746992110 Chr.11: 46650338-46650338 on Build GRCh38
rs753382891 Chr.11: 46650262-46650262 on Build GRCh38
rs1019201666 Chr.11: 46650128-46650128 on Build GRCh38
rs200781784 Chr.11: 46650135-46650135 on Build GRCh38
rs372339614 Chr.11: 46650362-46650362 on Build GRCh38
rs996707744 Chr.11: 46650349-46650351 on Build GRCh38
rs773284017 Chr.11: 46650180-46650180 on Build GRCh38
rs777970032 Chr.11: 46650124-46650124 on Build GRCh38
rs145889337 Chr.11: 46650206-46650206 on Build GRCh38
rs768611041 Chr.11: 46650169-46650169 on Build GRCh38
rs531241557 Chr.11: 46650456-46650456 on Build GRCh38
rs777401932 Chr.11: 46650130-46650130 on Build GRCh38
rs990141646 Chr.11: 46650473-46650473 on Build GRCh38
rs745399183 Chr.11: 46650184-46650187 on Build GRCh38
rs918084208 Chr.11: 46650358-46650358 on Build GRCh38
rs1045902725 Chr.11: 46650106-46650106 on Build GRCh38
rs1010571389 Chr.11: 46650511-46650511 on Build GRCh38

View Full Product Details