Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46719623-46720118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F2
Entrez Gene ID
2147
Gene Name
coagulation factor II, thrombin
Gene Aliases
PT, RPRGL2, THPH1
UniGene
Hs.655207
Gene Chromosome Location
Chr.11: 46719166-46739508 on Build GRCh38
Transcripts
NM_000506.4 NM_001311257.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4449466 c.240+1G>A p.? Chr.11: 46719863-46719863 on Build GRCh38
4746106 c.187G>A p.E63K Chr.11: 46719809-46719809 on Build GRCh38
6578159 c.114C>G p.L38L Chr.11: 46719736-46719736 on Build GRCh38
6657714 c.181G>A p.V61M Chr.11: 46719803-46719803 on Build GRCh38
7366884 c.94C>G p.Q32E Chr.11: 46719716-46719716 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780167560 Chr.11: 46719884-46719884 on Build GRCh38
rs781026103 Chr.11: 46719777-46719777 on Build GRCh38
rs1031279305 Chr.11: 46720019-46720019 on Build GRCh38
rs905074363 Chr.11: 46719796-46719796 on Build GRCh38
rs754825716 Chr.11: 46719670-46719670 on Build GRCh38
rs149847109 Chr.11: 46720066-46720066 on Build GRCh38
rs371479474 Chr.11: 46719671-46719671 on Build GRCh38
rs778412009 Chr.11: 46720087-46720087 on Build GRCh38
rs749497826 Chr.11: 46719681-46719681 on Build GRCh38
rs1043774209 Chr.11: 46719948-46719948 on Build GRCh38
rs954611364 Chr.11: 46719842-46719842 on Build GRCh38
rs371680056 Chr.11: 46719835-46719835 on Build GRCh38
rs772252088 Chr.11: 46719803-46719803 on Build GRCh38
rs756628713 Chr.11: 46719879-46719879 on Build GRCh38
rs376845267 Chr.11: 46719688-46719688 on Build GRCh38
rs755991682 Chr.11: 46719872-46719872 on Build GRCh38
rs766255526 Chr.11: 46719749-46719749 on Build GRCh38
rs556066325 Chr.11: 46719903-46719903 on Build GRCh38
rs754914460 Chr.11: 46719767-46719767 on Build GRCh38
rs894273773 Chr.11: 46719938-46719938 on Build GRCh38
rs200934494 Chr.11: 46719855-46719855 on Build GRCh38
rs1004064124 Chr.11: 46719844-46719844 on Build GRCh38
rs751494277 Chr.11: 46719759-46719759 on Build GRCh38
rs745681370 Chr.11: 46719683-46719683 on Build GRCh38
rs762660409 Chr.11: 46719863-46719864 on Build GRCh38
rs1056357375 Chr.11: 46720014-46720014 on Build GRCh38
rs866560182 Chr.11: 46719999-46719999 on Build GRCh38
rs781359748 Chr.11: 46720024-46720024 on Build GRCh38
rs1055231311 Chr.11: 46719934-46719934 on Build GRCh38
rs752493930 Chr.11: 46719871-46719871 on Build GRCh38
rs759788134 Chr.11: 46719692-46719692 on Build GRCh38
rs374353447 Chr.11: 46719802-46719802 on Build GRCh38
rs758759228 Chr.11: 46719883-46719883 on Build GRCh38
rs1042917828 Chr.11: 46719959-46719959 on Build GRCh38
rs769517071 Chr.11: 46719718-46719729 on Build GRCh38
rs1021513492 Chr.11: 46719775-46719775 on Build GRCh38
rs904439584 Chr.11: 46719960-46719960 on Build GRCh38
rs868768072 Chr.11: 46720051-46720051 on Build GRCh38
rs998768059 Chr.11: 46719964-46719964 on Build GRCh38
rs747075170 Chr.11: 46719889-46719889 on Build GRCh38
rs1015664856 Chr.11: 46719847-46719847 on Build GRCh38
rs759457145 Chr.11: 46719868-46719868 on Build GRCh38
rs753249964 Chr.11: 46719971-46719971 on Build GRCh38
rs894338780 Chr.11: 46719732-46719732 on Build GRCh38
rs896424682 Chr.11: 46720018-46720018 on Build GRCh38
rs773610151 Chr.11: 46719904-46719904 on Build GRCh38
rs1009034345 Chr.11: 46720038-46720038 on Build GRCh38
rs570442167 Chr.11: 46719684-46719684 on Build GRCh38
rs868768555 Chr.11: 46719840-46719840 on Build GRCh38
rs776390013 Chr.11: 46719682-46719683 on Build GRCh38
rs764029379 Chr.11: 46719880-46719880 on Build GRCh38
rs756816205 Chr.11: 46720068-46720068 on Build GRCh38
rs757486222 Chr.11: 46719789-46719789 on Build GRCh38
rs946360744 Chr.11: 46719757-46719757 on Build GRCh38
rs965209264 Chr.11: 46720046-46720046 on Build GRCh38
rs751272302 Chr.11: 46719666-46719666 on Build GRCh38
rs1002157797 Chr.11: 46719797-46719797 on Build GRCh38
rs772878615 Chr.11: 46719850-46719850 on Build GRCh38
rs775127693 Chr.11: 46719862-46719863 on Build GRCh38
rs368162633 Chr.11: 46719795-46719795 on Build GRCh38
rs148596790 Chr.11: 46719730-46719730 on Build GRCh38
rs769031043 Chr.11: 46719689-46719689 on Build GRCh38
rs776998025 Chr.11: 46719823-46719823 on Build GRCh38
rs772667220 Chr.11: 46719685-46719685 on Build GRCh38
rs142001812 Chr.11: 46719813-46719813 on Build GRCh38
rs748467283 Chr.11: 46719830-46719830 on Build GRCh38
rs1047809423 Chr.11: 46719653-46719653 on Build GRCh38
rs377331494 Chr.11: 46719890-46719890 on Build GRCh38
rs890155920 Chr.11: 46720026-46720026 on Build GRCh38
rs1013222460 Chr.11: 46719740-46719740 on Build GRCh38
rs767357179 Chr.11: 46719870-46719870 on Build GRCh38
rs141112085 Chr.11: 46719968-46719968 on Build GRCh38
rs939492961 Chr.11: 46719693-46719693 on Build GRCh38
rs944441422 Chr.11: 46719690-46719690 on Build GRCh38
rs547147827 Chr.11: 46719682-46719682 on Build GRCh38
rs769103147 Chr.11: 46719815-46719815 on Build GRCh38
rs773803065 Chr.11: 46719866-46719866 on Build GRCh38
rs147253030 Chr.11: 46719814-46719814 on Build GRCh38
rs765847453 Chr.11: 46719861-46719861 on Build GRCh38
rs144785536 Chr.11: 46719725-46719725 on Build GRCh38
rs752446552 Chr.11: 46719788-46719788 on Build GRCh38
rs764885504 Chr.11: 46719729-46719729 on Build GRCh38
rs937905659 Chr.11: 46719981-46719981 on Build GRCh38
rs762485669 Chr.11: 46719741-46719741 on Build GRCh38
rs1017358308 Chr.11: 46720043-46720043 on Build GRCh38
rs151121282 Chr.11: 46719856-46719856 on Build GRCh38
rs927977502 Chr.11: 46719917-46719917 on Build GRCh38
rs761190231 Chr.11: 46719864-46719865 on Build GRCh38
rs2070850 Chr.11: 46719945-46719945 on Build GRCh38
rs939420608 Chr.11: 46719920-46719920 on Build GRCh38

View Full Product Details