Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46720433-46720929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F2
Entrez Gene ID
2147
Gene Name
coagulation factor II, thrombin
Gene Aliases
PT, RPRGL2, THPH1
UniGene
Hs.655207
Gene Chromosome Location
Chr.11: 46719166-46739508 on Build GRCh38
Transcripts
NM_000506.4 NM_001311257.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7162317 c.279G>A p.A93A Chr.11: 46720803-46720803 on Build GRCh38
927278 c.253G>A p.A85T Chr.11: 46720535-46720535 on Build GRCh38
4145738 c.257A>T p.K86M Chr.11: 46720539-46720539 on Build GRCh38
7153097 c.289C>G p.R97G Chr.11: 46720813-46720813 on Build GRCh38
5453905 c.290G>A p.R97Q Chr.11: 46720814-46720814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376262187 Chr.11: 46720488-46720488 on Build GRCh38
rs578029112 Chr.11: 46720658-46720658 on Build GRCh38
rs532990120 Chr.11: 46720484-46720484 on Build GRCh38
rs569227147 Chr.11: 46720901-46720901 on Build GRCh38
rs968941106 Chr.11: 46720554-46720554 on Build GRCh38
rs980697463 Chr.11: 46720637-46720637 on Build GRCh38
rs911806952 Chr.11: 46720650-46720650 on Build GRCh38
rs758638621 Chr.11: 46720506-46720506 on Build GRCh38
rs756544259 Chr.11: 46720802-46720802 on Build GRCh38
rs147062971 Chr.11: 46720497-46720497 on Build GRCh38
rs968086215 Chr.11: 46720504-46720504 on Build GRCh38
rs759846816 Chr.11: 46720767-46720767 on Build GRCh38
rs771813897 Chr.11: 46720845-46720845 on Build GRCh38
rs754338884 Chr.11: 46720587-46720587 on Build GRCh38
rs770501121 Chr.11: 46720473-46720473 on Build GRCh38
rs144736001 Chr.11: 46720541-46720541 on Build GRCh38
rs751709961 Chr.11: 46720766-46720766 on Build GRCh38
rs764368187 Chr.11: 46720672-46720672 on Build GRCh38
rs867940810 Chr.11: 46720655-46720655 on Build GRCh38
rs201347110 Chr.11: 46720543-46720543 on Build GRCh38
rs773842485 Chr.11: 46720483-46720483 on Build GRCh38
rs779651171 Chr.11: 46720853-46720853 on Build GRCh38
rs373684363 Chr.11: 46720595-46720595 on Build GRCh38
rs778448750 Chr.11: 46720570-46720570 on Build GRCh38
rs939901054 Chr.11: 46720724-46720724 on Build GRCh38
rs746524034 Chr.11: 46720855-46720855 on Build GRCh38
rs201962988 Chr.11: 46720499-46720499 on Build GRCh38
rs751619623 Chr.11: 46720526-46720526 on Build GRCh38
rs189436668 Chr.11: 46720808-46720808 on Build GRCh38
rs756683451 Chr.11: 46720544-46720544 on Build GRCh38
rs1035279625 Chr.11: 46720465-46720465 on Build GRCh38
rs368019807 Chr.11: 46720748-46720748 on Build GRCh38
rs745502055 Chr.11: 46720839-46720839 on Build GRCh38
rs3136435 Chr.11: 46720876-46720876 on Build GRCh38
rs765766459 Chr.11: 46720752-46720752 on Build GRCh38
rs200034626 Chr.11: 46720593-46720593 on Build GRCh38
rs369279444 Chr.11: 46720492-46720492 on Build GRCh38
rs917629143 Chr.11: 46720706-46720706 on Build GRCh38
rs1005059949 Chr.11: 46720846-46720846 on Build GRCh38
rs771082282 Chr.11: 46720623-46720623 on Build GRCh38
rs959393525 Chr.11: 46720669-46720669 on Build GRCh38
rs761732579 Chr.11: 46720760-46720760 on Build GRCh38
rs769448614 Chr.11: 46720875-46720875 on Build GRCh38
rs370547285 Chr.11: 46720557-46720557 on Build GRCh38
rs775927207 Chr.11: 46720856-46720856 on Build GRCh38
rs749303506 Chr.11: 46720594-46720594 on Build GRCh38
rs375306913 Chr.11: 46720735-46720735 on Build GRCh38
rs745578381 Chr.11: 46720571-46720571 on Build GRCh38
rs778280660 Chr.11: 46720803-46720803 on Build GRCh38
rs748937156 Chr.11: 46720472-46720472 on Build GRCh38
rs558280278 Chr.11: 46720601-46720601 on Build GRCh38
rs934512313 Chr.11: 46720632-46720632 on Build GRCh38
rs569264167 Chr.11: 46720528-46720528 on Build GRCh38
rs766575627 Chr.11: 46720763-46720763 on Build GRCh38
rs755398788 Chr.11: 46720599-46720600 on Build GRCh38
rs3136434 Chr.11: 46720699-46720699 on Build GRCh38
rs889843584 Chr.11: 46720903-46720903 on Build GRCh38
rs770573470 Chr.11: 46720562-46720562 on Build GRCh38
rs200904690 Chr.11: 46720814-46720814 on Build GRCh38
rs1021728698 Chr.11: 46720520-46720520 on Build GRCh38
rs147892497 Chr.11: 46720809-46720809 on Build GRCh38
rs368155840 Chr.11: 46720865-46720865 on Build GRCh38
rs200777670 Chr.11: 46720556-46720556 on Build GRCh38
rs759667800 Chr.11: 46720882-46720882 on Build GRCh38
rs557393985 Chr.11: 46720695-46720695 on Build GRCh38
rs972674937 Chr.11: 46720755-46720755 on Build GRCh38
rs542676702 Chr.11: 46720661-46720661 on Build GRCh38
rs1048863627 Chr.11: 46720787-46720787 on Build GRCh38
rs766515978 Chr.11: 46720524-46720524 on Build GRCh38
rs752835821 Chr.11: 46720790-46720790 on Build GRCh38
rs767741988 Chr.11: 46720887-46720887 on Build GRCh38
rs776504352 Chr.11: 46720749-46720749 on Build GRCh38
rs371518691 Chr.11: 46720696-46720696 on Build GRCh38
rs761893175 Chr.11: 46720858-46720858 on Build GRCh38
rs367692335 Chr.11: 46720493-46720493 on Build GRCh38
rs202071883 Chr.11: 46720785-46720785 on Build GRCh38
rs1053412926 Chr.11: 46720633-46720633 on Build GRCh38

View Full Product Details