Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47161401-47161894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf49
Entrez Gene ID
79096
Gene Name
chromosome 11 open reading frame 49
Gene Aliases
-
UniGene
Hs.368296
Gene Chromosome Location
Chr.11: 46936524-47164381 on Build GRCh38
Transcripts
NM_001003676.2 NM_001003677.2 NM_001003678.2 NM_001278222.1 NM_024113.4 NR_103471.1 NR_103472.1 XM_006718315.1 XM_011520364.2 XM_011520365.2 XM_017018277.1 XM_017018278.1 XM_017018279.1 XM_017018280.1 XM_017018281.1 XM_017018282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1675967 c.840G>C p.R280S Chr.11: 47161464-47161464 on Build GRCh38
3809298 c.997G>A p.D333N Chr.11: 47161621-47161621 on Build GRCh38
5825026 c.914C>G p.P305R Chr.11: 47161538-47161538 on Build GRCh38
4801382 c.876A>T p.T292T Chr.11: 47161500-47161500 on Build GRCh38
1354026 c.889G>A p.A297T Chr.11: 47161513-47161513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891970796 Chr.11: 47161810-47161810 on Build GRCh38
rs975120487 Chr.11: 47161596-47161596 on Build GRCh38
rs768849403 Chr.11: 47161712-47161713 on Build GRCh38
rs752557162 Chr.11: 47161551-47161551 on Build GRCh38
rs767466212 Chr.11: 47161549-47161549 on Build GRCh38
rs764319510 Chr.11: 47161455-47161455 on Build GRCh38
rs368733444 Chr.11: 47161422-47161422 on Build GRCh38
rs895532194 Chr.11: 47161824-47161824 on Build GRCh38
rs766427786 Chr.11: 47161433-47161433 on Build GRCh38
rs112390825 Chr.11: 47161494-47161494 on Build GRCh38
rs746163551 Chr.11: 47161678-47161697 on Build GRCh38
rs745990435 Chr.11: 47161478-47161478 on Build GRCh38
rs897973767 Chr.11: 47161841-47161841 on Build GRCh38
rs762568619 Chr.11: 47161531-47161531 on Build GRCh38
rs764702588 Chr.11: 47161428-47161428 on Build GRCh38
rs895605679 Chr.11: 47161530-47161530 on Build GRCh38
rs780534182 Chr.11: 47161581-47161581 on Build GRCh38
rs781372291 Chr.11: 47161611-47161611 on Build GRCh38
rs747563601 Chr.11: 47161589-47161589 on Build GRCh38
rs780639867 Chr.11: 47161636-47161636 on Build GRCh38
rs75415432 Chr.11: 47161623-47161623 on Build GRCh38
rs1029096152 Chr.11: 47161604-47161604 on Build GRCh38
rs182856342 Chr.11: 47161747-47161747 on Build GRCh38
rs962576543 Chr.11: 47161624-47161624 on Build GRCh38
rs756393714 Chr.11: 47161679-47161698 on Build GRCh38
rs909826966 Chr.11: 47161432-47161432 on Build GRCh38
rs904468726 Chr.11: 47161785-47161786 on Build GRCh38
rs371464410 Chr.11: 47161510-47161510 on Build GRCh38
rs1043086043 Chr.11: 47161453-47161453 on Build GRCh38
rs554969196 Chr.11: 47161699-47161699 on Build GRCh38
rs909712460 Chr.11: 47161709-47161709 on Build GRCh38
rs781485489 Chr.11: 47161637-47161639 on Build GRCh38
rs551875443 Chr.11: 47161501-47161501 on Build GRCh38
rs767519805 Chr.11: 47161665-47161665 on Build GRCh38
rs757335628 Chr.11: 47161468-47161468 on Build GRCh38
rs762761437 Chr.11: 47161429-47161429 on Build GRCh38
rs572027810 Chr.11: 47161742-47161742 on Build GRCh38
rs765498677 Chr.11: 47161557-47161557 on Build GRCh38
rs151295568 Chr.11: 47161543-47161543 on Build GRCh38
rs201093564 Chr.11: 47161505-47161505 on Build GRCh38
rs758530873 Chr.11: 47161579-47161579 on Build GRCh38
rs563963949 Chr.11: 47161867-47161867 on Build GRCh38
rs945931795 Chr.11: 47161768-47161768 on Build GRCh38
rs778806441 Chr.11: 47161476-47161476 on Build GRCh38
rs549441275 Chr.11: 47161451-47161451 on Build GRCh38
rs911977087 Chr.11: 47161716-47161716 on Build GRCh38
rs902068814 Chr.11: 47161834-47161834 on Build GRCh38
rs758619643 Chr.11: 47161629-47161629 on Build GRCh38
rs755503108 Chr.11: 47161590-47161590 on Build GRCh38
rs1054667121 Chr.11: 47161808-47161808 on Build GRCh38
rs187494414 Chr.11: 47161764-47161764 on Build GRCh38
rs1015479640 Chr.11: 47161839-47161839 on Build GRCh38
rs983948309 Chr.11: 47161664-47161664 on Build GRCh38
rs949714338 Chr.11: 47161479-47161479 on Build GRCh38
rs775079577 Chr.11: 47161678-47161678 on Build GRCh38
rs972629304 Chr.11: 47161739-47161739 on Build GRCh38
rs780702369 Chr.11: 47161448-47161449 on Build GRCh38
rs201892175 Chr.11: 47161493-47161493 on Build GRCh38
rs79545080 Chr.11: 47161825-47161825 on Build GRCh38
rs939981130 Chr.11: 47161715-47161715 on Build GRCh38
rs768980987 Chr.11: 47161446-47161446 on Build GRCh38
rs927090568 Chr.11: 47161779-47161779 on Build GRCh38
rs748806995 Chr.11: 47161646-47161646 on Build GRCh38
rs867503543 Chr.11: 47161619-47161619 on Build GRCh38
rs564217634 Chr.11: 47161753-47161753 on Build GRCh38
rs748552174 Chr.11: 47161620-47161620 on Build GRCh38
rs759495210 Chr.11: 47161662-47161662 on Build GRCh38
rs200083506 Chr.11: 47161425-47161425 on Build GRCh38
rs541095717 Chr.11: 47161532-47161532 on Build GRCh38
rs201411218 Chr.11: 47161447-47161447 on Build GRCh38
rs935740310 Chr.11: 47161795-47161795 on Build GRCh38
rs11551406 Chr.11: 47161522-47161522 on Build GRCh38
rs1055428566 Chr.11: 47161814-47161814 on Build GRCh38
rs966077875 Chr.11: 47161560-47161560 on Build GRCh38
rs757378371 Chr.11: 47161553-47161553 on Build GRCh38
rs3740693 Chr.11: 47161434-47161434 on Build GRCh38
rs535065566 Chr.11: 47161696-47161696 on Build GRCh38
rs34722955 Chr.11: 47161688-47161688 on Build GRCh38
rs758834419 Chr.11: 47161487-47161487 on Build GRCh38
rs1040661184 Chr.11: 47161826-47161826 on Build GRCh38
rs921976111 Chr.11: 47161421-47161421 on Build GRCh38
rs750410747 Chr.11: 47161569-47161569 on Build GRCh38
rs1044160820 Chr.11: 47161488-47161488 on Build GRCh38
rs575970019 Chr.11: 47161650-47161650 on Build GRCh38
rs757510986 Chr.11: 47161515-47161516 on Build GRCh38
rs904207767 Chr.11: 47161485-47161485 on Build GRCh38
rs949763373 Chr.11: 47161755-47161755 on Build GRCh38
rs376866242 Chr.11: 47161545-47161545 on Build GRCh38
rs780552405 Chr.11: 47161490-47161490 on Build GRCh38

View Full Product Details