Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47466242-47466748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF1
Entrez Gene ID
10658
Gene Name
CUGBP, Elav-like family member 1
Gene Aliases
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
UniGene
Hs.595333
Gene Chromosome Location
Chr.11: 47465937-47565569 on Build GRCh38
Transcripts
NM_001025596.2 NM_001172639.1 NM_001172640.1 NM_006560.3 NM_198700.2 XM_011519848.2 XM_011519850.2 XM_011519852.2 XM_011519854.1 XM_011519855.1 XM_011519856.1 XM_011519857.1 XM_011519858.1 XM_011519859.1 XM_017017101.1 XM_017017102.1 XM_017017103.1 XM_017017104.1 XM_017017105.1 XM_017017106.1 XM_017017107.1 XM_017017108.1 XM_017017109.1 XM_017017110.1 XM_017017111.1 XM_017017112.1 XM_017017113.1 XM_017017114.1 XM_017017115.1 XM_017017116.1 XM_017017117.1 XM_017017118.1 XM_017017119.1 XM_017017120.1 XM_017017121.1 XM_017017122.1 XM_017017123.1 XM_017017124.1 XM_017017125.1 XM_017017126.1 XM_017017127.1 XM_017017128.1 XM_017017129.1 XM_017017130.1 XM_017017131.1 XM_017017132.1 XM_017017133.1 XM_017017134.1 XM_017017135.1

SNP Detail

SNP ID Chromosome Location
rs1037843737 Chr.11: 47466530-47466530 on Build GRCh38
rs906529000 Chr.11: 47466354-47466354 on Build GRCh38
rs1047797397 Chr.11: 47466624-47466624 on Build GRCh38
rs938719415 Chr.11: 47466626-47466626 on Build GRCh38
rs537472388 Chr.11: 47466702-47466702 on Build GRCh38
rs1021470255 Chr.11: 47466726-47466726 on Build GRCh38
rs568447899 Chr.11: 47466675-47466678 on Build GRCh38
rs914614689 Chr.11: 47466529-47466530 on Build GRCh38
rs1035590662 Chr.11: 47466366-47466366 on Build GRCh38
rs1004068008 Chr.11: 47466276-47466276 on Build GRCh38
rs12054 Chr.11: 47466697-47466697 on Build GRCh38
rs184537598 Chr.11: 47466608-47466608 on Build GRCh38
rs924306505 Chr.11: 47466490-47466490 on Build GRCh38
rs571834471 Chr.11: 47466674-47466674 on Build GRCh38
rs932989816 Chr.11: 47466505-47466505 on Build GRCh38
rs796168333 Chr.11: 47466266-47466266 on Build GRCh38
rs953789312 Chr.11: 47466377-47466377 on Build GRCh38
rs927273767 Chr.11: 47466620-47466620 on Build GRCh38
rs1037812639 Chr.11: 47466325-47466325 on Build GRCh38
rs182103465 Chr.11: 47466344-47466344 on Build GRCh38
rs1062801 Chr.11: 47466574-47466574 on Build GRCh38
rs555328608 Chr.11: 47466562-47466563 on Build GRCh38
rs943596618 Chr.11: 47466511-47466511 on Build GRCh38
rs947371267 Chr.11: 47466576-47466576 on Build GRCh38
rs576060036 Chr.11: 47466329-47466329 on Build GRCh38
rs566215535 Chr.11: 47466374-47466374 on Build GRCh38
rs528806821 Chr.11: 47466375-47466375 on Build GRCh38
rs934927639 Chr.11: 47466411-47466411 on Build GRCh38
rs991337982 Chr.11: 47466379-47466379 on Build GRCh38
rs989052209 Chr.11: 47466418-47466418 on Build GRCh38
rs555880730 Chr.11: 47466400-47466400 on Build GRCh38
rs1012804834 Chr.11: 47466707-47466707 on Build GRCh38
rs528944422 Chr.11: 47466343-47466343 on Build GRCh38
rs987707028 Chr.11: 47466507-47466507 on Build GRCh38
rs910823538 Chr.11: 47466510-47466510 on Build GRCh38
rs968880080 Chr.11: 47466409-47466409 on Build GRCh38
rs899294276 Chr.11: 47466619-47466619 on Build GRCh38
rs55702363 Chr.11: 47466563-47466564 on Build GRCh38
rs896411962 Chr.11: 47466275-47466276 on Build GRCh38
rs1003525625 Chr.11: 47466687-47466687 on Build GRCh38
rs929422025 Chr.11: 47466623-47466623 on Build GRCh38
rs397790422 Chr.11: 47466573-47466574 on Build GRCh38
rs977158845 Chr.11: 47466435-47466435 on Build GRCh38
rs376848481 Chr.11: 47466575-47466575 on Build GRCh38
rs1062802 Chr.11: 47466562-47466562 on Build GRCh38
rs773436905 Chr.11: 47466358-47466358 on Build GRCh38

View Full Product Details