Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47469740-47470242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF1
Entrez Gene ID
10658
Gene Name
CUGBP, Elav-like family member 1
Gene Aliases
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
UniGene
Hs.595333
Gene Chromosome Location
Chr.11: 47465937-47565569 on Build GRCh38
Transcripts
NM_001025596.2 NM_001172639.1 NM_001172640.1 NM_006560.3 NM_198700.2 XM_011519848.2 XM_011519850.2 XM_011519852.2 XM_011519854.1 XM_011519855.1 XM_011519856.1 XM_011519857.1 XM_011519858.1 XM_011519859.1 XM_017017101.1 XM_017017102.1 XM_017017103.1 XM_017017104.1 XM_017017105.1 XM_017017106.1 XM_017017107.1 XM_017017108.1 XM_017017109.1 XM_017017110.1 XM_017017111.1 XM_017017112.1 XM_017017113.1 XM_017017114.1 XM_017017115.1 XM_017017116.1 XM_017017117.1 XM_017017118.1 XM_017017119.1 XM_017017120.1 XM_017017121.1 XM_017017122.1 XM_017017123.1 XM_017017124.1 XM_017017125.1 XM_017017126.1 XM_017017127.1 XM_017017128.1 XM_017017129.1 XM_017017130.1 XM_017017131.1 XM_017017132.1 XM_017017133.1 XM_017017134.1 XM_017017135.1

SNP Detail

SNP ID Chromosome Location
rs900201634 Chr.11: 47469933-47469933 on Build GRCh38
rs1049015419 Chr.11: 47469965-47469965 on Build GRCh38
rs112953599 Chr.11: 47469779-47469779 on Build GRCh38
rs34290720 Chr.11: 47469909-47469909 on Build GRCh38
rs1046335360 Chr.11: 47470147-47470147 on Build GRCh38
rs775673854 Chr.11: 47469880-47469880 on Build GRCh38
rs187305704 Chr.11: 47469781-47469781 on Build GRCh38
rs1002392300 Chr.11: 47469830-47469830 on Build GRCh38
rs758168220 Chr.11: 47469928-47469928 on Build GRCh38
rs561425255 Chr.11: 47469856-47469856 on Build GRCh38
rs35408275 Chr.11: 47469831-47469831 on Build GRCh38
rs936634970 Chr.11: 47469775-47469775 on Build GRCh38
rs1030607856 Chr.11: 47470006-47470006 on Build GRCh38
rs560548607 Chr.11: 47469991-47469991 on Build GRCh38
rs777591448 Chr.11: 47470200-47470201 on Build GRCh38
rs764407002 Chr.11: 47469895-47469895 on Build GRCh38
rs905049920 Chr.11: 47470189-47470189 on Build GRCh38
rs142467461 Chr.11: 47469816-47469816 on Build GRCh38
rs1024854727 Chr.11: 47469788-47469788 on Build GRCh38
rs887540061 Chr.11: 47469921-47469921 on Build GRCh38
rs762417673 Chr.11: 47470190-47470190 on Build GRCh38
rs997752075 Chr.11: 47469937-47469939 on Build GRCh38
rs958146256 Chr.11: 47470112-47470112 on Build GRCh38
rs544860635 Chr.11: 47469854-47469854 on Build GRCh38
rs909978841 Chr.11: 47469974-47469974 on Build GRCh38
rs1032074408 Chr.11: 47469836-47469836 on Build GRCh38
rs987838497 Chr.11: 47469889-47469889 on Build GRCh38
rs958004649 Chr.11: 47469875-47469875 on Build GRCh38
rs575078194 Chr.11: 47469920-47469920 on Build GRCh38
rs916573450 Chr.11: 47470161-47470161 on Build GRCh38
rs993308662 Chr.11: 47469899-47469901 on Build GRCh38
rs1037038861 Chr.11: 47470072-47470072 on Build GRCh38
rs540541264 Chr.11: 47469978-47469978 on Build GRCh38
rs907026128 Chr.11: 47469898-47469900 on Build GRCh38
rs948632574 Chr.11: 47469821-47469821 on Build GRCh38
rs966841701 Chr.11: 47470015-47470015 on Build GRCh38
rs761091099 Chr.11: 47469884-47469884 on Build GRCh38
rs1032801936 Chr.11: 47470063-47470063 on Build GRCh38
rs895820013 Chr.11: 47470110-47470110 on Build GRCh38
rs765599285 Chr.11: 47470206-47470206 on Build GRCh38
rs558712185 Chr.11: 47469809-47469809 on Build GRCh38
rs776816543 Chr.11: 47469914-47469914 on Build GRCh38
rs978265834 Chr.11: 47470018-47470018 on Build GRCh38
rs532437123 Chr.11: 47470117-47470117 on Build GRCh38
rs921460446 Chr.11: 47469924-47469924 on Build GRCh38

View Full Product Details