Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57793924-57794430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNND1
Entrez Gene ID
1500
Gene Name
catenin delta 1
Gene Aliases
CAS, CTNND, P120CAS, P120CTN, p120, p120(CAS), p120(CTN)
UniGene
Hs.166011
Gene Chromosome Location
Chr.11: 57761762-57819180 on Build GRCh38
Transcripts
NM_001085458.1 NM_001085459.1 NM_001085460.1 NM_001085461.1 NM_001085462.1 NM_001085463.1 NM_001085464.1 NM_001085465.1 NM_001085466.1 NM_001085467.1 NM_001085468.1 NM_001085469.1 NM_001206883.1 NM_001206884.1 NM_001206885.1 NM_001206886.1 NM_001206887.1 NM_001206888.1 NM_001206889.1 NM_001206890.1 NM_001206891.1 NM_001331.2 NR_037646.1

Gene Symbol
TMX2-CTNND1
Entrez Gene ID
100528016
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.166011
Gene Chromosome Location
Chr.11: 57712523-57819180 on Build GRCh38
Transcripts
NM_001085458.1 NM_001085459.1 NM_001085460.1 NM_001085461.1 NM_001085462.1 NM_001085463.1 NM_001085464.1 NM_001085465.1 NM_001085466.1 NM_001085467.1 NM_001085468.1 NM_001085469.1 NM_001206883.1 NM_001206884.1 NM_001206885.1 NM_001206886.1 NM_001206887.1 NM_001206888.1 NM_001206889.1 NM_001206890.1 NM_001206891.1 NM_001331.2 NR_037646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1585820 c.203G>A p.R68Q Chr.11: 57794017-57794017 on Build GRCh38
6637626 c.267+2T>G p.? Chr.11: 57794083-57794083 on Build GRCh38
1585819 c.234G>T p.Q78H Chr.11: 57794048-57794048 on Build GRCh38
7425773 c.242C>T p.S81L Chr.11: 57794056-57794056 on Build GRCh38
24302 c.255C>T p.L85L Chr.11: 57794069-57794069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755730641 Chr.11: 57794124-57794124 on Build GRCh38
rs186648416 Chr.11: 57793989-57793989 on Build GRCh38
rs769344623 Chr.11: 57794280-57794280 on Build GRCh38
rs750337023 Chr.11: 57793971-57793971 on Build GRCh38
rs920179547 Chr.11: 57794013-57794013 on Build GRCh38
rs566085193 Chr.11: 57794012-57794012 on Build GRCh38
rs746192769 Chr.11: 57794127-57794127 on Build GRCh38
rs76817459 Chr.11: 57794071-57794071 on Build GRCh38
rs758274864 Chr.11: 57793975-57793975 on Build GRCh38
rs764955659 Chr.11: 57794083-57794083 on Build GRCh38
rs781653980 Chr.11: 57794155-57794155 on Build GRCh38
rs751511942 Chr.11: 57793963-57793963 on Build GRCh38
rs780684575 Chr.11: 57794009-57794009 on Build GRCh38
rs749685225 Chr.11: 57794132-57794138 on Build GRCh38
rs752297161 Chr.11: 57794101-57794101 on Build GRCh38
rs565495946 Chr.11: 57794039-57794039 on Build GRCh38
rs1018400431 Chr.11: 57794185-57794185 on Build GRCh38
rs554388200 Chr.11: 57794145-57794145 on Build GRCh38
rs879272002 Chr.11: 57794339-57794339 on Build GRCh38
rs528038934 Chr.11: 57794313-57794313 on Build GRCh38
rs374173249 Chr.11: 57794056-57794056 on Build GRCh38
rs908738086 Chr.11: 57794356-57794356 on Build GRCh38
rs779164596 Chr.11: 57794125-57794125 on Build GRCh38
rs998955981 Chr.11: 57794133-57794133 on Build GRCh38
rs973048994 Chr.11: 57794249-57794249 on Build GRCh38
rs754470095 Chr.11: 57794096-57794096 on Build GRCh38
rs751117746 Chr.11: 57794094-57794094 on Build GRCh38
rs779564095 Chr.11: 57793984-57793984 on Build GRCh38
rs1032660034 Chr.11: 57794135-57794135 on Build GRCh38
rs776716629 Chr.11: 57794073-57794073 on Build GRCh38
rs949833578 Chr.11: 57794344-57794344 on Build GRCh38
rs553795773 Chr.11: 57794027-57794027 on Build GRCh38
rs952981552 Chr.11: 57794295-57794295 on Build GRCh38
rs370894288 Chr.11: 57794090-57794090 on Build GRCh38
rs545644609 Chr.11: 57794281-57794281 on Build GRCh38
rs941447675 Chr.11: 57794299-57794299 on Build GRCh38
rs1008677246 Chr.11: 57794208-57794208 on Build GRCh38
rs762032009 Chr.11: 57794081-57794081 on Build GRCh38
rs375122285 Chr.11: 57794382-57794382 on Build GRCh38
rs191558740 Chr.11: 57794309-57794309 on Build GRCh38
rs746166291 Chr.11: 57794290-57794290 on Build GRCh38
rs577976135 Chr.11: 57794241-57794241 on Build GRCh38
rs867927890 Chr.11: 57794336-57794336 on Build GRCh38
rs941453865 Chr.11: 57793997-57793997 on Build GRCh38
rs926698328 Chr.11: 57794399-57794399 on Build GRCh38
rs922887625 Chr.11: 57794223-57794223 on Build GRCh38
rs201927451 Chr.11: 57794016-57794016 on Build GRCh38
rs779266022 Chr.11: 57794017-57794017 on Build GRCh38
rs549738133 Chr.11: 57793966-57793966 on Build GRCh38
rs1009870050 Chr.11: 57794141-57794141 on Build GRCh38
rs543236907 Chr.11: 57794338-57794338 on Build GRCh38
rs964158017 Chr.11: 57794202-57794202 on Build GRCh38
rs376551286 Chr.11: 57794242-57794242 on Build GRCh38
rs752176301 Chr.11: 57794093-57794093 on Build GRCh38
rs757805272 Chr.11: 57794148-57794148 on Build GRCh38
rs573787383 Chr.11: 57794177-57794177 on Build GRCh38
rs754662949 Chr.11: 57793991-57793991 on Build GRCh38
rs761502959 Chr.11: 57793963-57793963 on Build GRCh38
rs780381774 Chr.11: 57794005-57794006 on Build GRCh38
rs373862115 Chr.11: 57794099-57794099 on Build GRCh38
rs983229955 Chr.11: 57794246-57794246 on Build GRCh38
rs976985853 Chr.11: 57794215-57794215 on Build GRCh38
rs149449203 Chr.11: 57794237-57794237 on Build GRCh38
rs544412419 Chr.11: 57794220-57794220 on Build GRCh38
rs750185141 Chr.11: 57794085-57794085 on Build GRCh38
rs371264068 Chr.11: 57794028-57794028 on Build GRCh38

View Full Product Details