Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57804345-57804878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNND1
Entrez Gene ID
1500
Gene Name
catenin delta 1
Gene Aliases
CAS, CTNND, P120CAS, P120CTN, p120, p120(CAS), p120(CTN)
UniGene
Hs.166011
Gene Chromosome Location
Chr.11: 57761762-57819180 on Build GRCh38
Transcripts
NM_001085458.1 NM_001085459.1 NM_001085460.1 NM_001085461.1 NM_001085462.1 NM_001085463.1 NM_001085464.1 NM_001085465.1 NM_001085466.1 NM_001085467.1 NM_001085468.1 NM_001085469.1 NM_001206883.1 NM_001206884.1 NM_001206885.1 NM_001206886.1 NM_001206887.1 NM_001206888.1 NM_001206889.1 NM_001206890.1 NM_001206891.1 NM_001331.2 NR_037646.1

Gene Symbol
TMX2-CTNND1
Entrez Gene ID
100528016
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.166011
Gene Chromosome Location
Chr.11: 57712523-57819180 on Build GRCh38
Transcripts
NM_001085458.1 NM_001085459.1 NM_001085460.1 NM_001085461.1 NM_001085462.1 NM_001085463.1 NM_001085464.1 NM_001085465.1 NM_001085466.1 NM_001085467.1 NM_001085468.1 NM_001085469.1 NM_001206883.1 NM_001206884.1 NM_001206885.1 NM_001206886.1 NM_001206887.1 NM_001206888.1 NM_001206889.1 NM_001206890.1 NM_001206891.1 NM_001331.2 NR_037646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7180188 c.1712C>T p.S571L Chr.11: 57804770-57804770 on Build GRCh38
4626851 c.1626T>A p.S542R Chr.11: 57804684-57804684 on Build GRCh38
4626852 c.1626T>A p.S542R Chr.11: 57804684-57804684 on Build GRCh38
6069502 c.1652G>A p.C551Y Chr.11: 57804710-57804710 on Build GRCh38
928716 c.1640_1641insACTT p.R549fs*5 Chr.11: 57804698-57804699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776366066 Chr.11: 57804742-57804742 on Build GRCh38
rs559443966 Chr.11: 57804512-57804512 on Build GRCh38
rs900311833 Chr.11: 57804498-57804498 on Build GRCh38
rs1014592673 Chr.11: 57804598-57804598 on Build GRCh38
rs138371560 Chr.11: 57804399-57804399 on Build GRCh38
rs185366485 Chr.11: 57804463-57804463 on Build GRCh38
rs370211216 Chr.11: 57804694-57804694 on Build GRCh38
rs933863439 Chr.11: 57804692-57804692 on Build GRCh38
rs1038380278 Chr.11: 57804755-57804755 on Build GRCh38
rs746969374 Chr.11: 57804673-57804673 on Build GRCh38
rs890360235 Chr.11: 57804579-57804579 on Build GRCh38
rs1021226985 Chr.11: 57804520-57804520 on Build GRCh38
rs776819881 Chr.11: 57804774-57804774 on Build GRCh38
rs60769290 Chr.11: 57804810-57804810 on Build GRCh38
rs913351488 Chr.11: 57804467-57804467 on Build GRCh38
rs759489592 Chr.11: 57804823-57804824 on Build GRCh38
rs779939095 Chr.11: 57804629-57804629 on Build GRCh38
rs530107161 Chr.11: 57804536-57804536 on Build GRCh38
rs1004539435 Chr.11: 57804384-57804384 on Build GRCh38
rs759101108 Chr.11: 57804837-57804837 on Build GRCh38
rs777282942 Chr.11: 57804704-57804704 on Build GRCh38
rs770924563 Chr.11: 57804799-57804800 on Build GRCh38
rs200199564 Chr.11: 57804733-57804733 on Build GRCh38
rs763219126 Chr.11: 57804830-57804830 on Build GRCh38
rs563598014 Chr.11: 57804377-57804377 on Build GRCh38
rs11570200 Chr.11: 57804502-57804502 on Build GRCh38
rs769540092 Chr.11: 57804703-57804703 on Build GRCh38
rs898534201 Chr.11: 57804790-57804790 on Build GRCh38
rs775782206 Chr.11: 57804730-57804730 on Build GRCh38
rs920065785 Chr.11: 57804409-57804409 on Build GRCh38
rs149813159 Chr.11: 57804421-57804421 on Build GRCh38
rs974653911 Chr.11: 57804408-57804408 on Build GRCh38
rs1014706891 Chr.11: 57804397-57804397 on Build GRCh38
rs564303093 Chr.11: 57804454-57804454 on Build GRCh38
rs966988179 Chr.11: 57804526-57804526 on Build GRCh38
rs1034258223 Chr.11: 57804475-57804475 on Build GRCh38
rs769120408 Chr.11: 57804772-57804772 on Build GRCh38
rs947442692 Chr.11: 57804489-57804489 on Build GRCh38
rs10896647 Chr.11: 57804411-57804411 on Build GRCh38
rs1027580511 Chr.11: 57804681-57804681 on Build GRCh38
rs1055241455 Chr.11: 57804809-57804809 on Build GRCh38
rs748901736 Chr.11: 57804712-57804712 on Build GRCh38
rs765294247 Chr.11: 57804814-57804814 on Build GRCh38
rs938705805 Chr.11: 57804455-57804455 on Build GRCh38
rs1039946269 Chr.11: 57804390-57804390 on Build GRCh38
rs928865254 Chr.11: 57804435-57804435 on Build GRCh38
rs921146808 Chr.11: 57804394-57804394 on Build GRCh38
rs201092213 Chr.11: 57804666-57804666 on Build GRCh38
rs750626715 Chr.11: 57804823-57804823 on Build GRCh38
rs189735518 Chr.11: 57804591-57804591 on Build GRCh38
rs774877820 Chr.11: 57804456-57804456 on Build GRCh38
rs776485109 Chr.11: 57804822-57804824 on Build GRCh38
rs761795416 Chr.11: 57804808-57804808 on Build GRCh38
rs898586705 Chr.11: 57804452-57804452 on Build GRCh38
rs1038433391 Chr.11: 57804449-57804449 on Build GRCh38
rs896625472 Chr.11: 57804816-57804819 on Build GRCh38
rs547930024 Chr.11: 57804495-57804495 on Build GRCh38
rs988781333 Chr.11: 57804853-57804853 on Build GRCh38
rs145871725 Chr.11: 57804592-57804592 on Build GRCh38
rs779991605 Chr.11: 57804398-57804398 on Build GRCh38
rs747592582 Chr.11: 57804699-57804699 on Build GRCh38
rs552330982 Chr.11: 57804695-57804695 on Build GRCh38
rs772435976 Chr.11: 57804715-57804715 on Build GRCh38
rs758273710 Chr.11: 57804623-57804623 on Build GRCh38
rs748484315 Chr.11: 57804613-57804613 on Build GRCh38
rs149004167 Chr.11: 57804604-57804604 on Build GRCh38
rs1004232532 Chr.11: 57804430-57804430 on Build GRCh38
rs960271206 Chr.11: 57804848-57804848 on Build GRCh38
rs1053043799 Chr.11: 57804464-57804464 on Build GRCh38

View Full Product Details