Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 58731881-58732378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLYAT
Entrez Gene ID
10249
Gene Name
glycine-N-acyltransferase
Gene Aliases
ACGNAT, CAT, GAT
UniGene
Hs.145384
Gene Chromosome Location
Chr.11: 58708757-58731974 on Build GRCh38
Transcripts
NM_005838.3 NM_201648.2

SNP Detail

SNP ID Chromosome Location
rs59882006 Chr.11: 58732182-58732182 on Build GRCh38
rs548305290 Chr.11: 58732007-58732007 on Build GRCh38
rs915657046 Chr.11: 58732149-58732149 on Build GRCh38
rs1049744144 Chr.11: 58732221-58732221 on Build GRCh38
rs547486315 Chr.11: 58731928-58731928 on Build GRCh38
rs982231019 Chr.11: 58732114-58732114 on Build GRCh38
rs143454263 Chr.11: 58732240-58732240 on Build GRCh38
rs920518221 Chr.11: 58732089-58732089 on Build GRCh38
rs914253079 Chr.11: 58732161-58732161 on Build GRCh38
rs951652350 Chr.11: 58732092-58732093 on Build GRCh38
rs949858573 Chr.11: 58732154-58732154 on Build GRCh38
rs1051578378 Chr.11: 58732266-58732266 on Build GRCh38
rs536868495 Chr.11: 58732056-58732056 on Build GRCh38
rs183913343 Chr.11: 58732301-58732301 on Build GRCh38
rs565778896 Chr.11: 58731937-58731937 on Build GRCh38
rs900222183 Chr.11: 58732239-58732239 on Build GRCh38
rs996792923 Chr.11: 58732209-58732209 on Build GRCh38
rs886343227 Chr.11: 58732003-58732003 on Build GRCh38
rs886395629 Chr.11: 58732276-58732276 on Build GRCh38
rs955488587 Chr.11: 58732030-58732030 on Build GRCh38
rs769369306 Chr.11: 58732198-58732198 on Build GRCh38
rs149207605 Chr.11: 58732099-58732099 on Build GRCh38
rs770907048 Chr.11: 58731985-58731985 on Build GRCh38
rs1009131089 Chr.11: 58732041-58732041 on Build GRCh38
rs75425382 Chr.11: 58732234-58732234 on Build GRCh38
rs1039162617 Chr.11: 58731933-58731933 on Build GRCh38
rs1016149199 Chr.11: 58732290-58732290 on Build GRCh38
rs1054603772 Chr.11: 58732200-58732200 on Build GRCh38
rs993738819 Chr.11: 58732250-58732250 on Build GRCh38
rs991193819 Chr.11: 58732100-58732100 on Build GRCh38
rs746779327 Chr.11: 58731936-58731936 on Build GRCh38
rs187444063 Chr.11: 58732066-58732066 on Build GRCh38
rs934839974 Chr.11: 58732185-58732185 on Build GRCh38
rs530133532 Chr.11: 58731942-58731942 on Build GRCh38
rs970931322 Chr.11: 58732096-58732096 on Build GRCh38
rs57742826 Chr.11: 58732060-58732060 on Build GRCh38
rs191674543 Chr.11: 58732281-58732281 on Build GRCh38
rs188800887 Chr.11: 58732310-58732310 on Build GRCh38
rs1027569982 Chr.11: 58732280-58732280 on Build GRCh38
rs963674043 Chr.11: 58732085-58732085 on Build GRCh38
rs971490852 Chr.11: 58732345-58732345 on Build GRCh38
rs1024115260 Chr.11: 58732092-58732092 on Build GRCh38
rs999143232 Chr.11: 58731930-58731930 on Build GRCh38
rs997737130 Chr.11: 58731957-58731957 on Build GRCh38
rs981186711 Chr.11: 58732347-58732347 on Build GRCh38
rs892924880 Chr.11: 58731910-58731910 on Build GRCh38
rs1030267503 Chr.11: 58731960-58731960 on Build GRCh38
rs570662993 Chr.11: 58732061-58732061 on Build GRCh38
rs990921890 Chr.11: 58732087-58732087 on Build GRCh38
rs569500228 Chr.11: 58732034-58732034 on Build GRCh38
rs988472177 Chr.11: 58732156-58732156 on Build GRCh38
rs961871319 Chr.11: 58732245-58732245 on Build GRCh38
rs1046301381 Chr.11: 58732176-58732176 on Build GRCh38
rs935580586 Chr.11: 58732140-58732140 on Build GRCh38
rs924216851 Chr.11: 58732138-58732138 on Build GRCh38
rs915604584 Chr.11: 58732291-58732291 on Build GRCh38
rs893394967 Chr.11: 58732206-58732206 on Build GRCh38

View Full Product Details