Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 58834704-58835216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLYATL2
Entrez Gene ID
219970
Gene Name
glycine-N-acyltransferase like 2
Gene Aliases
BXMAS2-10, GATF-B
UniGene
Hs.254271
Gene Chromosome Location
Chr.11: 58834065-58905699 on Build GRCh38
Transcripts
NM_145016.3 XM_011544809.2 XM_017017337.1 XM_017017338.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7227117 c.574A>T p.I192F Chr.11: 58834740-58834740 on Build GRCh38
6014431 c.491C>T p.S164L Chr.11: 58834823-58834823 on Build GRCh38
4034214 c.586C>A p.L196I Chr.11: 58834728-58834728 on Build GRCh38
6519068 c.539G>T p.W180L Chr.11: 58834775-58834775 on Build GRCh38
3450066 c.482G>A p.G161E Chr.11: 58834832-58834832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375216857 Chr.11: 58835045-58835045 on Build GRCh38
rs755639436 Chr.11: 58834764-58834764 on Build GRCh38
rs970285711 Chr.11: 58835181-58835181 on Build GRCh38
rs746378135 Chr.11: 58834781-58834781 on Build GRCh38
rs928871876 Chr.11: 58834929-58834929 on Build GRCh38
rs200152129 Chr.11: 58834728-58834728 on Build GRCh38
rs535626394 Chr.11: 58834946-58834946 on Build GRCh38
rs777246922 Chr.11: 58834923-58834923 on Build GRCh38
rs774402769 Chr.11: 58834854-58834854 on Build GRCh38
rs752307700 Chr.11: 58834866-58834866 on Build GRCh38
rs997218790 Chr.11: 58835109-58835109 on Build GRCh38
rs371997267 Chr.11: 58835033-58835033 on Build GRCh38
rs552108196 Chr.11: 58834805-58834805 on Build GRCh38
rs111235124 Chr.11: 58834925-58834925 on Build GRCh38
rs73475770 Chr.11: 58834985-58834985 on Build GRCh38
rs879765076 Chr.11: 58834835-58834836 on Build GRCh38
rs980672983 Chr.11: 58835190-58835190 on Build GRCh38
rs755584294 Chr.11: 58834870-58834870 on Build GRCh38
rs111680745 Chr.11: 58835026-58835026 on Build GRCh38
rs75096485 Chr.11: 58834845-58834845 on Build GRCh38
rs748845012 Chr.11: 58834883-58834883 on Build GRCh38
rs375653649 Chr.11: 58834780-58834780 on Build GRCh38
rs11229651 Chr.11: 58834836-58834836 on Build GRCh38
rs61729332 Chr.11: 58834747-58834747 on Build GRCh38
rs745522716 Chr.11: 58834734-58834734 on Build GRCh38
rs17851433 Chr.11: 58834812-58834812 on Build GRCh38
rs903657271 Chr.11: 58835022-58835023 on Build GRCh38
rs980345335 Chr.11: 58834927-58834927 on Build GRCh38
rs901738216 Chr.11: 58835061-58835061 on Build GRCh38
rs777296998 Chr.11: 58834879-58834879 on Build GRCh38
rs767186891 Chr.11: 58834861-58834861 on Build GRCh38
rs374442344 Chr.11: 58834817-58834817 on Build GRCh38
rs1043584368 Chr.11: 58834988-58834988 on Build GRCh38
rs775732518 Chr.11: 58834972-58834972 on Build GRCh38
rs868520769 Chr.11: 58834832-58834832 on Build GRCh38
rs776773653 Chr.11: 58834808-58834808 on Build GRCh38
rs780751650 Chr.11: 58834742-58834742 on Build GRCh38
rs529021206 Chr.11: 58835165-58835165 on Build GRCh38
rs377489659 Chr.11: 58834733-58834733 on Build GRCh38
rs192840791 Chr.11: 58834960-58834960 on Build GRCh38
rs768793380 Chr.11: 58834778-58834779 on Build GRCh38
rs766305160 Chr.11: 58834730-58834730 on Build GRCh38
rs184824113 Chr.11: 58835121-58835121 on Build GRCh38
rs750599154 Chr.11: 58835138-58835138 on Build GRCh38
rs199566288 Chr.11: 58834788-58834788 on Build GRCh38
rs997588515 Chr.11: 58834801-58834801 on Build GRCh38
rs775789275 Chr.11: 58834797-58834797 on Build GRCh38
rs758871353 Chr.11: 58834884-58834884 on Build GRCh38
rs546127495 Chr.11: 58835020-58835020 on Build GRCh38
rs202103606 Chr.11: 58834851-58834851 on Build GRCh38
rs1043551877 Chr.11: 58834745-58834745 on Build GRCh38
rs765232447 Chr.11: 58834825-58834825 on Build GRCh38
rs1007437776 Chr.11: 58834760-58834760 on Build GRCh38
rs187660800 Chr.11: 58834799-58834799 on Build GRCh38
rs747413442 Chr.11: 58834800-58834800 on Build GRCh38
rs754800057 Chr.11: 58834852-58834852 on Build GRCh38
rs938720895 Chr.11: 58834937-58834937 on Build GRCh38
rs561806252 Chr.11: 58835142-58835142 on Build GRCh38

View Full Product Details