Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60850810-60851321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC86
Entrez Gene ID
79080
Gene Name
coiled-coil domain containing 86
Gene Aliases
-
UniGene
Hs.4253
Gene Chromosome Location
Chr.11: 60841956-60851088 on Build GRCh38
Transcripts
NM_024098.3 NM_004778.2

Gene Symbol
PTGDR2
Entrez Gene ID
11251
Gene Name
prostaglandin D2 receptor 2
Gene Aliases
CD294, CRTH2, DL1R, DP2, GPR44
UniGene
Hs.299567
Gene Chromosome Location
Chr.11: 60850925-60855971 on Build GRCh38
Transcripts
NM_024098.3 NM_004778.2

SNP Detail

SNP ID Chromosome Location
rs931369184 Chr.11: 60851046-60851047 on Build GRCh38
rs977815369 Chr.11: 60850958-60850958 on Build GRCh38
rs989230424 Chr.11: 60851168-60851168 on Build GRCh38
rs984569594 Chr.11: 60851134-60851134 on Build GRCh38
rs548263339 Chr.11: 60851093-60851093 on Build GRCh38
rs1012154601 Chr.11: 60850882-60850882 on Build GRCh38
rs1042374380 Chr.11: 60851248-60851248 on Build GRCh38
rs906384422 Chr.11: 60851120-60851120 on Build GRCh38
rs989526895 Chr.11: 60850985-60850985 on Build GRCh38
rs937364603 Chr.11: 60850981-60850981 on Build GRCh38
rs758029285 Chr.11: 60850922-60850923 on Build GRCh38
rs759189168 Chr.11: 60851052-60851052 on Build GRCh38
rs895807265 Chr.11: 60851207-60851207 on Build GRCh38
rs559280561 Chr.11: 60851023-60851023 on Build GRCh38
rs148839456 Chr.11: 60850994-60850994 on Build GRCh38
rs11824571 Chr.11: 60851088-60851088 on Build GRCh38
rs1032786430 Chr.11: 60851041-60851041 on Build GRCh38
rs138444219 Chr.11: 60850979-60850979 on Build GRCh38
rs575794040 Chr.11: 60850977-60850977 on Build GRCh38
rs903367311 Chr.11: 60851034-60851034 on Build GRCh38
rs550907114 Chr.11: 60851261-60851261 on Build GRCh38
rs919089640 Chr.11: 60850945-60850945 on Build GRCh38
rs1055848672 Chr.11: 60851193-60851193 on Build GRCh38
rs930456673 Chr.11: 60850964-60850964 on Build GRCh38
rs1000406199 Chr.11: 60851281-60851281 on Build GRCh38
rs937416798 Chr.11: 60851143-60851143 on Build GRCh38
rs752354396 Chr.11: 60851142-60851142 on Build GRCh38
rs895080715 Chr.11: 60850836-60850836 on Build GRCh38
rs545365765 Chr.11: 60851021-60851021 on Build GRCh38
rs930505724 Chr.11: 60851128-60851128 on Build GRCh38
rs184623748 Chr.11: 60851006-60851006 on Build GRCh38
rs903418840 Chr.11: 60851271-60851271 on Build GRCh38
rs188516745 Chr.11: 60851202-60851202 on Build GRCh38
rs1019036403 Chr.11: 60851055-60851055 on Build GRCh38
rs1025263437 Chr.11: 60850897-60850897 on Build GRCh38
rs932924232 Chr.11: 60850995-60850995 on Build GRCh38
rs571853221 Chr.11: 60851005-60851005 on Build GRCh38
rs369969136 Chr.11: 60851086-60851086 on Build GRCh38
rs753683564 Chr.11: 60851035-60851035 on Build GRCh38
rs971974673 Chr.11: 60850886-60850886 on Build GRCh38
rs372611424 Chr.11: 60850957-60850957 on Build GRCh38
rs760728112 Chr.11: 60850927-60850927 on Build GRCh38
rs150468620 Chr.11: 60850854-60850854 on Build GRCh38
rs919154551 Chr.11: 60851094-60851094 on Build GRCh38
rs912773108 Chr.11: 60851010-60851010 on Build GRCh38
rs11230506 Chr.11: 60851210-60851210 on Build GRCh38
rs371857294 Chr.11: 60851066-60851069 on Build GRCh38
rs957807541 Chr.11: 60851154-60851154 on Build GRCh38
rs775715987 Chr.11: 60850895-60850895 on Build GRCh38
rs766370934 Chr.11: 60850993-60850993 on Build GRCh38
rs970532659 Chr.11: 60850954-60850955 on Build GRCh38
rs944147374 Chr.11: 60851014-60851014 on Build GRCh38
rs953106526 Chr.11: 60851051-60851051 on Build GRCh38

View Full Product Details