Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61006382-61006899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD6
Entrez Gene ID
923
Gene Name
CD6 molecule
Gene Aliases
TP120
UniGene
Hs.744366
Gene Chromosome Location
Chr.11: 60971641-61020377 on Build GRCh38
Transcripts
NM_001254750.1 NM_001254751.1 NM_006725.4 NR_045638.1 XM_006718738.1 XM_006718739.1 XM_006718740.1 XM_006718741.1 XM_011545360.1 XM_011545362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
239229 c.118G>C p.G40R Chr.11: 61006642-61006642 on Build GRCh38
3980477 c.66C>G p.A22A Chr.11: 61006590-61006590 on Build GRCh38
4926527 c.65C>T p.A22V Chr.11: 61006589-61006589 on Build GRCh38
6898323 c.75C>A p.D25E Chr.11: 61006599-61006599 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370230503 Chr.11: 61006575-61006575 on Build GRCh38
rs774453541 Chr.11: 61006675-61006675 on Build GRCh38
rs773476485 Chr.11: 61006580-61006580 on Build GRCh38
rs751472643 Chr.11: 61006590-61006590 on Build GRCh38
rs929703552 Chr.11: 61006622-61006622 on Build GRCh38
rs958660910 Chr.11: 61006558-61006558 on Build GRCh38
rs747552304 Chr.11: 61006640-61006640 on Build GRCh38
rs769168823 Chr.11: 61006551-61006551 on Build GRCh38
rs778441031 Chr.11: 61006639-61006639 on Build GRCh38
rs751902504 Chr.11: 61006813-61006813 on Build GRCh38
rs770946545 Chr.11: 61006671-61006671 on Build GRCh38
rs116895811 Chr.11: 61006417-61006417 on Build GRCh38
rs749477451 Chr.11: 61006667-61006667 on Build GRCh38
rs1032129242 Chr.11: 61006497-61006497 on Build GRCh38
rs535179863 Chr.11: 61006722-61006722 on Build GRCh38
rs763720830 Chr.11: 61006630-61006630 on Build GRCh38
rs558319060 Chr.11: 61006662-61006662 on Build GRCh38
rs538390770 Chr.11: 61006540-61006540 on Build GRCh38
rs780434838 Chr.11: 61006537-61006537 on Build GRCh38
rs371589414 Chr.11: 61006683-61006683 on Build GRCh38
rs1055308896 Chr.11: 61006728-61006728 on Build GRCh38
rs965672166 Chr.11: 61006847-61006847 on Build GRCh38
rs1015781495 Chr.11: 61006478-61006478 on Build GRCh38
rs977275239 Chr.11: 61006692-61006692 on Build GRCh38
rs546567498 Chr.11: 61006660-61006660 on Build GRCh38
rs762942103 Chr.11: 61006581-61006581 on Build GRCh38
rs891248943 Chr.11: 61006815-61006815 on Build GRCh38
rs868475565 Chr.11: 61006426-61006426 on Build GRCh38
rs753313477 Chr.11: 61006464-61006464 on Build GRCh38
rs767412868 Chr.11: 61006617-61006617 on Build GRCh38
rs1055807440 Chr.11: 61006422-61006422 on Build GRCh38
rs957895978 Chr.11: 61006510-61006510 on Build GRCh38
rs753467206 Chr.11: 61006631-61006631 on Build GRCh38
rs142754995 Chr.11: 61006552-61006552 on Build GRCh38
rs755587886 Chr.11: 61006642-61006642 on Build GRCh38
rs767502590 Chr.11: 61006689-61006689 on Build GRCh38
rs764530610 Chr.11: 61006778-61006778 on Build GRCh38
rs137922382 Chr.11: 61006760-61006760 on Build GRCh38
rs777461107 Chr.11: 61006530-61006530 on Build GRCh38
rs1013973866 Chr.11: 61006449-61006449 on Build GRCh38
rs942150804 Chr.11: 61006712-61006712 on Build GRCh38
rs1015730503 Chr.11: 61006867-61006867 on Build GRCh38
rs74980262 Chr.11: 61006838-61006838 on Build GRCh38
rs374467904 Chr.11: 61006805-61006805 on Build GRCh38
rs868625131 Chr.11: 61006657-61006657 on Build GRCh38
rs752539359 Chr.11: 61006620-61006620 on Build GRCh38
rs1046829890 Chr.11: 61006761-61006761 on Build GRCh38
rs747464332 Chr.11: 61006539-61006539 on Build GRCh38
rs924508318 Chr.11: 61006701-61006701 on Build GRCh38
rs932692128 Chr.11: 61006790-61006790 on Build GRCh38
rs1007107732 Chr.11: 61006825-61006825 on Build GRCh38
rs150290463 Chr.11: 61006666-61006666 on Build GRCh38
rs770195748 Chr.11: 61006576-61006576 on Build GRCh38
rs949862843 Chr.11: 61006676-61006676 on Build GRCh38
rs770471070 Chr.11: 61006536-61006536 on Build GRCh38
rs138890229 Chr.11: 61006608-61006608 on Build GRCh38
rs572874059 Chr.11: 61006495-61006495 on Build GRCh38
rs902558452 Chr.11: 61006765-61006765 on Build GRCh38
rs895351002 Chr.11: 61006731-61006731 on Build GRCh38
rs138701575 Chr.11: 61006462-61006462 on Build GRCh38
rs966727763 Chr.11: 61006463-61006463 on Build GRCh38
rs756735248 Chr.11: 61006632-61006632 on Build GRCh38
rs766424297 Chr.11: 61006589-61006589 on Build GRCh38
rs935916727 Chr.11: 61006733-61006733 on Build GRCh38
rs999554226 Chr.11: 61006792-61006792 on Build GRCh38
rs1018114510 Chr.11: 61006843-61006843 on Build GRCh38
rs755890364 Chr.11: 61006623-61006623 on Build GRCh38
rs748808609 Chr.11: 61006533-61006533 on Build GRCh38
rs748586444 Chr.11: 61006664-61006664 on Build GRCh38

View Full Product Details